Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64986367-64986965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

Gene Symbol
FNTB
Entrez Gene ID
2342
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
FPTB
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64986789-65062652 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002028.3

SNP Detail

SNP ID Chromosome Location
rs537337979 Chr.14: 64986540-64986540 on Build GRCh38
rs1008161158 Chr.14: 64986900-64986900 on Build GRCh38
rs373810742 Chr.14: 64986937-64986937 on Build GRCh38
rs187061837 Chr.14: 64986720-64986720 on Build GRCh38
rs945444575 Chr.14: 64986412-64986412 on Build GRCh38
rs925321151 Chr.14: 64986454-64986454 on Build GRCh38
rs3215788 Chr.14: 64986776-64986776 on Build GRCh38
rs188813635 Chr.14: 64986823-64986823 on Build GRCh38
rs929460282 Chr.14: 64986622-64986622 on Build GRCh38
rs949549122 Chr.14: 64986620-64986620 on Build GRCh38
rs376226292 Chr.14: 64986909-64986909 on Build GRCh38
rs991263120 Chr.14: 64986777-64986777 on Build GRCh38
rs909479211 Chr.14: 64986665-64986665 on Build GRCh38
rs771816255 Chr.14: 64986921-64986921 on Build GRCh38
rs566127810 Chr.14: 64986486-64986486 on Build GRCh38
rs150661154 Chr.14: 64986566-64986566 on Build GRCh38
rs575075587 Chr.14: 64986539-64986539 on Build GRCh38
rs539673690 Chr.14: 64986932-64986932 on Build GRCh38
rs975799835 Chr.14: 64986616-64986616 on Build GRCh38
rs1034890067 Chr.14: 64986432-64986432 on Build GRCh38
rs891023979 Chr.14: 64986895-64986895 on Build GRCh38
rs1028234579 Chr.14: 64986718-64986718 on Build GRCh38
rs201739947 Chr.14: 64986924-64986924 on Build GRCh38
rs550898876 Chr.14: 64986764-64986764 on Build GRCh38
rs192403314 Chr.14: 64986775-64986775 on Build GRCh38
rs1049423880 Chr.14: 64986875-64986875 on Build GRCh38
rs996490068 Chr.14: 64986859-64986859 on Build GRCh38
rs986664594 Chr.14: 64986393-64986393 on Build GRCh38
rs965863134 Chr.14: 64986614-64986614 on Build GRCh38
rs182339022 Chr.14: 64986671-64986671 on Build GRCh38
rs375323849 Chr.14: 64986440-64986440 on Build GRCh38
rs777986327 Chr.14: 64986888-64986888 on Build GRCh38
rs535113042 Chr.14: 64986883-64986883 on Build GRCh38
rs1008024004 Chr.14: 64986723-64986723 on Build GRCh38
rs568122612 Chr.14: 64986899-64986899 on Build GRCh38
rs566067433 Chr.14: 64986827-64986827 on Build GRCh38
rs886821673 Chr.14: 64986692-64986692 on Build GRCh38
rs183356460 Chr.14: 64986779-64986779 on Build GRCh38
rs757093033 Chr.14: 64986933-64986936 on Build GRCh38
rs377745402 Chr.14: 64986715-64986715 on Build GRCh38
rs909334053 Chr.14: 64986868-64986868 on Build GRCh38
rs1003985996 Chr.14: 64986693-64986693 on Build GRCh38
rs570672073 Chr.14: 64986409-64986409 on Build GRCh38
rs896783477 Chr.14: 64986607-64986607 on Build GRCh38
rs370439296 Chr.14: 64986910-64986910 on Build GRCh38
rs774166152 Chr.14: 64986915-64986915 on Build GRCh38
rs959399375 Chr.14: 64986776-64986776 on Build GRCh38
rs979527940 Chr.14: 64986768-64986768 on Build GRCh38
rs769681975 Chr.14: 64986904-64986904 on Build GRCh38
rs144711621 Chr.14: 64986897-64986897 on Build GRCh38
rs918135918 Chr.14: 64986778-64986778 on Build GRCh38
rs572873700 Chr.14: 64986716-64986716 on Build GRCh38
rs1003329577 Chr.14: 64986431-64986431 on Build GRCh38
rs577025329 Chr.14: 64986574-64986574 on Build GRCh38
rs189672347 Chr.14: 64986506-64986506 on Build GRCh38
rs1020817121 Chr.14: 64986734-64986734 on Build GRCh38
rs1020911986 Chr.14: 64986902-64986902 on Build GRCh38
rs372708444 Chr.14: 64986930-64986930 on Build GRCh38
rs1038118078 Chr.14: 64986812-64986812 on Build GRCh38
rs139930341 Chr.14: 64986738-64986738 on Build GRCh38
rs984900783 Chr.14: 64986642-64986642 on Build GRCh38
rs770722527 Chr.14: 64986914-64986914 on Build GRCh38
rs781290528 Chr.14: 64986815-64986815 on Build GRCh38
rs975829958 Chr.14: 64986814-64986814 on Build GRCh38
rs879319639 Chr.14: 64986822-64986822 on Build GRCh38
rs759307173 Chr.14: 64986917-64986917 on Build GRCh38
rs980752878 Chr.14: 64986889-64986889 on Build GRCh38
rs577107021 Chr.14: 64986926-64986926 on Build GRCh38
rs1044762595 Chr.14: 64986410-64986410 on Build GRCh38
rs545959265 Chr.14: 64986584-64986584 on Build GRCh38
rs1022201145 Chr.14: 64986520-64986520 on Build GRCh38
rs892295305 Chr.14: 64986496-64986496 on Build GRCh38
rs1017652499 Chr.14: 64986586-64986586 on Build GRCh38
rs766780991 Chr.14: 64986935-64986935 on Build GRCh38
rs1038417675 Chr.14: 64986707-64986707 on Build GRCh38
rs942223745 Chr.14: 64986811-64986811 on Build GRCh38
rs765673443 Chr.14: 64986929-64986929 on Build GRCh38
rs755260685 Chr.14: 64986938-64986938 on Build GRCh38
rs534739967 Chr.14: 64986488-64986488 on Build GRCh38
rs898140676 Chr.14: 64986713-64986713 on Build GRCh38
rs558807312 Chr.14: 64986455-64986455 on Build GRCh38
rs747955254 Chr.14: 64986901-64986901 on Build GRCh38
rs763250558 Chr.14: 64986933-64986933 on Build GRCh38
rs751643582 Chr.14: 64986444-64986444 on Build GRCh38
rs908065251 Chr.14: 64986795-64986795 on Build GRCh38
rs1044404868 Chr.14: 64986681-64986681 on Build GRCh38
rs1032420395 Chr.14: 64986925-64986925 on Build GRCh38
rs979817987 Chr.14: 64986433-64986433 on Build GRCh38
rs553529626 Chr.14: 64986941-64986941 on Build GRCh38
rs530479347 Chr.14: 64986763-64986763 on Build GRCh38
rs775206745 Chr.14: 64986923-64986923 on Build GRCh38
rs1047863433 Chr.14: 64986783-64986783 on Build GRCh38
rs907293739 Chr.14: 64986427-64986427 on Build GRCh38

View Full Product Details