Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74904531-74905129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM219B
Entrez Gene ID
57184
Gene Name
family with sequence similarity 219 member B
Gene Aliases
C15orf17
UniGene
Hs.367690
Gene Chromosome Location
Chr.15: 74898763-74907228 on Build GRCh38
Transcripts
NM_001321920.1 NM_001321921.1 NM_001321922.1 NM_001321923.1 NM_020447.4 NR_135841.1 NR_135842.1 NR_135843.1 NR_135844.1 NR_135845.1 NR_135846.1 NR_135847.1 NR_135848.1 NR_135849.1 NR_135850.1 NR_135851.1 NR_135852.1 NR_135853.1 NR_135854.1 XM_017022433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7180675 c.401G>A p.R134K Chr.15: 74904692-74904692 on Build GRCh38
223799 c.385G>A p.G129R Chr.15: 74904708-74904708 on Build GRCh38
6324262 c.424G>T p.A142S Chr.15: 74904669-74904669 on Build GRCh38
6464075 c.398C>G p.S133C Chr.15: 74904695-74904695 on Build GRCh38
6993397 c.411C>T p.S137S Chr.15: 74904682-74904682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557116291 Chr.15: 74904561-74904561 on Build GRCh38
rs777095869 Chr.15: 74904627-74904627 on Build GRCh38
rs760302457 Chr.15: 74905106-74905106 on Build GRCh38
rs201851481 Chr.15: 74904682-74904682 on Build GRCh38
rs573495831 Chr.15: 74904741-74904741 on Build GRCh38
rs370919892 Chr.15: 74904740-74904740 on Build GRCh38
rs771010135 Chr.15: 74904712-74904712 on Build GRCh38
rs564030847 Chr.15: 74904977-74904977 on Build GRCh38
rs141614091 Chr.15: 74904699-74904699 on Build GRCh38
rs996290693 Chr.15: 74904959-74904959 on Build GRCh38
rs551409871 Chr.15: 74904932-74904932 on Build GRCh38
rs377112851 Chr.15: 74904662-74904662 on Build GRCh38
rs975216306 Chr.15: 74904709-74904709 on Build GRCh38
rs1054125273 Chr.15: 74904809-74904809 on Build GRCh38
rs138545501 Chr.15: 74904681-74904681 on Build GRCh38
rs774047339 Chr.15: 74904641-74904641 on Build GRCh38
rs974420507 Chr.15: 74904887-74904887 on Build GRCh38
rs1004013742 Chr.15: 74904845-74904845 on Build GRCh38
rs544343456 Chr.15: 74904787-74904787 on Build GRCh38
rs1029391204 Chr.15: 74904841-74904841 on Build GRCh38
rs906172533 Chr.15: 74904610-74904610 on Build GRCh38
rs781562694 Chr.15: 74905036-74905036 on Build GRCh38
rs376424699 Chr.15: 74905035-74905035 on Build GRCh38
rs1044827496 Chr.15: 74905054-74905054 on Build GRCh38
rs772823375 Chr.15: 74905053-74905053 on Build GRCh38
rs759319119 Chr.15: 74904747-74904747 on Build GRCh38
rs770633758 Chr.15: 74905092-74905092 on Build GRCh38
rs1012402587 Chr.15: 74904605-74904605 on Build GRCh38
rs925483486 Chr.15: 74904990-74904990 on Build GRCh38
rs770491887 Chr.15: 74904733-74904733 on Build GRCh38
rs759129067 Chr.15: 74905099-74905099 on Build GRCh38
rs899524913 Chr.15: 74904828-74904828 on Build GRCh38
rs906221970 Chr.15: 74904821-74904821 on Build GRCh38
rs771339856 Chr.15: 74904621-74904621 on Build GRCh38
rs923931051 Chr.15: 74905030-74905030 on Build GRCh38
rs575279806 Chr.15: 74904574-74904574 on Build GRCh38
rs777656598 Chr.15: 74904692-74904692 on Build GRCh38
rs781724101 Chr.15: 74904713-74904713 on Build GRCh38
rs752994432 Chr.15: 74904955-74904955 on Build GRCh38
rs529512809 Chr.15: 74904782-74904782 on Build GRCh38
rs545840111 Chr.15: 74904591-74904591 on Build GRCh38
rs773257556 Chr.15: 74904570-74904570 on Build GRCh38
rs1044765013 Chr.15: 74904609-74904609 on Build GRCh38
rs1053948882 Chr.15: 74904584-74904584 on Build GRCh38
rs757594855 Chr.15: 74905017-74905017 on Build GRCh38
rs774807997 Chr.15: 74904740-74904741 on Build GRCh38
rs962653126 Chr.15: 74904877-74904877 on Build GRCh38
rs373998090 Chr.15: 74904622-74904622 on Build GRCh38
rs372171726 Chr.15: 74904761-74904761 on Build GRCh38
rs759210453 Chr.15: 74904844-74904844 on Build GRCh38
rs752119433 Chr.15: 74904958-74904958 on Build GRCh38
rs962953842 Chr.15: 74904558-74904558 on Build GRCh38
rs746466509 Chr.15: 74905059-74905059 on Build GRCh38
rs992137788 Chr.15: 74904949-74904949 on Build GRCh38
rs938078081 Chr.15: 74904644-74904644 on Build GRCh38
rs779437658 Chr.15: 74904623-74904635 on Build GRCh38
rs752692286 Chr.15: 74904764-74904764 on Build GRCh38
rs756563330 Chr.15: 74905046-74905046 on Build GRCh38
rs932283753 Chr.15: 74905069-74905069 on Build GRCh38
rs118007749 Chr.15: 74904743-74904743 on Build GRCh38
rs767136299 Chr.15: 74904652-74904652 on Build GRCh38
rs954667114 Chr.15: 74904715-74904717 on Build GRCh38
rs771596042 Chr.15: 74904730-74904733 on Build GRCh38
rs964197879 Chr.15: 74904939-74904939 on Build GRCh38
rs768264890 Chr.15: 74905072-74905073 on Build GRCh38
rs767992593 Chr.15: 74904779-74904779 on Build GRCh38
rs562676314 Chr.15: 74904835-74904835 on Build GRCh38
rs959337170 Chr.15: 74904921-74904921 on Build GRCh38
rs144575000 Chr.15: 74904742-74904742 on Build GRCh38
rs765421342 Chr.15: 74904987-74904987 on Build GRCh38
rs533135914 Chr.15: 74904856-74904856 on Build GRCh38
rs147883291 Chr.15: 74905095-74905095 on Build GRCh38
rs766189641 Chr.15: 74904676-74904676 on Build GRCh38
rs751569894 Chr.15: 74905011-74905011 on Build GRCh38
rs746336816 Chr.15: 74904729-74904730 on Build GRCh38
rs535604993 Chr.15: 74904726-74904726 on Build GRCh38
rs746205162 Chr.15: 74904719-74904719 on Build GRCh38
rs1033627353 Chr.15: 74904897-74904897 on Build GRCh38
rs368080759 Chr.15: 74904743-74904744 on Build GRCh38
rs758489824 Chr.15: 74904688-74904688 on Build GRCh38
rs760143659 Chr.15: 74904633-74904633 on Build GRCh38
rs752785603 Chr.15: 74904686-74904686 on Build GRCh38
rs945472690 Chr.15: 74904984-74904984 on Build GRCh38
rs746295129 Chr.15: 74905037-74905037 on Build GRCh38
rs963527459 Chr.15: 74904683-74904683 on Build GRCh38
rs866118684 Chr.15: 74904674-74904674 on Build GRCh38
rs761384109 Chr.15: 74904646-74904646 on Build GRCh38
rs750107659 Chr.15: 74904659-74904659 on Build GRCh38
rs1005873328 Chr.15: 74904876-74904876 on Build GRCh38
rs749836486 Chr.15: 74905052-74905052 on Build GRCh38
rs528334285 Chr.15: 74904985-74904985 on Build GRCh38
rs776129458 Chr.15: 74904739-74904739 on Build GRCh38

View Full Product Details