Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74655606-74656170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
UniGene
Hs.682454
Gene Chromosome Location
Chr.15: 74630558-74696045 on Build GRCh38
Transcripts
NM_001142443.1 NM_001142444.1 NM_025083.3 XM_011522092.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153427 c.485-1G>T p.? Chr.15: 74656069-74656069 on Build GRCh38
6651551 c.485-7_485-6insT p.? Chr.15: 74656074-74656075 on Build GRCh38
5000933 c.572G>T p.C191F Chr.15: 74655981-74655981 on Build GRCh38
3503953 c.699C>T p.S233S Chr.15: 74655854-74655854 on Build GRCh38
6840809 c.646delG p.A216fs*47 Chr.15: 74655907-74655907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752566564 Chr.15: 74655880-74655880 on Build GRCh38
rs772170307 Chr.15: 74655777-74655777 on Build GRCh38
rs775891360 Chr.15: 74655825-74655825 on Build GRCh38
rs932039438 Chr.15: 74655676-74655676 on Build GRCh38
rs749664984 Chr.15: 74655967-74655967 on Build GRCh38
rs893973149 Chr.15: 74656087-74656087 on Build GRCh38
rs953361566 Chr.15: 74655757-74655757 on Build GRCh38
rs776787766 Chr.15: 74655793-74655793 on Build GRCh38
rs772333439 Chr.15: 74655708-74655708 on Build GRCh38
rs755570696 Chr.15: 74655760-74655760 on Build GRCh38
rs762291691 Chr.15: 74655985-74655985 on Build GRCh38
rs775641228 Chr.15: 74655966-74655966 on Build GRCh38
rs767052992 Chr.15: 74655756-74655756 on Build GRCh38
rs1052995494 Chr.15: 74656134-74656134 on Build GRCh38
rs759824978 Chr.15: 74655747-74655747 on Build GRCh38
rs771012149 Chr.15: 74656112-74656112 on Build GRCh38
rs75726871 Chr.15: 74655690-74655690 on Build GRCh38
rs370322738 Chr.15: 74655908-74655908 on Build GRCh38
rs76943949 Chr.15: 74655734-74655734 on Build GRCh38
rs1046670582 Chr.15: 74655660-74655660 on Build GRCh38
rs754633529 Chr.15: 74655878-74655878 on Build GRCh38
rs765905853 Chr.15: 74655860-74655860 on Build GRCh38
rs369652432 Chr.15: 74655755-74655755 on Build GRCh38
rs944902233 Chr.15: 74656126-74656126 on Build GRCh38
rs770978061 Chr.15: 74655791-74655791 on Build GRCh38
rs746117381 Chr.15: 74655947-74655947 on Build GRCh38
rs758212150 Chr.15: 74655893-74655893 on Build GRCh38
rs760273319 Chr.15: 74655986-74655986 on Build GRCh38
rs145915480 Chr.15: 74655815-74655815 on Build GRCh38
rs758336758 Chr.15: 74656049-74656049 on Build GRCh38
rs778751798 Chr.15: 74655774-74655774 on Build GRCh38
rs753131832 Chr.15: 74656025-74656025 on Build GRCh38
rs1001939113 Chr.15: 74655851-74655851 on Build GRCh38
rs766985395 Chr.15: 74655686-74655689 on Build GRCh38
rs758925076 Chr.15: 74656038-74656038 on Build GRCh38
rs930460930 Chr.15: 74655890-74655890 on Build GRCh38
rs776240001 Chr.15: 74655702-74655702 on Build GRCh38
rs753487087 Chr.15: 74655862-74655862 on Build GRCh38
rs141268344 Chr.15: 74655986-74655986 on Build GRCh38
rs764605310 Chr.15: 74655853-74655853 on Build GRCh38
rs748690391 Chr.15: 74655785-74655785 on Build GRCh38
rs776173074 Chr.15: 74656013-74656013 on Build GRCh38
rs971982773 Chr.15: 74655850-74655850 on Build GRCh38
rs949684656 Chr.15: 74655644-74655644 on Build GRCh38
rs561328785 Chr.15: 74655794-74655794 on Build GRCh38
rs7497342 Chr.15: 74655722-74655722 on Build GRCh38
rs556581951 Chr.15: 74655709-74655709 on Build GRCh38
rs368361738 Chr.15: 74656113-74656113 on Build GRCh38
rs140728378 Chr.15: 74655787-74655787 on Build GRCh38
rs960566889 Chr.15: 74655872-74655872 on Build GRCh38
rs562794805 Chr.15: 74656055-74656055 on Build GRCh38
rs920681952 Chr.15: 74655632-74655632 on Build GRCh38
rs920583730 Chr.15: 74656044-74656044 on Build GRCh38
rs377491253 Chr.15: 74655879-74655879 on Build GRCh38
rs202125728 Chr.15: 74656081-74656081 on Build GRCh38
rs768940764 Chr.15: 74655976-74655976 on Build GRCh38
rs143286435 Chr.15: 74656039-74656039 on Build GRCh38
rs986177905 Chr.15: 74656123-74656123 on Build GRCh38
rs892679695 Chr.15: 74655735-74655735 on Build GRCh38
rs760801853 Chr.15: 74655718-74655718 on Build GRCh38
rs748142645 Chr.15: 74655775-74655775 on Build GRCh38
rs755521800 Chr.15: 74655753-74655753 on Build GRCh38
rs147487761 Chr.15: 74655754-74655754 on Build GRCh38
rs746940876 Chr.15: 74655911-74655911 on Build GRCh38
rs763263440 Chr.15: 74655852-74655852 on Build GRCh38
rs145840933 Chr.15: 74655769-74655769 on Build GRCh38
rs544450073 Chr.15: 74655977-74655977 on Build GRCh38
rs912082243 Chr.15: 74656124-74656124 on Build GRCh38
rs779584456 Chr.15: 74655763-74655763 on Build GRCh38
rs906815203 Chr.15: 74655999-74655999 on Build GRCh38
rs745982305 Chr.15: 74656102-74656102 on Build GRCh38
rs1034873520 Chr.15: 74655855-74655855 on Build GRCh38
rs920437460 Chr.15: 74655861-74655861 on Build GRCh38
rs752333584 Chr.15: 74656048-74656048 on Build GRCh38
rs189497300 Chr.15: 74655859-74655859 on Build GRCh38
rs764782564 Chr.15: 74655716-74655716 on Build GRCh38
rs538490167 Chr.15: 74655696-74655696 on Build GRCh38
rs752378157 Chr.15: 74655708-74655708 on Build GRCh38
rs777808860 Chr.15: 74655778-74655778 on Build GRCh38
rs770084918 Chr.15: 74655949-74655949 on Build GRCh38
rs138195822 Chr.15: 74655920-74655920 on Build GRCh38
rs780950249 Chr.15: 74656097-74656097 on Build GRCh38
rs1009603616 Chr.15: 74655723-74655723 on Build GRCh38
rs753460829 Chr.15: 74655768-74655768 on Build GRCh38
rs991651963 Chr.15: 74655914-74655914 on Build GRCh38

View Full Product Details