Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67208285-67208797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383724 c.651A>G p.E217E Chr.15: 67208626-67208626 on Build GRCh38
3672100 c.668G>T p.R223L Chr.15: 67208609-67208609 on Build GRCh38
4555774 c.668G>A p.R223Q Chr.15: 67208609-67208609 on Build GRCh38
3678250 c.669G>A p.R223R Chr.15: 67208608-67208608 on Build GRCh38
6077879 c.699G>C p.Q233H Chr.15: 67208578-67208578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758419763 Chr.15: 67208538-67208539 on Build GRCh38
rs371443433 Chr.15: 67208522-67208522 on Build GRCh38
rs762124174 Chr.15: 67208604-67208604 on Build GRCh38
rs1034703212 Chr.15: 67208770-67208770 on Build GRCh38
rs957325140 Chr.15: 67208606-67208606 on Build GRCh38
rs758520652 Chr.15: 67208517-67208517 on Build GRCh38
rs933835191 Chr.15: 67208623-67208623 on Build GRCh38
rs201284578 Chr.15: 67208609-67208609 on Build GRCh38
rs567107219 Chr.15: 67208765-67208765 on Build GRCh38
rs150867081 Chr.15: 67208753-67208753 on Build GRCh38
rs770712735 Chr.15: 67208676-67208676 on Build GRCh38
rs1017462562 Chr.15: 67208736-67208736 on Build GRCh38
rs200248264 Chr.15: 67208581-67208581 on Build GRCh38
rs758992814 Chr.15: 67208710-67208710 on Build GRCh38
rs200297750 Chr.15: 67208655-67208655 on Build GRCh38
rs1014862723 Chr.15: 67208746-67208746 on Build GRCh38
rs544916355 Chr.15: 67208639-67208639 on Build GRCh38
rs189653983 Chr.15: 67208619-67208619 on Build GRCh38
rs564806767 Chr.15: 67208640-67208640 on Build GRCh38
rs368643210 Chr.15: 67208636-67208636 on Build GRCh38
rs780272145 Chr.15: 67208574-67208574 on Build GRCh38
rs199749230 Chr.15: 67208610-67208610 on Build GRCh38
rs1039093002 Chr.15: 67208660-67208660 on Build GRCh38
rs545489939 Chr.15: 67208744-67208744 on Build GRCh38
rs747915052 Chr.15: 67208551-67208551 on Build GRCh38
rs768479452 Chr.15: 67208539-67208539 on Build GRCh38
rs765473955 Chr.15: 67208607-67208607 on Build GRCh38
rs201133630 Chr.15: 67208603-67208603 on Build GRCh38
rs770626942 Chr.15: 67208587-67208587 on Build GRCh38
rs199567850 Chr.15: 67208674-67208674 on Build GRCh38
rs1009135875 Chr.15: 67208696-67208696 on Build GRCh38
rs899166516 Chr.15: 67208669-67208669 on Build GRCh38
rs747015082 Chr.15: 67208538-67208538 on Build GRCh38
rs2289262 Chr.15: 67208695-67208695 on Build GRCh38
rs757911123 Chr.15: 67208739-67208739 on Build GRCh38
rs1051160639 Chr.15: 67208678-67208678 on Build GRCh38
rs1029760899 Chr.15: 67208418-67208418 on Build GRCh38
rs36096355 Chr.15: 67208595-67208595 on Build GRCh38
rs560715437 Chr.15: 67208731-67208731 on Build GRCh38
rs569290502 Chr.15: 67208508-67208508 on Build GRCh38
rs775130747 Chr.15: 67208756-67208756 on Build GRCh38
rs1001238912 Chr.15: 67208769-67208769 on Build GRCh38
rs559278371 Chr.15: 67208463-67208463 on Build GRCh38
rs950958444 Chr.15: 67208370-67208370 on Build GRCh38
rs12904837 Chr.15: 67208378-67208378 on Build GRCh38
rs761177603 Chr.15: 67208597-67208597 on Build GRCh38
rs1014396136 Chr.15: 67208423-67208423 on Build GRCh38
rs367950785 Chr.15: 67208700-67208700 on Build GRCh38
rs994288733 Chr.15: 67208391-67208391 on Build GRCh38
rs773041947 Chr.15: 67208579-67208579 on Build GRCh38
rs774420450 Chr.15: 67208510-67208510 on Build GRCh38
rs769444076 Chr.15: 67208576-67208576 on Build GRCh38
rs989531679 Chr.15: 67208550-67208550 on Build GRCh38
rs547214794 Chr.15: 67208323-67208323 on Build GRCh38
rs539371258 Chr.15: 67208350-67208350 on Build GRCh38
rs377511353 Chr.15: 67208548-67208548 on Build GRCh38
rs139418005 Chr.15: 67208763-67208763 on Build GRCh38
rs567070752 Chr.15: 67208348-67208348 on Build GRCh38
rs1022173818 Chr.15: 67208747-67208747 on Build GRCh38
rs117853536 Chr.15: 67208608-67208608 on Build GRCh38

View Full Product Details