Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23758375-23758905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHP2
Entrez Gene ID
63928
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
-
UniGene
Hs.178589
Gene Chromosome Location
Chr.16: 23754627-23758951 on Build GRCh38
Transcripts
NM_022097.3

SNP Detail

SNP ID Chromosome Location
rs1007497524 Chr.16: 23758659-23758659 on Build GRCh38
rs559732908 Chr.16: 23758524-23758524 on Build GRCh38
rs993777704 Chr.16: 23758600-23758600 on Build GRCh38
rs1032210022 Chr.16: 23758714-23758714 on Build GRCh38
rs890409298 Chr.16: 23758650-23758650 on Build GRCh38
rs925206209 Chr.16: 23758826-23758826 on Build GRCh38
rs192983978 Chr.16: 23758787-23758787 on Build GRCh38
rs926309011 Chr.16: 23758546-23758546 on Build GRCh38
rs960108838 Chr.16: 23758780-23758780 on Build GRCh38
rs139517450 Chr.16: 23758407-23758407 on Build GRCh38
rs917520442 Chr.16: 23758491-23758491 on Build GRCh38
rs577487793 Chr.16: 23758825-23758825 on Build GRCh38
rs1006433878 Chr.16: 23758706-23758706 on Build GRCh38
rs8051073 Chr.16: 23758803-23758803 on Build GRCh38
rs993090335 Chr.16: 23758462-23758462 on Build GRCh38
rs1051663760 Chr.16: 23758625-23758625 on Build GRCh38
rs556266495 Chr.16: 23758802-23758802 on Build GRCh38
rs188649581 Chr.16: 23758765-23758765 on Build GRCh38
rs969129868 Chr.16: 23758704-23758704 on Build GRCh38
rs766339738 Chr.16: 23758747-23758747 on Build GRCh38
rs943035220 Chr.16: 23758596-23758596 on Build GRCh38
rs755074815 Chr.16: 23758791-23758791 on Build GRCh38
rs943296865 Chr.16: 23758832-23758832 on Build GRCh38
rs974388609 Chr.16: 23758839-23758839 on Build GRCh38
rs866065104 Chr.16: 23758863-23758863 on Build GRCh38
rs1017594961 Chr.16: 23758691-23758691 on Build GRCh38
rs1000275916 Chr.16: 23758710-23758710 on Build GRCh38
rs773292367 Chr.16: 23758543-23758543 on Build GRCh38
rs555095443 Chr.16: 23758613-23758613 on Build GRCh38
rs956211924 Chr.16: 23758764-23758764 on Build GRCh38
rs763981383 Chr.16: 23758824-23758824 on Build GRCh38
rs796839191 Chr.16: 23758574-23758574 on Build GRCh38
rs926258314 Chr.16: 23758813-23758813 on Build GRCh38
rs979767510 Chr.16: 23758564-23758564 on Build GRCh38
rs773604991 Chr.16: 23758443-23758443 on Build GRCh38
rs1016487515 Chr.16: 23758716-23758716 on Build GRCh38
rs753968851 Chr.16: 23758782-23758782 on Build GRCh38
rs941749272 Chr.16: 23758560-23758560 on Build GRCh38
rs1047887960 Chr.16: 23758649-23758649 on Build GRCh38
rs565842345 Chr.16: 23758446-23758446 on Build GRCh38
rs1035748725 Chr.16: 23758421-23758421 on Build GRCh38
rs922970858 Chr.16: 23758848-23758848 on Build GRCh38
rs538276591 Chr.16: 23758715-23758715 on Build GRCh38
rs374639812 Chr.16: 23758783-23758783 on Build GRCh38
rs1035675289 Chr.16: 23758772-23758772 on Build GRCh38
rs770777226 Chr.16: 23758503-23758503 on Build GRCh38
rs761808579 Chr.16: 23758671-23758671 on Build GRCh38
rs945307815 Chr.16: 23758547-23758548 on Build GRCh38
rs1044681951 Chr.16: 23758506-23758506 on Build GRCh38
rs960141044 Chr.16: 23758437-23758437 on Build GRCh38
rs1020754083 Chr.16: 23758786-23758786 on Build GRCh38
rs933089398 Chr.16: 23758623-23758623 on Build GRCh38
rs979351641 Chr.16: 23758816-23758816 on Build GRCh38
rs145147975 Chr.16: 23758597-23758597 on Build GRCh38
rs956437497 Chr.16: 23758458-23758458 on Build GRCh38
rs761026113 Chr.16: 23758595-23758595 on Build GRCh38

View Full Product Details