Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132196233-132196808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT9
Entrez Gene ID
50614
Gene Name
polypeptide N-acetylgalactosaminyltransferase 9
Gene Aliases
GALNAC-T9, GALNACT9
UniGene
Hs.301062
Gene Chromosome Location
Chr.12: 132195829-132329364 on Build GRCh38
Transcripts
NM_001122636.1 NM_021808.3 XM_017019371.1 XM_017019372.1 XM_017019373.1 XM_017019374.1

SNP Detail

SNP ID Chromosome Location
rs534581396 Chr.12: 132196401-132196401 on Build GRCh38
rs911879089 Chr.12: 132196584-132196584 on Build GRCh38
rs556939645 Chr.12: 132196756-132196756 on Build GRCh38
rs991994516 Chr.12: 132196660-132196660 on Build GRCh38
rs76055951 Chr.12: 132196623-132196623 on Build GRCh38
rs184026614 Chr.12: 132196585-132196585 on Build GRCh38
rs1008519328 Chr.12: 132196299-132196299 on Build GRCh38
rs1015822823 Chr.12: 132196322-132196322 on Build GRCh38
rs903927439 Chr.12: 132196303-132196303 on Build GRCh38
rs1032016808 Chr.12: 132196433-132196433 on Build GRCh38
rs769879395 Chr.12: 132196600-132196600 on Build GRCh38
rs1051770577 Chr.12: 132196387-132196387 on Build GRCh38
rs995695417 Chr.12: 132196778-132196778 on Build GRCh38
rs375431554 Chr.12: 132196398-132196398 on Build GRCh38
rs566076000 Chr.12: 132196503-132196503 on Build GRCh38
rs113805462 Chr.12: 132196337-132196337 on Build GRCh38
rs188763457 Chr.12: 132196647-132196647 on Build GRCh38
rs202077963 Chr.12: 132196397-132196398 on Build GRCh38
rs550280202 Chr.12: 132196461-132196461 on Build GRCh38
rs746351826 Chr.12: 132196593-132196593 on Build GRCh38
rs1031987177 Chr.12: 132196306-132196306 on Build GRCh38
rs961943135 Chr.12: 132196327-132196327 on Build GRCh38
rs919383254 Chr.12: 132196525-132196525 on Build GRCh38
rs1053547390 Chr.12: 132196642-132196642 on Build GRCh38
rs528665884 Chr.12: 132196509-132196509 on Build GRCh38
rs929401675 Chr.12: 132196551-132196551 on Build GRCh38
rs899653381 Chr.12: 132196423-132196423 on Build GRCh38
rs911894948 Chr.12: 132196780-132196781 on Build GRCh38
rs551983886 Chr.12: 132196569-132196569 on Build GRCh38
rs897809699 Chr.12: 132196269-132196269 on Build GRCh38
rs980521663 Chr.12: 132196366-132196366 on Build GRCh38
rs926380803 Chr.12: 132196382-132196382 on Build GRCh38
rs1027151803 Chr.12: 132196779-132196779 on Build GRCh38
rs139915363 Chr.12: 132196438-132196438 on Build GRCh38
rs771171367 Chr.12: 132196468-132196468 on Build GRCh38
rs768814109 Chr.12: 132196717-132196717 on Build GRCh38
rs951794447 Chr.12: 132196350-132196350 on Build GRCh38
rs763188957 Chr.12: 132196630-132196630 on Build GRCh38
rs950888082 Chr.12: 132196772-132196772 on Build GRCh38
rs994113715 Chr.12: 132196270-132196270 on Build GRCh38
rs779697988 Chr.12: 132196395-132196395 on Build GRCh38
rs943972792 Chr.12: 132196599-132196599 on Build GRCh38
rs945316486 Chr.12: 132196428-132196428 on Build GRCh38
rs118000178 Chr.12: 132196305-132196305 on Build GRCh38
rs1016204658 Chr.12: 132196481-132196481 on Build GRCh38
rs955558958 Chr.12: 132196309-132196309 on Build GRCh38
rs370172380 Chr.12: 132196341-132196341 on Build GRCh38
rs1028494663 Chr.12: 132196308-132196308 on Build GRCh38
rs191551041 Chr.12: 132196472-132196472 on Build GRCh38
rs996926139 Chr.12: 132196462-132196462 on Build GRCh38
rs897346855 Chr.12: 132196775-132196775 on Build GRCh38
rs1009302572 Chr.12: 132196662-132196662 on Build GRCh38
rs963621403 Chr.12: 132196749-132196749 on Build GRCh38
rs757160716 Chr.12: 132196261-132196261 on Build GRCh38
rs377689608 Chr.12: 132196304-132196304 on Build GRCh38
rs181549382 Chr.12: 132196673-132196673 on Build GRCh38
rs143416725 Chr.12: 132196711-132196711 on Build GRCh38
rs1037764629 Chr.12: 132196338-132196338 on Build GRCh38
rs111971104 Chr.12: 132196315-132196315 on Build GRCh38
rs960458760 Chr.12: 132196652-132196652 on Build GRCh38
rs1027267816 Chr.12: 132196576-132196576 on Build GRCh38
rs1022732974 Chr.12: 132196282-132196282 on Build GRCh38
rs765379820 Chr.12: 132196676-132196676 on Build GRCh38
rs187963491 Chr.12: 132196442-132196442 on Build GRCh38
rs111429378 Chr.12: 132196383-132196383 on Build GRCh38
rs961631290 Chr.12: 132196550-132196550 on Build GRCh38
rs77604987 Chr.12: 132196677-132196677 on Build GRCh38
rs377000630 Chr.12: 132196474-132196474 on Build GRCh38
rs906141949 Chr.12: 132196781-132196781 on Build GRCh38
rs890966887 Chr.12: 132196289-132196289 on Build GRCh38
rs1049769720 Chr.12: 132196720-132196720 on Build GRCh38
rs754637403 Chr.12: 132196427-132196427 on Build GRCh38
rs891202737 Chr.12: 132196745-132196745 on Build GRCh38
rs116383641 Chr.12: 132196328-132196328 on Build GRCh38
rs1025083563 Chr.12: 132196672-132196672 on Build GRCh38
rs1047389116 Chr.12: 132196284-132196284 on Build GRCh38
rs114971933 Chr.12: 132196463-132196463 on Build GRCh38
rs73166822 Chr.12: 132196755-132196755 on Build GRCh38
rs139836376 Chr.12: 132196520-132196521 on Build GRCh38
rs1024635039 Chr.12: 132196453-132196453 on Build GRCh38
rs1044069683 Chr.12: 132196686-132196686 on Build GRCh38
rs899682828 Chr.12: 132196608-132196608 on Build GRCh38

View Full Product Details