Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66614601-66615125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
UniGene
Hs.643961
Gene Chromosome Location
Chr.16: 66598178-66696707 on Build GRCh38
Transcripts
NM_178818.2 NM_181521.2 XM_011522882.1 XM_017022954.1

SNP Detail

SNP ID Chromosome Location
rs772440218 Chr.16: 66615065-66615065 on Build GRCh38
rs958582093 Chr.16: 66614703-66614703 on Build GRCh38
rs138828819 Chr.16: 66614640-66614640 on Build GRCh38
rs200202076 Chr.16: 66614957-66614957 on Build GRCh38
rs1016715429 Chr.16: 66614988-66614988 on Build GRCh38
rs559477550 Chr.16: 66615094-66615094 on Build GRCh38
rs972396708 Chr.16: 66615017-66615017 on Build GRCh38
rs950928116 Chr.16: 66615054-66615054 on Build GRCh38
rs912061477 Chr.16: 66614743-66614743 on Build GRCh38
rs888242976 Chr.16: 66615056-66615056 on Build GRCh38
rs993977911 Chr.16: 66614634-66614634 on Build GRCh38
rs943579020 Chr.16: 66615098-66615098 on Build GRCh38
rs1003470200 Chr.16: 66614933-66614933 on Build GRCh38
rs370514063 Chr.16: 66615047-66615047 on Build GRCh38
rs936879005 Chr.16: 66614931-66614931 on Build GRCh38
rs528075161 Chr.16: 66614784-66614784 on Build GRCh38
rs918141973 Chr.16: 66614748-66614748 on Build GRCh38
rs901197714 Chr.16: 66614971-66614971 on Build GRCh38
rs987720000 Chr.16: 66615056-66615056 on Build GRCh38
rs868603402 Chr.16: 66614648-66614648 on Build GRCh38
rs994077727 Chr.16: 66614979-66614979 on Build GRCh38
rs952558229 Chr.16: 66614665-66614665 on Build GRCh38
rs541509181 Chr.16: 66614702-66614702 on Build GRCh38
rs962675668 Chr.16: 66614997-66614997 on Build GRCh38
rs748971539 Chr.16: 66614952-66614952 on Build GRCh38
rs554686940 Chr.16: 66614811-66614811 on Build GRCh38
rs192135166 Chr.16: 66614846-66614846 on Build GRCh38
rs1045617 Chr.16: 66614818-66614818 on Build GRCh38
rs765026922 Chr.16: 66614625-66614625 on Build GRCh38
rs866091830 Chr.16: 66614698-66614698 on Build GRCh38
rs1039152530 Chr.16: 66614776-66614776 on Build GRCh38
rs756812057 Chr.16: 66614694-66614694 on Build GRCh38
rs750100839 Chr.16: 66614731-66614731 on Build GRCh38
rs974052644 Chr.16: 66614675-66614675 on Build GRCh38
rs946760656 Chr.16: 66614829-66614829 on Build GRCh38
rs559090055 Chr.16: 66614628-66614628 on Build GRCh38
rs548163404 Chr.16: 66614956-66614956 on Build GRCh38
rs545813058 Chr.16: 66615066-66615066 on Build GRCh38
rs902403058 Chr.16: 66614915-66614915 on Build GRCh38
rs530168638 Chr.16: 66614697-66614697 on Build GRCh38
rs943452443 Chr.16: 66614759-66614759 on Build GRCh38
rs1045604 Chr.16: 66614889-66614889 on Build GRCh38
rs4538003 Chr.16: 66615093-66615093 on Build GRCh38
rs748552754 Chr.16: 66615012-66615012 on Build GRCh38
rs532241114 Chr.16: 66615037-66615037 on Build GRCh38
rs978344822 Chr.16: 66614842-66614842 on Build GRCh38
rs780781556 Chr.16: 66614808-66614808 on Build GRCh38
rs949623022 Chr.16: 66614796-66614796 on Build GRCh38
rs779245756 Chr.16: 66614958-66614958 on Build GRCh38

View Full Product Details