Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39470711-39471097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK12
Entrez Gene ID
51755
Gene Name
cyclin dependent kinase 12
Gene Aliases
CRK7, CRKR, CRKRS
UniGene
Hs.345028
Gene Chromosome Location
Chr.17: 39461486-39567560 on Build GRCh38
Transcripts
NM_015083.2 NM_016507.3 XM_005257456.3 XM_005257458.4 XM_011524892.2 XM_011524893.2 XM_011524894.2 XM_011524895.2 XM_011524896.2 XM_011524897.2 XM_011524898.2 XM_011524899.2 XM_011524900.2 XM_011524901.2 XM_011524902.2 XM_011524903.2 XM_011524905.2 XM_011524906.2 XM_011524907.2 XM_017024744.1 XM_017024745.1 XM_017024746.1 XM_017024747.1 XM_017024748.1 XM_017024749.1 XM_017024750.1 XM_017024751.1 XM_017024752.1 XM_017024753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971026 c.1259_1262delCCAA p.S420fs*15 Chr.17: 39471091-39471094 on Build GRCh38
6924366 c.1097C>G p.S366* Chr.17: 39470929-39470929 on Build GRCh38
4065968 c.1054T>G p.S352A Chr.17: 39470886-39470886 on Build GRCh38
4065969 c.1126A>G p.S376G Chr.17: 39470958-39470958 on Build GRCh38
4065970 c.1132C>T p.R378C Chr.17: 39470964-39470964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939275643 Chr.17: 39470816-39470816 on Build GRCh38
rs113134934 Chr.17: 39471004-39471004 on Build GRCh38
rs867025919 Chr.17: 39470833-39470833 on Build GRCh38
rs961048592 Chr.17: 39470761-39470761 on Build GRCh38
rs1016267621 Chr.17: 39471067-39471067 on Build GRCh38
rs200488621 Chr.17: 39470934-39470934 on Build GRCh38
rs765737274 Chr.17: 39470981-39470981 on Build GRCh38
rs573194227 Chr.17: 39470768-39470768 on Build GRCh38
rs148279954 Chr.17: 39471002-39471002 on Build GRCh38
rs760288133 Chr.17: 39470869-39470869 on Build GRCh38
rs764336794 Chr.17: 39471056-39471056 on Build GRCh38
rs113172957 Chr.17: 39470770-39470770 on Build GRCh38
rs771747794 Chr.17: 39470961-39470961 on Build GRCh38
rs754381959 Chr.17: 39470886-39470886 on Build GRCh38
rs766724817 Chr.17: 39470904-39470904 on Build GRCh38
rs138292741 Chr.17: 39470877-39470877 on Build GRCh38
rs774313790 Chr.17: 39470847-39470847 on Build GRCh38
rs1043468837 Chr.17: 39470867-39470867 on Build GRCh38
rs373258085 Chr.17: 39470837-39470837 on Build GRCh38
rs751710276 Chr.17: 39470916-39470916 on Build GRCh38
rs947889462 Chr.17: 39470836-39470836 on Build GRCh38
rs749257918 Chr.17: 39470832-39470832 on Build GRCh38
rs755536892 Chr.17: 39471053-39471054 on Build GRCh38
rs765683870 Chr.17: 39470890-39470890 on Build GRCh38
rs764436473 Chr.17: 39470914-39470914 on Build GRCh38
rs1042434482 Chr.17: 39470871-39470871 on Build GRCh38
rs758987935 Chr.17: 39470979-39470979 on Build GRCh38
rs748037335 Chr.17: 39470951-39470951 on Build GRCh38
rs150224634 Chr.17: 39470887-39470887 on Build GRCh38
rs376212098 Chr.17: 39470932-39470932 on Build GRCh38
rs142798057 Chr.17: 39471066-39471066 on Build GRCh38
rs540707885 Chr.17: 39470771-39470771 on Build GRCh38
rs375280392 Chr.17: 39470965-39470965 on Build GRCh38
rs200075664 Chr.17: 39470971-39470971 on Build GRCh38
rs753119390 Chr.17: 39470899-39470899 on Build GRCh38
rs183513859 Chr.17: 39471052-39471052 on Build GRCh38
rs367868824 Chr.17: 39471005-39471005 on Build GRCh38
rs772732127 Chr.17: 39470864-39470864 on Build GRCh38
rs768678277 Chr.17: 39470846-39470846 on Build GRCh38
rs1056424565 Chr.17: 39470826-39470826 on Build GRCh38
rs866483767 Chr.17: 39471011-39471011 on Build GRCh38
rs749210177 Chr.17: 39470840-39470840 on Build GRCh38
rs377487664 Chr.17: 39470855-39470855 on Build GRCh38
rs144663238 Chr.17: 39470978-39470978 on Build GRCh38
rs760065247 Chr.17: 39470889-39470889 on Build GRCh38
rs191130503 Chr.17: 39470809-39470809 on Build GRCh38
rs563325093 Chr.17: 39470938-39470938 on Build GRCh38
rs991434899 Chr.17: 39470825-39470825 on Build GRCh38
rs946819913 Chr.17: 39470863-39470863 on Build GRCh38
rs1015290178 Chr.17: 39470760-39470760 on Build GRCh38
rs778932012 Chr.17: 39470944-39470944 on Build GRCh38
rs370304179 Chr.17: 39470964-39470964 on Build GRCh38
rs746609194 Chr.17: 39470970-39470970 on Build GRCh38
rs200070621 Chr.17: 39470861-39470861 on Build GRCh38

View Full Product Details