Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42482930-42483408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A1
Entrez Gene ID
535
Gene Name
ATPase H+ transporting V0 subunit a1
Gene Aliases
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
UniGene
Hs.463074
Gene Chromosome Location
Chr.17: 42458844-42522579 on Build GRCh38
Transcripts
NM_001130020.1 NM_001130021.1 NM_005177.3 XM_005257459.1 XM_005257461.1 XM_005257463.2 XM_011524908.1 XM_017024766.1 XM_017024767.1 XM_017024768.1 XM_017024769.1 XM_017024770.1 XM_017024771.1 XM_017024772.1 XM_017024773.1 XM_017024774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7138166 c.749C>T p.S250L Chr.17: 42483049-42483049 on Build GRCh38
706304 c.728C>T p.S243L Chr.17: 42483049-42483049 on Build GRCh38
7132411 c.763C>A p.P255T Chr.17: 42483063-42483063 on Build GRCh38
1302844 c.742C>A p.P248T Chr.17: 42483063-42483063 on Build GRCh38
4507829 c.758C>T p.P253L Chr.17: 42483058-42483058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768977132 Chr.17: 42483071-42483071 on Build GRCh38
rs765000931 Chr.17: 42483008-42483008 on Build GRCh38
rs35520937 Chr.17: 42483101-42483101 on Build GRCh38
rs114909848 Chr.17: 42482975-42482975 on Build GRCh38
rs921398138 Chr.17: 42483304-42483304 on Build GRCh38
rs532994313 Chr.17: 42483219-42483219 on Build GRCh38
rs759371429 Chr.17: 42483018-42483018 on Build GRCh38
rs1021975227 Chr.17: 42483221-42483221 on Build GRCh38
rs533877949 Chr.17: 42483343-42483343 on Build GRCh38
rs773467067 Chr.17: 42483182-42483182 on Build GRCh38
rs762123399 Chr.17: 42483022-42483022 on Build GRCh38
rs746452583 Chr.17: 42482990-42482990 on Build GRCh38
rs765122384 Chr.17: 42483019-42483019 on Build GRCh38
rs779368097 Chr.17: 42483114-42483114 on Build GRCh38
rs746654866 Chr.17: 42483128-42483128 on Build GRCh38
rs542366227 Chr.17: 42483110-42483110 on Build GRCh38
rs932880332 Chr.17: 42482959-42482959 on Build GRCh38
rs775357466 Chr.17: 42483169-42483169 on Build GRCh38
rs774811493 Chr.17: 42483186-42483186 on Build GRCh38
rs766068403 Chr.17: 42483371-42483371 on Build GRCh38
rs1048615334 Chr.17: 42483363-42483363 on Build GRCh38
rs878912897 Chr.17: 42483215-42483215 on Build GRCh38
rs745837894 Chr.17: 42483138-42483138 on Build GRCh38
rs1023379542 Chr.17: 42483245-42483245 on Build GRCh38
rs143549371 Chr.17: 42483118-42483118 on Build GRCh38
rs929433637 Chr.17: 42483053-42483053 on Build GRCh38
rs770561379 Chr.17: 42482995-42482995 on Build GRCh38
rs1032185296 Chr.17: 42483302-42483302 on Build GRCh38
rs957961464 Chr.17: 42483330-42483330 on Build GRCh38
rs1003664902 Chr.17: 42483142-42483142 on Build GRCh38
rs777626938 Chr.17: 42483123-42483123 on Build GRCh38
rs756257957 Chr.17: 42483075-42483075 on Build GRCh38
rs888422651 Chr.17: 42483076-42483076 on Build GRCh38
rs562325041 Chr.17: 42483143-42483143 on Build GRCh38
rs748395848 Chr.17: 42483117-42483117 on Build GRCh38
rs566532526 Chr.17: 42483341-42483341 on Build GRCh38
rs924313429 Chr.17: 42483270-42483270 on Build GRCh38
rs763876960 Chr.17: 42483180-42483180 on Build GRCh38
rs373733840 Chr.17: 42483137-42483137 on Build GRCh38
rs760982256 Chr.17: 42483004-42483008 on Build GRCh38
rs879414446 Chr.17: 42483284-42483284 on Build GRCh38
rs62075812 Chr.17: 42483383-42483383 on Build GRCh38
rs142427885 Chr.17: 42483258-42483258 on Build GRCh38
rs899426119 Chr.17: 42483038-42483038 on Build GRCh38
rs913612258 Chr.17: 42482970-42482970 on Build GRCh38
rs775110321 Chr.17: 42483020-42483020 on Build GRCh38
rs762674642 Chr.17: 42483179-42483179 on Build GRCh38
rs766418874 Chr.17: 42483088-42483088 on Build GRCh38
rs1012802488 Chr.17: 42483207-42483207 on Build GRCh38
rs965096739 Chr.17: 42483282-42483282 on Build GRCh38
rs72823091 Chr.17: 42483256-42483256 on Build GRCh38
rs376470573 Chr.17: 42483003-42483003 on Build GRCh38
rs754421311 Chr.17: 42483093-42483093 on Build GRCh38
rs750510954 Chr.17: 42483074-42483074 on Build GRCh38
rs776688997 Chr.17: 42482997-42482997 on Build GRCh38
rs551292997 Chr.17: 42483135-42483135 on Build GRCh38

View Full Product Details