Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5392294-5392889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

SNP Detail

SNP ID Chromosome Location
rs189939395 Chr.18: 5392646-5392646 on Build GRCh38
rs543792657 Chr.18: 5392685-5392685 on Build GRCh38
rs995735973 Chr.18: 5392748-5392748 on Build GRCh38
rs141378527 Chr.18: 5392812-5392812 on Build GRCh38
rs148690503 Chr.18: 5392777-5392777 on Build GRCh38
rs982793208 Chr.18: 5392459-5392459 on Build GRCh38
rs144097441 Chr.18: 5392463-5392463 on Build GRCh38
rs892601863 Chr.18: 5392359-5392359 on Build GRCh38
rs9953490 Chr.18: 5392654-5392654 on Build GRCh38
rs914201372 Chr.18: 5392641-5392641 on Build GRCh38
rs574223972 Chr.18: 5392733-5392733 on Build GRCh38
rs958789607 Chr.18: 5392848-5392848 on Build GRCh38
rs552139619 Chr.18: 5392813-5392813 on Build GRCh38
rs1140836 Chr.18: 5392524-5392524 on Build GRCh38
rs371333000 Chr.18: 5392499-5392499 on Build GRCh38
rs368268009 Chr.18: 5392562-5392562 on Build GRCh38
rs571174738 Chr.18: 5392404-5392404 on Build GRCh38
rs776566296 Chr.18: 5392645-5392645 on Build GRCh38
rs937306739 Chr.18: 5392598-5392602 on Build GRCh38
rs535934864 Chr.18: 5392482-5392482 on Build GRCh38
rs138082368 Chr.18: 5392348-5392348 on Build GRCh38
rs556434184 Chr.18: 5392556-5392556 on Build GRCh38
rs1009135068 Chr.18: 5392321-5392321 on Build GRCh38
rs1015292957 Chr.18: 5392816-5392816 on Build GRCh38
rs1050110405 Chr.18: 5392640-5392640 on Build GRCh38
rs1053083324 Chr.18: 5392650-5392650 on Build GRCh38
rs73937119 Chr.18: 5392450-5392450 on Build GRCh38
rs964357295 Chr.18: 5392393-5392393 on Build GRCh38
rs768631531 Chr.18: 5392639-5392639 on Build GRCh38
rs1140837 Chr.18: 5392469-5392469 on Build GRCh38
rs572972201 Chr.18: 5392527-5392527 on Build GRCh38
rs367633963 Chr.18: 5392392-5392392 on Build GRCh38
rs532349975 Chr.18: 5392771-5392771 on Build GRCh38
rs1024995618 Chr.18: 5392801-5392801 on Build GRCh38
rs920373373 Chr.18: 5392601-5392601 on Build GRCh38
rs900083176 Chr.18: 5392739-5392739 on Build GRCh38
rs58241380 Chr.18: 5392344-5392344 on Build GRCh38
rs1020054893 Chr.18: 5392323-5392323 on Build GRCh38
rs1018924950 Chr.18: 5392386-5392386 on Build GRCh38
rs369819844 Chr.18: 5392805-5392805 on Build GRCh38
rs1022764077 Chr.18: 5392412-5392412 on Build GRCh38
rs867634245 Chr.18: 5392844-5392844 on Build GRCh38
rs531108860 Chr.18: 5392862-5392862 on Build GRCh38
rs969443565 Chr.18: 5392438-5392438 on Build GRCh38
rs764248459 Chr.18: 5392629-5392630 on Build GRCh38
rs529512019 Chr.18: 5392319-5392319 on Build GRCh38
rs72865098 Chr.18: 5392832-5392832 on Build GRCh38
rs974474330 Chr.18: 5392441-5392441 on Build GRCh38
rs962144800 Chr.18: 5392590-5392590 on Build GRCh38
rs563602296 Chr.18: 5392734-5392734 on Build GRCh38
rs528815116 Chr.18: 5392383-5392383 on Build GRCh38
rs905317488 Chr.18: 5392755-5392755 on Build GRCh38
rs945707511 Chr.18: 5392642-5392642 on Build GRCh38
rs973256782 Chr.18: 5392591-5392591 on Build GRCh38
rs371511364 Chr.18: 5392507-5392507 on Build GRCh38
rs567071753 Chr.18: 5392477-5392477 on Build GRCh38
rs559721929 Chr.18: 5392778-5392778 on Build GRCh38
rs563668426 Chr.18: 5392710-5392710 on Build GRCh38

View Full Product Details