Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82255303-82255899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1D
Entrez Gene ID
1453
Gene Name
casein kinase 1 delta
Gene Aliases
ASPS, CKIdelta, FASPS2, HCKID
UniGene
Hs.631725
Gene Chromosome Location
Chr.17: 82239317-82273742 on Build GRCh38
Transcripts
NM_001893.4 NM_139062.2 NR_110578.1 XM_005256336.3 XM_005256337.4 XM_017024199.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636729 c.214G>A p.G72R Chr.17: 82255551-82255551 on Build GRCh38
3523824 c.306C>T p.L102L Chr.17: 82255459-82255459 on Build GRCh38
5882961 c.276C>G p.L92L Chr.17: 82255489-82255489 on Build GRCh38
6636726 c.288C>A p.C96* Chr.17: 82255477-82255477 on Build GRCh38
4070892 c.274C>A p.L92I Chr.17: 82255491-82255491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533632187 Chr.17: 82255492-82255492 on Build GRCh38
rs991086603 Chr.17: 82255695-82255695 on Build GRCh38
rs979186833 Chr.17: 82255658-82255658 on Build GRCh38
rs753408118 Chr.17: 82255453-82255453 on Build GRCh38
rs919414858 Chr.17: 82255383-82255383 on Build GRCh38
rs1053806428 Chr.17: 82255416-82255416 on Build GRCh38
rs911027320 Chr.17: 82255869-82255869 on Build GRCh38
rs771648482 Chr.17: 82255412-82255413 on Build GRCh38
rs759476027 Chr.17: 82255407-82255407 on Build GRCh38
rs375506434 Chr.17: 82255716-82255716 on Build GRCh38
rs757868957 Chr.17: 82255441-82255441 on Build GRCh38
rs959396720 Chr.17: 82255795-82255795 on Build GRCh38
rs918505131 Chr.17: 82255856-82255856 on Build GRCh38
rs889868846 Chr.17: 82255741-82255741 on Build GRCh38
rs770436735 Chr.17: 82255531-82255533 on Build GRCh38
rs943448923 Chr.17: 82255821-82255821 on Build GRCh38
rs751323232 Chr.17: 82255777-82255777 on Build GRCh38
rs909444075 Chr.17: 82255349-82255349 on Build GRCh38
rs1051850112 Chr.17: 82255389-82255389 on Build GRCh38
rs747737044 Chr.17: 82255484-82255484 on Build GRCh38
rs1019570752 Chr.17: 82255535-82255535 on Build GRCh38
rs1041115965 Chr.17: 82255874-82255874 on Build GRCh38
rs751850801 Chr.17: 82255528-82255528 on Build GRCh38
rs766688524 Chr.17: 82255512-82255512 on Build GRCh38
rs990900057 Chr.17: 82255681-82255681 on Build GRCh38
rs764370790 Chr.17: 82255561-82255561 on Build GRCh38
rs760578322 Chr.17: 82255391-82255391 on Build GRCh38
rs1011487831 Chr.17: 82255474-82255474 on Build GRCh38
rs771944737 Chr.17: 82255490-82255490 on Build GRCh38
rs373904715 Chr.17: 82255618-82255618 on Build GRCh38
rs751992018 Chr.17: 82255596-82255596 on Build GRCh38
rs116389388 Chr.17: 82255756-82255756 on Build GRCh38
rs1036880177 Chr.17: 82255392-82255392 on Build GRCh38
rs549082888 Chr.17: 82255720-82255720 on Build GRCh38
rs368602756 Chr.17: 82255353-82255353 on Build GRCh38
rs1002684525 Chr.17: 82255858-82255858 on Build GRCh38
rs770868697 Chr.17: 82255531-82255531 on Build GRCh38
rs1035405140 Chr.17: 82255679-82255679 on Build GRCh38
rs766145837 Chr.17: 82255603-82255604 on Build GRCh38
rs917562284 Chr.17: 82255699-82255699 on Build GRCh38
rs919536486 Chr.17: 82255781-82255781 on Build GRCh38
rs370494013 Chr.17: 82255616-82255616 on Build GRCh38
rs1052367647 Chr.17: 82255815-82255815 on Build GRCh38
rs968847040 Chr.17: 82255656-82255656 on Build GRCh38
rs781744472 Chr.17: 82255450-82255450 on Build GRCh38
rs1000324183 Chr.17: 82255785-82255785 on Build GRCh38
rs1034446923 Chr.17: 82255791-82255791 on Build GRCh38
rs540581308 Chr.17: 82255782-82255782 on Build GRCh38
rs756570635 Chr.17: 82255834-82255834 on Build GRCh38
rs770819449 Chr.17: 82255402-82255402 on Build GRCh38
rs910615271 Chr.17: 82255817-82255817 on Build GRCh38
rs759730221 Chr.17: 82255541-82255541 on Build GRCh38
rs745497844 Chr.17: 82255404-82255405 on Build GRCh38
rs889207780 Chr.17: 82255401-82255401 on Build GRCh38
rs1035943607 Chr.17: 82255868-82255868 on Build GRCh38
rs1010474784 Chr.17: 82255767-82255767 on Build GRCh38
rs773186560 Chr.17: 82255381-82255381 on Build GRCh38
rs774889675 Chr.17: 82255413-82255413 on Build GRCh38
rs186931747 Chr.17: 82255452-82255452 on Build GRCh38
rs1009612562 Chr.17: 82255833-82255833 on Build GRCh38
rs776484545 Chr.17: 82255540-82255540 on Build GRCh38
rs893795620 Chr.17: 82255829-82255829 on Build GRCh38
rs931125903 Chr.17: 82255717-82255717 on Build GRCh38
rs149005084 Chr.17: 82255873-82255873 on Build GRCh38
rs764529977 Chr.17: 82255421-82255421 on Build GRCh38
rs752859448 Chr.17: 82255410-82255410 on Build GRCh38
rs564627454 Chr.17: 82255705-82255705 on Build GRCh38
rs901148805 Chr.17: 82255758-82255758 on Build GRCh38
rs1007527113 Chr.17: 82255414-82255414 on Build GRCh38
rs1006939010 Chr.17: 82255743-82255743 on Build GRCh38
rs778690634 Chr.17: 82255471-82255471 on Build GRCh38
rs759717634 Chr.17: 82255371-82255371 on Build GRCh38
rs1041598502 Chr.17: 82255849-82255849 on Build GRCh38
rs967236793 Chr.17: 82255530-82255530 on Build GRCh38
rs917598520 Chr.17: 82255827-82255827 on Build GRCh38
rs764625001 Chr.17: 82255624-82255624 on Build GRCh38
rs958716597 Chr.17: 82255680-82255680 on Build GRCh38
rs773786515 Chr.17: 82255620-82255621 on Build GRCh38
rs769750217 Chr.17: 82255551-82255551 on Build GRCh38
rs763431194 Chr.17: 82255412-82255412 on Build GRCh38
rs1053903021 Chr.17: 82255823-82255823 on Build GRCh38
rs765502737 Chr.17: 82255409-82255409 on Build GRCh38
rs754568292 Chr.17: 82255619-82255619 on Build GRCh38
rs1010191636 Chr.17: 82255876-82255877 on Build GRCh38
rs933524353 Chr.17: 82255387-82255387 on Build GRCh38
rs752151724 Chr.17: 82255426-82255426 on Build GRCh38
rs942299294 Chr.17: 82255370-82255370 on Build GRCh38
rs750128193 Chr.17: 82255416-82255416 on Build GRCh38
rs116794634 Chr.17: 82255875-82255875 on Build GRCh38
rs754377044 Chr.17: 82255459-82255459 on Build GRCh38
rs762206641 Chr.17: 82255610-82255610 on Build GRCh38
rs1036932824 Chr.17: 82255711-82255711 on Build GRCh38
rs546740896 Chr.17: 82255857-82255857 on Build GRCh38
rs896968522 Chr.17: 82255393-82255393 on Build GRCh38
rs925033422 Chr.17: 82255675-82255675 on Build GRCh38
rs563266748 Chr.17: 82255644-82255644 on Build GRCh38
rs746760351 Chr.17: 82255494-82255494 on Build GRCh38
rs763052778 Chr.17: 82255558-82255558 on Build GRCh38
rs11552086 Chr.17: 82255552-82255552 on Build GRCh38
rs1027056874 Chr.17: 82255465-82255465 on Build GRCh38
rs764961361 Chr.17: 82255331-82255331 on Build GRCh38

View Full Product Details