Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35793174-35793707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK1
Entrez Gene ID
9201
Gene Name
doublecortin like kinase 1
Gene Aliases
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK
UniGene
Hs.507755
Gene Chromosome Location
Chr.13: 35768652-36132020 on Build GRCh38
Transcripts
NM_001195415.1 NM_001195416.1 NM_004734.4 XM_005266592.2 XM_006719893.2 XM_017020847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249793 c.1945-2A>C p.? Chr.13: 35793481-35793481 on Build GRCh38
5893722 c.1102C>T p.P368S Chr.13: 35793401-35793401 on Build GRCh38
5893723 c.2023C>T p.P675S Chr.13: 35793401-35793401 on Build GRCh38
4047284 c.1064C>G p.A355G Chr.13: 35793439-35793439 on Build GRCh38
6253150 c.2024C>A p.P675Q Chr.13: 35793400-35793400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761282174 Chr.13: 35793478-35793478 on Build GRCh38
rs749998565 Chr.13: 35793494-35793494 on Build GRCh38
rs368749570 Chr.13: 35793541-35793541 on Build GRCh38
rs781374868 Chr.13: 35793502-35793502 on Build GRCh38
rs756375140 Chr.13: 35793625-35793625 on Build GRCh38
rs549979203 Chr.13: 35793326-35793326 on Build GRCh38
rs181860743 Chr.13: 35793399-35793399 on Build GRCh38
rs566488208 Chr.13: 35793316-35793316 on Build GRCh38
rs1055989115 Chr.13: 35793307-35793307 on Build GRCh38
rs117067436 Chr.13: 35793665-35793665 on Build GRCh38
rs149526371 Chr.13: 35793232-35793232 on Build GRCh38
rs750207795 Chr.13: 35793339-35793339 on Build GRCh38
rs531747698 Chr.13: 35793268-35793268 on Build GRCh38
rs564283780 Chr.13: 35793257-35793257 on Build GRCh38
rs1018135787 Chr.13: 35793513-35793513 on Build GRCh38
rs1010017950 Chr.13: 35793245-35793245 on Build GRCh38
rs935289961 Chr.13: 35793266-35793266 on Build GRCh38
rs755359793 Chr.13: 35793221-35793221 on Build GRCh38
rs146043148 Chr.13: 35793467-35793467 on Build GRCh38
rs950132374 Chr.13: 35793333-35793333 on Build GRCh38
rs1034107742 Chr.13: 35793314-35793314 on Build GRCh38
rs773822732 Chr.13: 35793473-35793473 on Build GRCh38
rs891553383 Chr.13: 35793623-35793623 on Build GRCh38
rs755814838 Chr.13: 35793349-35793349 on Build GRCh38
rs891671287 Chr.13: 35793242-35793242 on Build GRCh38
rs373569186 Chr.13: 35793249-35793249 on Build GRCh38
rs959398710 Chr.13: 35793673-35793673 on Build GRCh38
rs756388992 Chr.13: 35793518-35793518 on Build GRCh38
rs1001432654 Chr.13: 35793302-35793302 on Build GRCh38
rs910913476 Chr.13: 35793585-35793585 on Build GRCh38
rs775847658 Chr.13: 35793460-35793460 on Build GRCh38
rs931569724 Chr.13: 35793537-35793537 on Build GRCh38
rs547235201 Chr.13: 35793459-35793459 on Build GRCh38
rs746020830 Chr.13: 35793517-35793517 on Build GRCh38
rs1040808387 Chr.13: 35793614-35793614 on Build GRCh38
rs529067405 Chr.13: 35793452-35793452 on Build GRCh38
rs781338577 Chr.13: 35793506-35793506 on Build GRCh38
rs558193525 Chr.13: 35793642-35793642 on Build GRCh38
rs112083991 Chr.13: 35793280-35793280 on Build GRCh38
rs539305195 Chr.13: 35793491-35793491 on Build GRCh38
rs780395198 Chr.13: 35793524-35793524 on Build GRCh38
rs763724666 Chr.13: 35793485-35793486 on Build GRCh38
rs377145626 Chr.13: 35793495-35793495 on Build GRCh38
rs372376341 Chr.13: 35793443-35793443 on Build GRCh38
rs894536867 Chr.13: 35793309-35793309 on Build GRCh38
rs573723385 Chr.13: 35793655-35793655 on Build GRCh38
rs555156022 Chr.13: 35793651-35793651 on Build GRCh38
rs1021423113 Chr.13: 35793272-35793272 on Build GRCh38
rs1926459 Chr.13: 35793504-35793504 on Build GRCh38
rs771151820 Chr.13: 35793424-35793424 on Build GRCh38
rs767118101 Chr.13: 35793320-35793320 on Build GRCh38
rs763441974 Chr.13: 35793466-35793466 on Build GRCh38
rs1007963984 Chr.13: 35793225-35793225 on Build GRCh38
rs941505759 Chr.13: 35793264-35793264 on Build GRCh38
rs191002717 Chr.13: 35793199-35793199 on Build GRCh38
rs368298500 Chr.13: 35793500-35793500 on Build GRCh38
rs780022912 Chr.13: 35793395-35793395 on Build GRCh38
rs537080353 Chr.13: 35793600-35793600 on Build GRCh38
rs1043298465 Chr.13: 35793644-35793644 on Build GRCh38
rs754979350 Chr.13: 35793401-35793401 on Build GRCh38
rs747122620 Chr.13: 35793409-35793409 on Build GRCh38
rs969040262 Chr.13: 35793274-35793274 on Build GRCh38

View Full Product Details