Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 33224530-33225013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
UniGene
Hs.115461
Gene Chromosome Location
Chr.18: 32937402-33441040 on Build GRCh38
Transcripts
NM_001105528.1 NM_001308126.1 NM_198995.2 XM_011525948.1 XM_011525951.1 XM_017025721.1 XM_017025722.1 XM_017025723.1 XM_017025724.1 XM_017025725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6502720 c.1771G>C p.E591Q Chr.18: 33224822-33224822 on Build GRCh38
3525363 c.1697G>A p.R566Q Chr.18: 33224896-33224896 on Build GRCh38
3525361 c.1730C>T p.A577V Chr.18: 33224863-33224863 on Build GRCh38
6083041 c.1724T>C p.I575T Chr.18: 33224869-33224869 on Build GRCh38
1303697 c.1657-1G>C p.? Chr.18: 33224937-33224937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141176123 Chr.18: 33224928-33224928 on Build GRCh38
rs764013511 Chr.18: 33224889-33224889 on Build GRCh38
rs1042943674 Chr.18: 33224723-33224723 on Build GRCh38
rs762291735 Chr.18: 33224751-33224751 on Build GRCh38
rs457896 Chr.18: 33224794-33224794 on Build GRCh38
rs113589354 Chr.18: 33224795-33224796 on Build GRCh38
rs768290189 Chr.18: 33224816-33224816 on Build GRCh38
rs777122360 Chr.18: 33224856-33224856 on Build GRCh38
rs779887543 Chr.18: 33224901-33224901 on Build GRCh38
rs776190666 Chr.18: 33224729-33224729 on Build GRCh38
rs271429 Chr.18: 33224640-33224640 on Build GRCh38
rs576466204 Chr.18: 33224674-33224674 on Build GRCh38
rs973737590 Chr.18: 33224560-33224560 on Build GRCh38
rs765706382 Chr.18: 33224930-33224931 on Build GRCh38
rs112779048 Chr.18: 33224896-33224896 on Build GRCh38
rs386802235 Chr.18: 33224792-33224794 on Build GRCh38
rs754677102 Chr.18: 33224911-33224911 on Build GRCh38
rs778651870 Chr.18: 33224915-33224915 on Build GRCh38
rs759906598 Chr.18: 33224866-33224866 on Build GRCh38
rs922947433 Chr.18: 33224699-33224699 on Build GRCh38
rs747718980 Chr.18: 33224916-33224916 on Build GRCh38
rs984628752 Chr.18: 33224615-33224615 on Build GRCh38
rs768039911 Chr.18: 33224753-33224753 on Build GRCh38
rs934202476 Chr.18: 33224709-33224709 on Build GRCh38
rs1000038821 Chr.18: 33224743-33224743 on Build GRCh38
rs775154383 Chr.18: 33224734-33224734 on Build GRCh38
rs757189280 Chr.18: 33224793-33224793 on Build GRCh38
rs372662327 Chr.18: 33224963-33224963 on Build GRCh38
rs759944357 Chr.18: 33224925-33224928 on Build GRCh38
rs916380666 Chr.18: 33224569-33224569 on Build GRCh38
rs61735129 Chr.18: 33224795-33224795 on Build GRCh38
rs763456731 Chr.18: 33224906-33224906 on Build GRCh38
rs200469422 Chr.18: 33224672-33224672 on Build GRCh38
rs542359437 Chr.18: 33224684-33224684 on Build GRCh38
rs779996608 Chr.18: 33224611-33224611 on Build GRCh38
rs951884323 Chr.18: 33224583-33224583 on Build GRCh38
rs549403400 Chr.18: 33224931-33224931 on Build GRCh38
rs76742118 Chr.18: 33224641-33224641 on Build GRCh38
rs143411483 Chr.18: 33224663-33224663 on Build GRCh38
rs761128297 Chr.18: 33224772-33224772 on Build GRCh38
rs771619759 Chr.18: 33224926-33224926 on Build GRCh38
rs932294523 Chr.18: 33224653-33224653 on Build GRCh38
rs144365664 Chr.18: 33224875-33224875 on Build GRCh38
rs762524624 Chr.18: 33224736-33224736 on Build GRCh38
rs777177324 Chr.18: 33224947-33224947 on Build GRCh38
rs139736043 Chr.18: 33224897-33224897 on Build GRCh38
rs61734218 Chr.18: 33224823-33224823 on Build GRCh38
rs1011074726 Chr.18: 33224724-33224724 on Build GRCh38
rs748994198 Chr.18: 33224814-33224814 on Build GRCh38
rs572714033 Chr.18: 33224716-33224716 on Build GRCh38
rs768165219 Chr.18: 33224744-33224744 on Build GRCh38
rs536205887 Chr.18: 33224555-33224555 on Build GRCh38
rs766814350 Chr.18: 33224774-33224774 on Build GRCh38
rs745651862 Chr.18: 33224727-33224727 on Build GRCh38
rs776967776 Chr.18: 33224917-33224918 on Build GRCh38
rs547922194 Chr.18: 33224853-33224853 on Build GRCh38
rs466113 Chr.18: 33224792-33224792 on Build GRCh38
rs753180261 Chr.18: 33224931-33224931 on Build GRCh38
rs1040021084 Chr.18: 33224668-33224668 on Build GRCh38
rs758534698 Chr.18: 33224877-33224877 on Build GRCh38
rs539468486 Chr.18: 33224645-33224645 on Build GRCh38
rs564793450 Chr.18: 33224791-33224791 on Build GRCh38
rs562260372 Chr.18: 33224714-33224714 on Build GRCh38
rs779763134 Chr.18: 33224802-33224802 on Build GRCh38
rs771562844 Chr.18: 33224943-33224943 on Build GRCh38
rs533824961 Chr.18: 33224572-33224572 on Build GRCh38
rs370103065 Chr.18: 33224754-33224754 on Build GRCh38
rs747675216 Chr.18: 33224831-33224831 on Build GRCh38
rs766863605 Chr.18: 33224851-33224851 on Build GRCh38
rs77014111 Chr.18: 33224717-33224717 on Build GRCh38
rs761024594 Chr.18: 33224846-33224846 on Build GRCh38
rs755194112 Chr.18: 33224790-33224790 on Build GRCh38
rs907610649 Chr.18: 33224626-33224626 on Build GRCh38
rs771437193 Chr.18: 33224837-33224837 on Build GRCh38
rs773803908 Chr.18: 33224843-33224843 on Build GRCh38
rs567960141 Chr.18: 33224568-33224568 on Build GRCh38

View Full Product Details