Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17763948-17764525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
UniGene
Hs.96485
Gene Chromosome Location
Chr.19: 17747718-17788568 on Build GRCh38
Transcripts
NM_001161357.1 NM_001161358.1 NM_001161359.1 NM_015122.2 XM_005259831.3 XM_006722701.3 XM_006722702.3 XM_011527824.2 XM_011527825.2 XM_011527826.2 XM_011527827.2 XM_011527828.2 XM_017026519.1 XM_017026520.1 XM_017026521.1 XM_017026522.1 XM_017026523.1 XM_017026524.1 XM_017026525.1 XM_017026526.1 XM_017026527.1 XM_017026528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611796 c.180C>T p.N60N Chr.19: 17764435-17764435 on Build GRCh38
711189 c.125C>A p.T42N Chr.19: 17764380-17764380 on Build GRCh38
7079332 c.130G>A p.E44K Chr.19: 17764385-17764385 on Build GRCh38
7279696 c.150G>A p.A50A Chr.19: 17764405-17764405 on Build GRCh38
3530555 c.156G>A p.A52A Chr.19: 17764411-17764411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776461187 Chr.19: 17764474-17764474 on Build GRCh38
rs773739214 Chr.19: 17764336-17764337 on Build GRCh38
rs145154295 Chr.19: 17764424-17764424 on Build GRCh38
rs751201281 Chr.19: 17764429-17764429 on Build GRCh38
rs752191239 Chr.19: 17764423-17764423 on Build GRCh38
rs573351971 Chr.19: 17764116-17764116 on Build GRCh38
rs1039264352 Chr.19: 17764128-17764128 on Build GRCh38
rs571603619 Chr.19: 17763978-17763978 on Build GRCh38
rs775480992 Chr.19: 17764364-17764364 on Build GRCh38
rs148702519 Chr.19: 17764165-17764165 on Build GRCh38
rs748433915 Chr.19: 17764470-17764470 on Build GRCh38
rs752242567 Chr.19: 17764333-17764333 on Build GRCh38
rs11458516 Chr.19: 17764045-17764046 on Build GRCh38
rs974333892 Chr.19: 17764272-17764272 on Build GRCh38
rs1039637391 Chr.19: 17764365-17764365 on Build GRCh38
rs571002372 Chr.19: 17764158-17764158 on Build GRCh38
rs981097248 Chr.19: 17764284-17764284 on Build GRCh38
rs982569429 Chr.19: 17763974-17763974 on Build GRCh38
rs541585673 Chr.19: 17764266-17764266 on Build GRCh38
rs199537461 Chr.19: 17764405-17764405 on Build GRCh38
rs780568471 Chr.19: 17764458-17764458 on Build GRCh38
rs747410983 Chr.19: 17764340-17764340 on Build GRCh38
rs754484609 Chr.19: 17764331-17764331 on Build GRCh38
rs995338446 Chr.19: 17764469-17764469 on Build GRCh38
rs986406047 Chr.19: 17764316-17764316 on Build GRCh38
rs377559160 Chr.19: 17764337-17764337 on Build GRCh38
rs148025294 Chr.19: 17764411-17764411 on Build GRCh38
rs760473923 Chr.19: 17764399-17764399 on Build GRCh38
rs199720023 Chr.19: 17764463-17764463 on Build GRCh38
rs142054668 Chr.19: 17764465-17764465 on Build GRCh38
rs746311456 Chr.19: 17764354-17764354 on Build GRCh38
rs951918529 Chr.19: 17764199-17764199 on Build GRCh38
rs907138370 Chr.19: 17764155-17764155 on Build GRCh38
rs934322309 Chr.19: 17764325-17764325 on Build GRCh38
rs141727484 Chr.19: 17764435-17764435 on Build GRCh38
rs760418831 Chr.19: 17764326-17764326 on Build GRCh38
rs928357104 Chr.19: 17764093-17764093 on Build GRCh38
rs7256565 Chr.19: 17764214-17764214 on Build GRCh38
rs572547540 Chr.19: 17764224-17764224 on Build GRCh38
rs753712854 Chr.19: 17764330-17764330 on Build GRCh38
rs747956233 Chr.19: 17764390-17764390 on Build GRCh38
rs973984962 Chr.19: 17764021-17764021 on Build GRCh38
rs936805859 Chr.19: 17764006-17764006 on Build GRCh38
rs553459609 Chr.19: 17764496-17764496 on Build GRCh38
rs772271019 Chr.19: 17764471-17764471 on Build GRCh38
rs147597811 Chr.19: 17764457-17764457 on Build GRCh38
rs757648231 Chr.19: 17764349-17764349 on Build GRCh38
rs766322957 Chr.19: 17764328-17764328 on Build GRCh38
rs138999727 Chr.19: 17764283-17764283 on Build GRCh38
rs759209210 Chr.19: 17764417-17764442 on Build GRCh38
rs928326017 Chr.19: 17764287-17764287 on Build GRCh38
rs200418103 Chr.19: 17764242-17764242 on Build GRCh38
rs774580596 Chr.19: 17764398-17764398 on Build GRCh38
rs915829874 Chr.19: 17764033-17764033 on Build GRCh38
rs893270207 Chr.19: 17764117-17764117 on Build GRCh38
rs769578227 Chr.19: 17764439-17764440 on Build GRCh38
rs749354985 Chr.19: 17764367-17764367 on Build GRCh38
rs962549496 Chr.19: 17764267-17764267 on Build GRCh38
rs759300421 Chr.19: 17764482-17764482 on Build GRCh38
rs200177505 Chr.19: 17764350-17764350 on Build GRCh38
rs770319954 Chr.19: 17764358-17764358 on Build GRCh38
rs953061953 Chr.19: 17764285-17764285 on Build GRCh38
rs557478200 Chr.19: 17764070-17764070 on Build GRCh38
rs150720896 Chr.19: 17764279-17764279 on Build GRCh38
rs556778709 Chr.19: 17764157-17764157 on Build GRCh38
rs200398288 Chr.19: 17764369-17764369 on Build GRCh38
rs1013598396 Chr.19: 17764092-17764092 on Build GRCh38
rs934432979 Chr.19: 17764103-17764103 on Build GRCh38
rs757701170 Chr.19: 17764432-17764432 on Build GRCh38
rs201119223 Chr.19: 17764332-17764332 on Build GRCh38
rs536539991 Chr.19: 17764124-17764124 on Build GRCh38
rs1014634785 Chr.19: 17764230-17764230 on Build GRCh38
rs962865581 Chr.19: 17764017-17764017 on Build GRCh38
rs377654716 Chr.19: 17764460-17764460 on Build GRCh38
rs145996010 Chr.19: 17764037-17764037 on Build GRCh38
rs544951040 Chr.19: 17764044-17764045 on Build GRCh38
rs553331889 Chr.19: 17764081-17764081 on Build GRCh38
rs771736285 Chr.19: 17764436-17764436 on Build GRCh38
rs900771910 Chr.19: 17764132-17764132 on Build GRCh38
rs551538160 Chr.19: 17764410-17764410 on Build GRCh38
rs182807069 Chr.19: 17764336-17764336 on Build GRCh38
rs765203913 Chr.19: 17764418-17764418 on Build GRCh38

View Full Product Details