Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49630516-49631063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL11
Entrez Gene ID
115761
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
ARLTS1
UniGene
Hs.558599
Gene Chromosome Location
Chr.13: 49628299-49633872 on Build GRCh38
Transcripts
NM_138450.5 XM_005266253.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293285 c.198G>A p.G66G Chr.13: 49630645-49630645 on Build GRCh38
6139080 c.252C>A p.L84L Chr.13: 49630699-49630699 on Build GRCh38
1942720 c.526C>T p.R176C Chr.13: 49630973-49630973 on Build GRCh38
5971004 c.165G>T p.G55G Chr.13: 49630612-49630612 on Build GRCh38
295257 c.216C>A p.A72A Chr.13: 49630663-49630663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769095684 Chr.13: 49630739-49630739 on Build GRCh38
rs781514079 Chr.13: 49630728-49630728 on Build GRCh38
rs755100942 Chr.13: 49630894-49630894 on Build GRCh38
rs374531869 Chr.13: 49630700-49630700 on Build GRCh38
rs759152923 Chr.13: 49630854-49630854 on Build GRCh38
rs747335286 Chr.13: 49630916-49630916 on Build GRCh38
rs867820824 Chr.13: 49630712-49630712 on Build GRCh38
rs775860107 Chr.13: 49630568-49630568 on Build GRCh38
rs756928549 Chr.13: 49630589-49630589 on Build GRCh38
rs370030859 Chr.13: 49630706-49630706 on Build GRCh38
rs923965064 Chr.13: 49630872-49630872 on Build GRCh38
rs201969290 Chr.13: 49630566-49630566 on Build GRCh38
rs377295532 Chr.13: 49630705-49630705 on Build GRCh38
rs574095059 Chr.13: 49631016-49631016 on Build GRCh38
rs115070474 Chr.13: 49630995-49630995 on Build GRCh38
rs745519193 Chr.13: 49631001-49631001 on Build GRCh38
rs778849305 Chr.13: 49630901-49630901 on Build GRCh38
rs763888661 Chr.13: 49630952-49630952 on Build GRCh38
rs569508133 Chr.13: 49630936-49630936 on Build GRCh38
rs866872755 Chr.13: 49630827-49630827 on Build GRCh38
rs377089928 Chr.13: 49630558-49630558 on Build GRCh38
rs768182542 Chr.13: 49631005-49631005 on Build GRCh38
rs780120181 Chr.13: 49630802-49630802 on Build GRCh38
rs911644636 Chr.13: 49630941-49630941 on Build GRCh38
rs753334860 Chr.13: 49630772-49630772 on Build GRCh38
rs777416619 Chr.13: 49630784-49630784 on Build GRCh38
rs115584350 Chr.13: 49631003-49631003 on Build GRCh38
rs758972109 Chr.13: 49630991-49630991 on Build GRCh38
rs748581908 Chr.13: 49630833-49630833 on Build GRCh38
rs147120792 Chr.13: 49630839-49630839 on Build GRCh38
rs758766792 Chr.13: 49630907-49630907 on Build GRCh38
rs138452698 Chr.13: 49630843-49630843 on Build GRCh38
rs751003114 Chr.13: 49630840-49630840 on Build GRCh38
rs113027020 Chr.13: 49630653-49630653 on Build GRCh38
rs145617860 Chr.13: 49630699-49630699 on Build GRCh38
rs117251022 Chr.13: 49630641-49630641 on Build GRCh38
rs779514414 Chr.13: 49630832-49630832 on Build GRCh38
rs747926100 Chr.13: 49630719-49630719 on Build GRCh38
rs771389167 Chr.13: 49630921-49630921 on Build GRCh38
rs562619284 Chr.13: 49631018-49631018 on Build GRCh38
rs773042047 Chr.13: 49630702-49630702 on Build GRCh38
rs3803186 Chr.13: 49630744-49630744 on Build GRCh38
rs927795583 Chr.13: 49630696-49630696 on Build GRCh38
rs771123171 Chr.13: 49630652-49630652 on Build GRCh38
rs1051854026 Chr.13: 49630748-49630752 on Build GRCh38
rs748760701 Chr.13: 49630904-49630904 on Build GRCh38
rs958511445 Chr.13: 49630628-49630628 on Build GRCh38
rs759972408 Chr.13: 49630564-49630564 on Build GRCh38
rs766738083 Chr.13: 49630863-49630863 on Build GRCh38
rs772969476 Chr.13: 49630747-49630747 on Build GRCh38
rs762794413 Chr.13: 49630871-49630871 on Build GRCh38
rs776309613 Chr.13: 49630848-49630848 on Build GRCh38
rs764419423 Chr.13: 49630573-49630573 on Build GRCh38
rs761636757 Chr.13: 49630963-49630963 on Build GRCh38
rs759566544 Chr.13: 49630773-49630773 on Build GRCh38
rs769272757 Chr.13: 49631002-49631002 on Build GRCh38
rs745498908 Chr.13: 49630934-49630934 on Build GRCh38
rs763346893 Chr.13: 49630571-49630571 on Build GRCh38
rs766056897 Chr.13: 49630771-49630771 on Build GRCh38
rs994780789 Chr.13: 49630790-49630790 on Build GRCh38
rs34301344 Chr.13: 49630893-49630893 on Build GRCh38
rs143660006 Chr.13: 49630637-49630637 on Build GRCh38
rs35363768 Chr.13: 49631017-49631017 on Build GRCh38
rs778216279 Chr.13: 49630914-49630914 on Build GRCh38
rs551289381 Chr.13: 49630807-49630807 on Build GRCh38
rs146850453 Chr.13: 49630937-49630937 on Build GRCh38
rs35835937 Chr.13: 49631029-49631029 on Build GRCh38
rs577463512 Chr.13: 49631021-49631021 on Build GRCh38
rs767279791 Chr.13: 49630973-49630973 on Build GRCh38
rs769184018 Chr.13: 49630823-49630823 on Build GRCh38
rs772293845 Chr.13: 49630746-49630746 on Build GRCh38
rs1031176774 Chr.13: 49630822-49630822 on Build GRCh38
rs747265258 Chr.13: 49630650-49630650 on Build GRCh38
rs114223009 Chr.13: 49631004-49631004 on Build GRCh38
rs140526352 Chr.13: 49630974-49630974 on Build GRCh38
rs369494120 Chr.13: 49630615-49630615 on Build GRCh38
rs763436324 Chr.13: 49630677-49630677 on Build GRCh38
rs200112422 Chr.13: 49631022-49631022 on Build GRCh38
rs774163543 Chr.13: 49630955-49630955 on Build GRCh38
rs762961641 Chr.13: 49630946-49630946 on Build GRCh38
rs780101758 Chr.13: 49630738-49630738 on Build GRCh38
rs567358654 Chr.13: 49630891-49630891 on Build GRCh38
rs996300560 Chr.13: 49630604-49630604 on Build GRCh38
rs915492243 Chr.13: 49630902-49630902 on Build GRCh38
rs770655477 Chr.13: 49630844-49630844 on Build GRCh38
rs757752049 Chr.13: 49630996-49630996 on Build GRCh38
rs139544934 Chr.13: 49630636-49630636 on Build GRCh38
rs756352209 Chr.13: 49630882-49630882 on Build GRCh38
rs764627150 Chr.13: 49630858-49630858 on Build GRCh38
rs752542486 Chr.13: 49630778-49630778 on Build GRCh38
rs761431152 Chr.13: 49630781-49630781 on Build GRCh38
rs576032054 Chr.13: 49630536-49630536 on Build GRCh38
rs781685676 Chr.13: 49630998-49630998 on Build GRCh38
rs776025417 Chr.13: 49630668-49630668 on Build GRCh38
rs867095637 Chr.13: 49630895-49630895 on Build GRCh38
rs3803185 Chr.13: 49630889-49630889 on Build GRCh38
rs752737224 Chr.13: 49630859-49630859 on Build GRCh38
rs551214371 Chr.13: 49630927-49630927 on Build GRCh38
rs369303916 Chr.13: 49631015-49631015 on Build GRCh38
rs763952974 Chr.13: 49630878-49630878 on Build GRCh38
rs778840264 Chr.13: 49630643-49630643 on Build GRCh38
rs765365695 Chr.13: 49630982-49630982 on Build GRCh38
rs751269133 Chr.13: 49630588-49630588 on Build GRCh38
rs751295835 Chr.13: 49630880-49630880 on Build GRCh38
rs759584416 Chr.13: 49630658-49630659 on Build GRCh38
rs532945151 Chr.13: 49630775-49630775 on Build GRCh38
rs149745507 Chr.13: 49630679-49630679 on Build GRCh38
rs769920049 Chr.13: 49630731-49630731 on Build GRCh38
rs970805802 Chr.13: 49630856-49630856 on Build GRCh38
rs200663291 Chr.13: 49630718-49630718 on Build GRCh38
rs756260176 Chr.13: 49630795-49630795 on Build GRCh38
rs757447757 Chr.13: 49630721-49630721 on Build GRCh38
rs761386601 Chr.13: 49630682-49630682 on Build GRCh38
rs749892930 Chr.13: 49630616-49630616 on Build GRCh38
rs776703350 Chr.13: 49630547-49630547 on Build GRCh38
rs752665403 Chr.13: 49630983-49630983 on Build GRCh38
rs868285879 Chr.13: 49630808-49630808 on Build GRCh38
rs760395038 Chr.13: 49630765-49630765 on Build GRCh38
rs544417615 Chr.13: 49630730-49630730 on Build GRCh38
rs35712316 Chr.13: 49630805-49630805 on Build GRCh38
rs201132357 Chr.13: 49630542-49630542 on Build GRCh38
rs745917227 Chr.13: 49630654-49630654 on Build GRCh38
rs767150266 Chr.13: 49630685-49630685 on Build GRCh38
rs769830175 Chr.13: 49630666-49630666 on Build GRCh38
rs765332310 Chr.13: 49631025-49631025 on Build GRCh38
rs750723672 Chr.13: 49630785-49630785 on Build GRCh38
rs774603360 Chr.13: 49630743-49630743 on Build GRCh38

View Full Product Details