Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55349253-55349805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6B2
Entrez Gene ID
125965
Gene Name
cytochrome c oxidase subunit 6B2
Gene Aliases
COXVIB2, CT59
UniGene
Hs.550544
Gene Chromosome Location
Chr.19: 55349702-55354814 on Build GRCh38
Transcripts
NM_144613.4

SNP Detail

SNP ID Chromosome Location
rs913466433 Chr.19: 55349662-55349662 on Build GRCh38
rs928728511 Chr.19: 55349760-55349760 on Build GRCh38
rs894892000 Chr.19: 55349738-55349738 on Build GRCh38
rs933414141 Chr.19: 55349344-55349344 on Build GRCh38
rs538064114 Chr.19: 55349391-55349391 on Build GRCh38
rs1045816541 Chr.19: 55349435-55349435 on Build GRCh38
rs771478760 Chr.19: 55349540-55349540 on Build GRCh38
rs937390083 Chr.19: 55349474-55349474 on Build GRCh38
rs1055814273 Chr.19: 55349778-55349778 on Build GRCh38
rs796912266 Chr.19: 55349564-55349564 on Build GRCh38
rs967555989 Chr.19: 55349664-55349664 on Build GRCh38
rs959004525 Chr.19: 55349284-55349294 on Build GRCh38
rs981565320 Chr.19: 55349676-55349676 on Build GRCh38
rs760815893 Chr.19: 55349283-55349284 on Build GRCh38
rs990351094 Chr.19: 55349350-55349350 on Build GRCh38
rs1022385389 Chr.19: 55349772-55349772 on Build GRCh38
rs899410570 Chr.19: 55349477-55349477 on Build GRCh38
rs779636071 Chr.19: 55349625-55349625 on Build GRCh38
rs946204991 Chr.19: 55349432-55349432 on Build GRCh38
rs530211282 Chr.19: 55349456-55349456 on Build GRCh38
rs57934925 Chr.19: 55349623-55349623 on Build GRCh38
rs990834503 Chr.19: 55349640-55349640 on Build GRCh38
rs922441831 Chr.19: 55349571-55349571 on Build GRCh38
rs1038099349 Chr.19: 55349621-55349621 on Build GRCh38
rs567792300 Chr.19: 55349414-55349414 on Build GRCh38
rs987529321 Chr.19: 55349575-55349575 on Build GRCh38
rs937401786 Chr.19: 55349770-55349770 on Build GRCh38
rs185590150 Chr.19: 55349521-55349521 on Build GRCh38
rs977999545 Chr.19: 55349569-55349569 on Build GRCh38
rs190441260 Chr.19: 55349549-55349549 on Build GRCh38
rs1047986883 Chr.19: 55349646-55349646 on Build GRCh38
rs759674347 Chr.19: 55349479-55349479 on Build GRCh38
rs936210577 Chr.19: 55349343-55349343 on Build GRCh38
rs111466436 Chr.19: 55349275-55349275 on Build GRCh38
rs893080119 Chr.19: 55349278-55349278 on Build GRCh38
rs928718617 Chr.19: 55349471-55349471 on Build GRCh38
rs386389288 Chr.19: 55349303-55349304 on Build GRCh38
rs111872256 Chr.19: 55349279-55349279 on Build GRCh38
rs1022249242 Chr.19: 55349554-55349554 on Build GRCh38
rs915131597 Chr.19: 55349292-55349292 on Build GRCh38
rs978325910 Chr.19: 55349314-55349314 on Build GRCh38
rs761307209 Chr.19: 55349704-55349704 on Build GRCh38
rs869079965 Chr.19: 55349305-55349306 on Build GRCh38
rs995135196 Chr.19: 55349492-55349492 on Build GRCh38
rs529536209 Chr.19: 55349485-55349485 on Build GRCh38
rs910623619 Chr.19: 55349578-55349578 on Build GRCh38
rs146959385 Chr.19: 55349638-55349638 on Build GRCh38
rs33929957 Chr.19: 55349305-55349305 on Build GRCh38
rs910571417 Chr.19: 55349399-55349399 on Build GRCh38
rs1054907303 Chr.19: 55349731-55349731 on Build GRCh38
rs966590349 Chr.19: 55349313-55349313 on Build GRCh38
rs943335624 Chr.19: 55349429-55349429 on Build GRCh38
rs868275448 Chr.19: 55349572-55349572 on Build GRCh38
rs995858765 Chr.19: 55349573-55349573 on Build GRCh38
rs569171583 Chr.19: 55349641-55349641 on Build GRCh38
rs886297431 Chr.19: 55349503-55349503 on Build GRCh38
rs989428608 Chr.19: 55349284-55349296 on Build GRCh38
rs922216722 Chr.19: 55349274-55349274 on Build GRCh38
rs548042480 Chr.19: 55349502-55349502 on Build GRCh38
rs1029052094 Chr.19: 55349579-55349579 on Build GRCh38
rs924730713 Chr.19: 55349318-55349318 on Build GRCh38
rs1018740409 Chr.19: 55349558-55349558 on Build GRCh38

View Full Product Details