Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132594503-132595075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX31
Entrez Gene ID
64794
Gene Name
DEAD-box helicase 31
Gene Aliases
PPP1R25
UniGene
Hs.660767
Gene Chromosome Location
Chr.9: 132592997-132670403 on Build GRCh38
Transcripts
NM_001322340.1 NM_001322341.1 NM_001322342.1 NM_001322343.1 NM_022779.8 NR_136309.1 XM_005272207.4 XM_006717236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641717 c.2366C>T p.S789L Chr.9: 132595056-132595056 on Build GRCh38
4163463 c.2395A>G p.I799V Chr.9: 132595027-132595027 on Build GRCh38
6641718 c.2534G>T p.S845I Chr.9: 132594888-132594888 on Build GRCh38
5812437 c.2455A>T p.S819C Chr.9: 132594967-132594967 on Build GRCh38
3212612 c.2552T>C p.V851A Chr.9: 132594870-132594870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911787129 Chr.9: 132594648-132594648 on Build GRCh38
rs1045920652 Chr.9: 132594607-132594607 on Build GRCh38
rs1029537432 Chr.9: 132594771-132594771 on Build GRCh38
rs968128525 Chr.9: 132594580-132594580 on Build GRCh38
rs911833170 Chr.9: 132594904-132594904 on Build GRCh38
rs556141300 Chr.9: 132594753-132594753 on Build GRCh38
rs1005401048 Chr.9: 132594703-132594703 on Build GRCh38
rs985236831 Chr.9: 132594813-132594813 on Build GRCh38
rs932443129 Chr.9: 132594563-132594563 on Build GRCh38
rs763665487 Chr.9: 132594923-132594923 on Build GRCh38
rs774131131 Chr.9: 132595043-132595043 on Build GRCh38
rs201737152 Chr.9: 132595007-132595007 on Build GRCh38
rs1014907616 Chr.9: 132594997-132594997 on Build GRCh38
rs73560853 Chr.9: 132594832-132594832 on Build GRCh38
rs201133051 Chr.9: 132595048-132595048 on Build GRCh38
rs766265730 Chr.9: 132594889-132594889 on Build GRCh38
rs758000606 Chr.9: 132594915-132594915 on Build GRCh38
rs768097189 Chr.9: 132595042-132595042 on Build GRCh38
rs936205827 Chr.9: 132594578-132594578 on Build GRCh38
rs138687182 Chr.9: 132595040-132595040 on Build GRCh38
rs960871633 Chr.9: 132594546-132594546 on Build GRCh38
rs775005678 Chr.9: 132594991-132594991 on Build GRCh38
rs937390295 Chr.9: 132594895-132594895 on Build GRCh38
rs1053369791 Chr.9: 132594584-132594584 on Build GRCh38
rs868674087 Chr.9: 132594673-132594673 on Build GRCh38
rs770709112 Chr.9: 132595028-132595028 on Build GRCh38
rs763389978 Chr.9: 132594553-132594553 on Build GRCh38
rs950315577 Chr.9: 132594858-132594858 on Build GRCh38
rs776536280 Chr.9: 132594982-132594982 on Build GRCh38
rs144575550 Chr.9: 132594645-132594645 on Build GRCh38
rs1001660090 Chr.9: 132594628-132594628 on Build GRCh38
rs752443098 Chr.9: 132594846-132594846 on Build GRCh38
rs372566843 Chr.9: 132594857-132594857 on Build GRCh38
rs306549 Chr.9: 132594789-132594789 on Build GRCh38
rs961305143 Chr.9: 132594731-132594731 on Build GRCh38
rs553394587 Chr.9: 132594828-132594828 on Build GRCh38
rs756590257 Chr.9: 132595013-132595013 on Build GRCh38
rs745675409 Chr.9: 132595031-132595031 on Build GRCh38
rs113038356 Chr.9: 132595018-132595018 on Build GRCh38
rs190560671 Chr.9: 132594531-132594531 on Build GRCh38
rs374984597 Chr.9: 132594995-132594995 on Build GRCh38
rs780449546 Chr.9: 132595019-132595019 on Build GRCh38
rs774470115 Chr.9: 132594959-132594959 on Build GRCh38
rs752081223 Chr.9: 132594984-132594984 on Build GRCh38
rs934579318 Chr.9: 132594621-132594621 on Build GRCh38
rs150458058 Chr.9: 132595022-132595022 on Build GRCh38
rs780897840 Chr.9: 132595030-132595030 on Build GRCh38
rs752372890 Chr.9: 132594951-132594951 on Build GRCh38
rs751295139 Chr.9: 132595003-132595003 on Build GRCh38
rs138252105 Chr.9: 132595024-132595024 on Build GRCh38
rs306548 Chr.9: 132594894-132594894 on Build GRCh38
rs749596077 Chr.9: 132594872-132594872 on Build GRCh38
rs896711968 Chr.9: 132594825-132594825 on Build GRCh38
rs377747166 Chr.9: 132594940-132594940 on Build GRCh38
rs267602151 Chr.9: 132594978-132594978 on Build GRCh38
rs367642142 Chr.9: 132595005-132595005 on Build GRCh38
rs374708193 Chr.9: 132594887-132594887 on Build GRCh38
rs966327973 Chr.9: 132594921-132594921 on Build GRCh38
rs561676028 Chr.9: 132595001-132595001 on Build GRCh38
rs186334076 Chr.9: 132594805-132594805 on Build GRCh38
rs768768089 Chr.9: 132594873-132594873 on Build GRCh38
rs979389973 Chr.9: 132594589-132594589 on Build GRCh38
rs976442828 Chr.9: 132594556-132594556 on Build GRCh38
rs371161695 Chr.9: 132594950-132594950 on Build GRCh38
rs757778902 Chr.9: 132594850-132594850 on Build GRCh38
rs759535820 Chr.9: 132594988-132594988 on Build GRCh38
rs306547 Chr.9: 132595027-132595027 on Build GRCh38
rs976536388 Chr.9: 132594751-132594751 on Build GRCh38
rs189935935 Chr.9: 132594848-132594848 on Build GRCh38
rs950181247 Chr.9: 132594605-132594605 on Build GRCh38
rs746776524 Chr.9: 132594972-132594972 on Build GRCh38
rs749343290 Chr.9: 132594956-132594956 on Build GRCh38
rs780118639 Chr.9: 132594955-132594955 on Build GRCh38
rs781769612 Chr.9: 132594853-132594853 on Build GRCh38
rs370819702 Chr.9: 132594928-132594928 on Build GRCh38
rs751920815 Chr.9: 132594930-132594930 on Build GRCh38
rs73560850 Chr.9: 132594557-132594557 on Build GRCh38
rs183529459 Chr.9: 132594656-132594656 on Build GRCh38
rs753297952 Chr.9: 132594831-132594831 on Build GRCh38
rs140473313 Chr.9: 132594939-132594939 on Build GRCh38
rs776591466 Chr.9: 132594892-132594892 on Build GRCh38
rs769720993 Chr.9: 132595039-132595039 on Build GRCh38
rs114397779 Chr.9: 132595025-132595025 on Build GRCh38
rs985528588 Chr.9: 132594575-132594575 on Build GRCh38
rs374961217 Chr.9: 132594576-132594576 on Build GRCh38
rs747338832 Chr.9: 132594849-132594849 on Build GRCh38
rs773005920 Chr.9: 132594875-132594875 on Build GRCh38
rs369132072 Chr.9: 132594730-132594730 on Build GRCh38
rs767407679 Chr.9: 132594815-132594815 on Build GRCh38
rs1000864619 Chr.9: 132594876-132594876 on Build GRCh38
rs770762452 Chr.9: 132594975-132594975 on Build GRCh38
rs762241533 Chr.9: 132595004-132595004 on Build GRCh38
rs762491372 Chr.9: 132595041-132595041 on Build GRCh38
rs369912738 Chr.9: 132594901-132594901 on Build GRCh38
rs778446111 Chr.9: 132594714-132594716 on Build GRCh38
rs182619230 Chr.9: 132594860-132594860 on Build GRCh38
rs752287250 Chr.9: 132594903-132594903 on Build GRCh38
rs886145280 Chr.9: 132594754-132594754 on Build GRCh38
rs769622687 Chr.9: 132594990-132594990 on Build GRCh38
rs570289130 Chr.9: 132594706-132594706 on Build GRCh38
rs372911669 Chr.9: 132594829-132594829 on Build GRCh38
rs745899775 Chr.9: 132594856-132594856 on Build GRCh38
rs961212889 Chr.9: 132594548-132594548 on Build GRCh38
rs912942946 Chr.9: 132595046-132595046 on Build GRCh38
rs768877501 Chr.9: 132594957-132594957 on Build GRCh38
rs750790221 Chr.9: 132594593-132594593 on Build GRCh38
rs1024193585 Chr.9: 132594838-132594838 on Build GRCh38
rs905939064 Chr.9: 132594609-132594609 on Build GRCh38
rs774664252 Chr.9: 132594692-132594692 on Build GRCh38

View Full Product Details