Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23706600-23706966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT9
Entrez Gene ID
23597
Gene Name
acyl-CoA thioesterase 9
Gene Aliases
ACATE2, CGI-16, MT-ACT48, MTACT48
UniGene
Hs.298885
Gene Chromosome Location
Chr.X: 23701055-23743290 on Build GRCh38
Transcripts
NM_001033583.2 NM_001037171.1 XM_005274472.2

SNP Detail

SNP ID Chromosome Location
rs377512509 Chr.X: 23706735-23706735 on Build GRCh38
rs746934885 Chr.X: 23706682-23706682 on Build GRCh38
rs754657325 Chr.X: 23706782-23706782 on Build GRCh38
rs774492451 Chr.X: 23706732-23706732 on Build GRCh38
rs761150147 Chr.X: 23706749-23706749 on Build GRCh38
rs757337263 Chr.X: 23706793-23706793 on Build GRCh38
rs139220036 Chr.X: 23706680-23706680 on Build GRCh38
rs200394258 Chr.X: 23706709-23706709 on Build GRCh38
rs972114762 Chr.X: 23706857-23706857 on Build GRCh38
rs377359478 Chr.X: 23706936-23706936 on Build GRCh38
rs372326977 Chr.X: 23706779-23706779 on Build GRCh38
rs368436703 Chr.X: 23706773-23706773 on Build GRCh38
rs35865928 Chr.X: 23706711-23706711 on Build GRCh38
rs370991464 Chr.X: 23706743-23706743 on Build GRCh38
rs193009863 Chr.X: 23706891-23706891 on Build GRCh38
rs546321 Chr.X: 23706867-23706867 on Build GRCh38
rs779581561 Chr.X: 23706786-23706786 on Build GRCh38
rs750543146 Chr.X: 23706804-23706804 on Build GRCh38
rs1043102493 Chr.X: 23706905-23706905 on Build GRCh38
rs771016382 Chr.X: 23706718-23706718 on Build GRCh38
rs375253099 Chr.X: 23706766-23706766 on Build GRCh38
rs891744670 Chr.X: 23706870-23706870 on Build GRCh38
rs142007269 Chr.X: 23706797-23706797 on Build GRCh38
rs201643422 Chr.X: 23706706-23706706 on Build GRCh38
rs745310560 Chr.X: 23706654-23706654 on Build GRCh38
rs965586685 Chr.X: 23706689-23706689 on Build GRCh38
rs971470671 Chr.X: 23706843-23706843 on Build GRCh38
rs375391179 Chr.X: 23706780-23706780 on Build GRCh38
rs754163613 Chr.X: 23706772-23706772 on Build GRCh38
rs149412949 Chr.X: 23706681-23706681 on Build GRCh38
rs941822652 Chr.X: 23706794-23706794 on Build GRCh38
rs938579750 Chr.X: 23706860-23706860 on Build GRCh38
rs750081033 Chr.X: 23706776-23706776 on Build GRCh38
rs977020326 Chr.X: 23706703-23706703 on Build GRCh38
rs1024811805 Chr.X: 23706811-23706811 on Build GRCh38
rs768840125 Chr.X: 23706743-23706752 on Build GRCh38
rs764741406 Chr.X: 23706775-23706775 on Build GRCh38
rs759475709 Chr.X: 23706734-23706734 on Build GRCh38
rs764466281 Chr.X: 23706758-23706758 on Build GRCh38

View Full Product Details