Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172295207-172295798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
UniGene
Hs.744888
Gene Chromosome Location
Chr.3: 172039628-172400703 on Build GRCh38
Transcripts
NM_001135095.1 NM_022763.3 XM_017007062.1 XM_017007063.1 XM_017007064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999797 c.973C>T p.R325* Chr.3: 172295486-172295486 on Build GRCh38
1041313 c.973C>A p.R325R Chr.3: 172295486-172295486 on Build GRCh38
4395253 c.984A>G p.K328K Chr.3: 172295497-172295497 on Build GRCh38
4685246 c.892delC p.P299fs*33 Chr.3: 172295405-172295405 on Build GRCh38
5863940 c.908C>T p.S303F Chr.3: 172295421-172295421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199826814 Chr.3: 172295532-172295532 on Build GRCh38
rs747440007 Chr.3: 172295389-172295389 on Build GRCh38
rs140664919 Chr.3: 172295435-172295435 on Build GRCh38
rs772592498 Chr.3: 172295381-172295381 on Build GRCh38
rs1034155463 Chr.3: 172295260-172295260 on Build GRCh38
rs139237887 Chr.3: 172295461-172295461 on Build GRCh38
rs750888712 Chr.3: 172295299-172295299 on Build GRCh38
rs747762602 Chr.3: 172295759-172295759 on Build GRCh38
rs1000538393 Chr.3: 172295757-172295757 on Build GRCh38
rs371917385 Chr.3: 172295382-172295382 on Build GRCh38
rs1021166963 Chr.3: 172295339-172295339 on Build GRCh38
rs765061009 Chr.3: 172295242-172295242 on Build GRCh38
rs910255195 Chr.3: 172295518-172295518 on Build GRCh38
rs758606800 Chr.3: 172295534-172295534 on Build GRCh38
rs1001048865 Chr.3: 172295668-172295668 on Build GRCh38
rs767368402 Chr.3: 172295336-172295336 on Build GRCh38
rs902712705 Chr.3: 172295653-172295653 on Build GRCh38
rs780595387 Chr.3: 172295509-172295509 on Build GRCh38
rs546230381 Chr.3: 172295686-172295686 on Build GRCh38
rs557492566 Chr.3: 172295284-172295284 on Build GRCh38
rs1030924389 Chr.3: 172295248-172295248 on Build GRCh38
rs771578611 Chr.3: 172295314-172295319 on Build GRCh38
rs919132553 Chr.3: 172295544-172295544 on Build GRCh38
rs529516574 Chr.3: 172295742-172295742 on Build GRCh38
rs746335098 Chr.3: 172295377-172295377 on Build GRCh38
rs748822053 Chr.3: 172295346-172295346 on Build GRCh38
rs572467276 Chr.3: 172295593-172295593 on Build GRCh38
rs1002222908 Chr.3: 172295230-172295230 on Build GRCh38
rs765187309 Chr.3: 172295432-172295432 on Build GRCh38
rs776767343 Chr.3: 172295405-172295405 on Build GRCh38
rs745397891 Chr.3: 172295484-172295484 on Build GRCh38
rs367903462 Chr.3: 172295520-172295520 on Build GRCh38
rs773357124 Chr.3: 172295560-172295560 on Build GRCh38
rs555695775 Chr.3: 172295450-172295450 on Build GRCh38
rs779444816 Chr.3: 172295374-172295374 on Build GRCh38
rs756967110 Chr.3: 172295465-172295465 on Build GRCh38
rs753474526 Chr.3: 172295462-172295462 on Build GRCh38
rs768941719 Chr.3: 172295517-172295517 on Build GRCh38
rs186484557 Chr.3: 172295334-172295334 on Build GRCh38
rs757226329 Chr.3: 172295514-172295515 on Build GRCh38
rs372899194 Chr.3: 172295297-172295297 on Build GRCh38
rs752552061 Chr.3: 172295340-172295340 on Build GRCh38
rs756766201 Chr.3: 172295358-172295358 on Build GRCh38
rs1040614544 Chr.3: 172295598-172295598 on Build GRCh38
rs201895621 Chr.3: 172295546-172295546 on Build GRCh38
rs932134567 Chr.3: 172295632-172295632 on Build GRCh38
rs754993753 Chr.3: 172295318-172295318 on Build GRCh38
rs1032573783 Chr.3: 172295676-172295676 on Build GRCh38
rs909151716 Chr.3: 172295601-172295601 on Build GRCh38
rs1000525581 Chr.3: 172295235-172295235 on Build GRCh38
rs975881788 Chr.3: 172295761-172295761 on Build GRCh38
rs73031398 Chr.3: 172295587-172295587 on Build GRCh38
rs374959823 Chr.3: 172295469-172295469 on Build GRCh38
rs113373621 Chr.3: 172295545-172295545 on Build GRCh38
rs758900070 Chr.3: 172295508-172295508 on Build GRCh38
rs768908474 Chr.3: 172295391-172295391 on Build GRCh38
rs1042559923 Chr.3: 172295647-172295647 on Build GRCh38
rs112002468 Chr.3: 172295609-172295609 on Build GRCh38
rs755923971 Chr.3: 172295454-172295454 on Build GRCh38
rs368660656 Chr.3: 172295343-172295343 on Build GRCh38
rs1017304617 Chr.3: 172295745-172295745 on Build GRCh38
rs376109959 Chr.3: 172295659-172295659 on Build GRCh38
rs758953163 Chr.3: 172295427-172295427 on Build GRCh38
rs747352220 Chr.3: 172295512-172295512 on Build GRCh38
rs896570768 Chr.3: 172295302-172295302 on Build GRCh38
rs56732464 Chr.3: 172295731-172295731 on Build GRCh38
rs374191084 Chr.3: 172295530-172295530 on Build GRCh38
rs767568739 Chr.3: 172295446-172295446 on Build GRCh38
rs761917415 Chr.3: 172295420-172295420 on Build GRCh38
rs780023942 Chr.3: 172295752-172295752 on Build GRCh38
rs902033588 Chr.3: 172295619-172295619 on Build GRCh38
rs200783931 Chr.3: 172295347-172295347 on Build GRCh38
rs755764265 Chr.3: 172295341-172295341 on Build GRCh38

View Full Product Details