Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44176031-44176590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
UniGene
Hs.521953
Gene Chromosome Location
Chr.X: 44146254-44343677 on Build GRCh38
Transcripts
NM_025184.3 XM_006724562.2

SNP Detail

SNP ID Chromosome Location
rs1001563053 Chr.X: 44176138-44176138 on Build GRCh38
rs776459837 Chr.X: 44176273-44176273 on Build GRCh38
rs374536009 Chr.X: 44176311-44176311 on Build GRCh38
rs941953490 Chr.X: 44176359-44176359 on Build GRCh38
rs779516860 Chr.X: 44176297-44176297 on Build GRCh38
rs922873524 Chr.X: 44176112-44176112 on Build GRCh38
rs754149543 Chr.X: 44176343-44176343 on Build GRCh38
rs774719233 Chr.X: 44176432-44176432 on Build GRCh38
rs367878877 Chr.X: 44176361-44176361 on Build GRCh38
rs772009254 Chr.X: 44176465-44176465 on Build GRCh38
rs917648614 Chr.X: 44176281-44176281 on Build GRCh38
rs1041621836 Chr.X: 44176498-44176498 on Build GRCh38
rs753395222 Chr.X: 44176408-44176408 on Build GRCh38
rs903169853 Chr.X: 44176531-44176531 on Build GRCh38
rs907498489 Chr.X: 44176218-44176218 on Build GRCh38
rs760784512 Chr.X: 44176315-44176315 on Build GRCh38
rs983062550 Chr.X: 44176188-44176188 on Build GRCh38
rs780176825 Chr.X: 44176391-44176391 on Build GRCh38
rs940355694 Chr.X: 44176270-44176270 on Build GRCh38
rs150446077 Chr.X: 44176099-44176099 on Build GRCh38
rs773847145 Chr.X: 44176283-44176283 on Build GRCh38
rs758365646 Chr.X: 44176384-44176384 on Build GRCh38
rs371762068 Chr.X: 44176423-44176423 on Build GRCh38
rs576164324 Chr.X: 44176059-44176059 on Build GRCh38
rs991888146 Chr.X: 44176240-44176240 on Build GRCh38
rs1009609486 Chr.X: 44176475-44176475 on Build GRCh38
rs932632330 Chr.X: 44176419-44176419 on Build GRCh38
rs774713964 Chr.X: 44176292-44176292 on Build GRCh38
rs7887384 Chr.X: 44176428-44176428 on Build GRCh38
rs1014555771 Chr.X: 44176427-44176427 on Build GRCh38
rs755854300 Chr.X: 44176205-44176205 on Build GRCh38
rs760020443 Chr.X: 44176303-44176303 on Build GRCh38
rs746483056 Chr.X: 44176399-44176399 on Build GRCh38
rs374567264 Chr.X: 44176360-44176360 on Build GRCh38
rs768194061 Chr.X: 44176202-44176202 on Build GRCh38
rs754580202 Chr.X: 44176400-44176400 on Build GRCh38
rs959053664 Chr.X: 44176208-44176208 on Build GRCh38
rs764280140 Chr.X: 44176336-44176336 on Build GRCh38
rs771382524 Chr.X: 44176289-44176289 on Build GRCh38
rs1021548171 Chr.X: 44176092-44176092 on Build GRCh38
rs1033341645 Chr.X: 44176176-44176176 on Build GRCh38
rs560465146 Chr.X: 44176234-44176234 on Build GRCh38
rs750202120 Chr.X: 44176375-44176375 on Build GRCh38
rs765018227 Chr.X: 44176351-44176351 on Build GRCh38

View Full Product Details