Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119545021-119545598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
UniGene
Hs.248572
Gene Chromosome Location
Chr.X: 119538149-119565434 on Build GRCh38
Transcripts
NM_001170569.1 NM_001170570.1 NM_022101.3

SNP Detail

SNP ID Chromosome Location
rs913825706 Chr.X: 119545382-119545382 on Build GRCh38
rs775201817 Chr.X: 119545430-119545430 on Build GRCh38
rs759026785 Chr.X: 119545196-119545197 on Build GRCh38
rs868336779 Chr.X: 119545352-119545352 on Build GRCh38
rs12556540 Chr.X: 119545105-119545105 on Build GRCh38
rs191963467 Chr.X: 119545425-119545425 on Build GRCh38
rs1043829446 Chr.X: 119545091-119545091 on Build GRCh38
rs202035774 Chr.X: 119545538-119545538 on Build GRCh38
rs751101464 Chr.X: 119545150-119545150 on Build GRCh38
rs377323466 Chr.X: 119545197-119545197 on Build GRCh38
rs751910333 Chr.X: 119545510-119545510 on Build GRCh38
rs373279148 Chr.X: 119545196-119545196 on Build GRCh38
rs141334748 Chr.X: 119545541-119545541 on Build GRCh38
rs58787648 Chr.X: 119545191-119545192 on Build GRCh38
rs1049306459 Chr.X: 119545076-119545076 on Build GRCh38
rs1034810927 Chr.X: 119545563-119545563 on Build GRCh38
rs879013517 Chr.X: 119545352-119545353 on Build GRCh38
rs372221185 Chr.X: 119545353-119545353 on Build GRCh38
rs1000743888 Chr.X: 119545126-119545126 on Build GRCh38
rs932109945 Chr.X: 119545057-119545057 on Build GRCh38
rs778042405 Chr.X: 119545385-119545385 on Build GRCh38
rs146602854 Chr.X: 119545434-119545434 on Build GRCh38
rs894726009 Chr.X: 119545129-119545129 on Build GRCh38
rs373822295 Chr.X: 119545415-119545415 on Build GRCh38
rs764570781 Chr.X: 119545503-119545503 on Build GRCh38
rs933888532 Chr.X: 119545370-119545370 on Build GRCh38
rs986682610 Chr.X: 119545374-119545374 on Build GRCh38
rs867453084 Chr.X: 119545198-119545198 on Build GRCh38
rs772679351 Chr.X: 119545551-119545551 on Build GRCh38
rs750518695 Chr.X: 119545540-119545540 on Build GRCh38
rs769368904 Chr.X: 119545427-119545427 on Build GRCh38
rs755928167 Chr.X: 119545353-119545355 on Build GRCh38
rs903623234 Chr.X: 119545112-119545112 on Build GRCh38
rs867364517 Chr.X: 119545132-119545132 on Build GRCh38
rs922141640 Chr.X: 119545046-119545046 on Build GRCh38
rs183412836 Chr.X: 119545087-119545087 on Build GRCh38
rs750105956 Chr.X: 119545079-119545079 on Build GRCh38
rs962856213 Chr.X: 119545279-119545279 on Build GRCh38
rs762445714 Chr.X: 119545518-119545518 on Build GRCh38
rs982822028 Chr.X: 119545164-119545164 on Build GRCh38
rs921148444 Chr.X: 119545315-119545315 on Build GRCh38
rs907170780 Chr.X: 119545522-119545522 on Build GRCh38
rs112264917 Chr.X: 119545128-119545128 on Build GRCh38
rs752504138 Chr.X: 119545304-119545304 on Build GRCh38
rs397950658 Chr.X: 119545195-119545196 on Build GRCh38
rs754862845 Chr.X: 119545189-119545189 on Build GRCh38
rs951342538 Chr.X: 119545152-119545152 on Build GRCh38

View Full Product Details