Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102749154-102749726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHLHB9
Entrez Gene ID
80823
Gene Name
basic helix-loop-helix domain containing, class B, 9
Gene Aliases
GASP3, p60TRP
UniGene
Hs.4276
Gene Chromosome Location
Chr.X: 102720714-102752441 on Build GRCh38
Transcripts
NM_001142524.1 NM_001142525.1 NM_001142526.1 NM_001142527.1 NM_001142528.1 NM_001142529.1 NM_001142530.1 NM_030639.2

SNP Detail

SNP ID Chromosome Location
rs372717305 Chr.X: 102749658-102749658 on Build GRCh38
rs777083653 Chr.X: 102749276-102749276 on Build GRCh38
rs752470520 Chr.X: 102749593-102749593 on Build GRCh38
rs755948490 Chr.X: 102749215-102749215 on Build GRCh38
rs766182016 Chr.X: 102749334-102749334 on Build GRCh38
rs780429755 Chr.X: 102749664-102749664 on Build GRCh38
rs189045157 Chr.X: 102749351-102749351 on Build GRCh38
rs767068278 Chr.X: 102749300-102749300 on Build GRCh38
rs2179675 Chr.X: 102749389-102749389 on Build GRCh38
rs1036118088 Chr.X: 102749204-102749204 on Build GRCh38
rs747018279 Chr.X: 102749427-102749427 on Build GRCh38
rs771586540 Chr.X: 102749470-102749470 on Build GRCh38
rs17854123 Chr.X: 102749700-102749700 on Build GRCh38
rs749607896 Chr.X: 102749668-102749668 on Build GRCh38
rs760701478 Chr.X: 102749320-102749320 on Build GRCh38
rs777911515 Chr.X: 102749408-102749408 on Build GRCh38
rs141213425 Chr.X: 102749456-102749456 on Build GRCh38
rs368463899 Chr.X: 102749560-102749560 on Build GRCh38
rs753738502 Chr.X: 102749340-102749340 on Build GRCh38
rs867262335 Chr.X: 102749461-102749461 on Build GRCh38
rs771273673 Chr.X: 102749270-102749270 on Build GRCh38
rs754794333 Chr.X: 102749345-102749345 on Build GRCh38
rs184677680 Chr.X: 102749235-102749235 on Build GRCh38
rs138015098 Chr.X: 102749650-102749650 on Build GRCh38
rs770329576 Chr.X: 102749702-102749702 on Build GRCh38
rs776708971 Chr.X: 102749555-102749555 on Build GRCh38
rs766383489 Chr.X: 102749533-102749533 on Build GRCh38
rs375334052 Chr.X: 102749660-102749660 on Build GRCh38
rs761291178 Chr.X: 102749183-102749183 on Build GRCh38
rs779905239 Chr.X: 102749225-102749225 on Build GRCh38
rs747561089 Chr.X: 102749264-102749264 on Build GRCh38
rs781527721 Chr.X: 102749433-102749433 on Build GRCh38
rs751707842 Chr.X: 102749639-102749639 on Build GRCh38
rs757804218 Chr.X: 102749431-102749431 on Build GRCh38
rs367874505 Chr.X: 102749295-102749295 on Build GRCh38
rs748282084 Chr.X: 102749248-102749248 on Build GRCh38
rs146968166 Chr.X: 102749562-102749562 on Build GRCh38
rs760084844 Chr.X: 102749506-102749506 on Build GRCh38
rs759844076 Chr.X: 102749278-102749278 on Build GRCh38
rs764877521 Chr.X: 102749563-102749563 on Build GRCh38
rs772711917 Chr.X: 102749675-102749675 on Build GRCh38
rs747805882 Chr.X: 102749467-102749467 on Build GRCh38
rs772270165 Chr.X: 102749249-102749249 on Build GRCh38
rs772531860 Chr.X: 102749319-102749319 on Build GRCh38
rs769450022 Chr.X: 102749491-102749493 on Build GRCh38
rs746600310 Chr.X: 102749683-102749683 on Build GRCh38
rs1033656815 Chr.X: 102749674-102749674 on Build GRCh38
rs754307795 Chr.X: 102749263-102749263 on Build GRCh38
rs772802916 Chr.X: 102749482-102749482 on Build GRCh38
rs773961613 Chr.X: 102749292-102749292 on Build GRCh38
rs112563174 Chr.X: 102749380-102749380 on Build GRCh38
rs149011689 Chr.X: 102749228-102749228 on Build GRCh38
rs767510675 Chr.X: 102749194-102749194 on Build GRCh38
rs779259163 Chr.X: 102749237-102749237 on Build GRCh38
rs970772618 Chr.X: 102749579-102749579 on Build GRCh38
rs768345325 Chr.X: 102749283-102749283 on Build GRCh38
rs889399480 Chr.X: 102749393-102749393 on Build GRCh38
rs375161860 Chr.X: 102749457-102749457 on Build GRCh38
rs371712485 Chr.X: 102749395-102749395 on Build GRCh38
rs893974811 Chr.X: 102749477-102749477 on Build GRCh38
rs193009805 Chr.X: 102749466-102749466 on Build GRCh38
rs145845995 Chr.X: 102749205-102749205 on Build GRCh38
rs758675977 Chr.X: 102749597-102749597 on Build GRCh38
rs764171456 Chr.X: 102749612-102749612 on Build GRCh38

View Full Product Details