Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110674116-110674675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL1
Entrez Gene ID
91851
Gene Name
chordin like 1
Gene Aliases
CHL, MGC1, MGCN, NRLN1, VOPT, dA141H5.1
UniGene
Hs.496587
Gene Chromosome Location
Chr.X: 110673856-110796058 on Build GRCh38
Transcripts
NM_001143981.1 NM_001143982.1 NM_001143983.2 NM_145234.3 XM_005262221.1 XM_005262222.3 XM_005262223.1 XM_005262224.1 XM_017029959.1

SNP Detail

SNP ID Chromosome Location
rs796916849 Chr.X: 110674458-110674459 on Build GRCh38
rs1002744284 Chr.X: 110674342-110674342 on Build GRCh38
rs886523349 Chr.X: 110674282-110674282 on Build GRCh38
rs768070199 Chr.X: 110674499-110674506 on Build GRCh38
rs12007691 Chr.X: 110674462-110674462 on Build GRCh38
rs982272922 Chr.X: 110674143-110674143 on Build GRCh38
rs12012172 Chr.X: 110674506-110674506 on Build GRCh38
rs928087331 Chr.X: 110674531-110674531 on Build GRCh38
rs10526 Chr.X: 110674445-110674445 on Build GRCh38
rs1008735913 Chr.X: 110674390-110674390 on Build GRCh38
rs988874872 Chr.X: 110674478-110674478 on Build GRCh38
rs61447153 Chr.X: 110674461-110674462 on Build GRCh38
rs16986076 Chr.X: 110674421-110674421 on Build GRCh38
rs938261044 Chr.X: 110674542-110674542 on Build GRCh38
rs991092334 Chr.X: 110674602-110674602 on Build GRCh38
rs904029587 Chr.X: 110674318-110674318 on Build GRCh38
rs59391928 Chr.X: 110674505-110674506 on Build GRCh38
rs1033953830 Chr.X: 110674359-110674359 on Build GRCh38
rs961367637 Chr.X: 110674631-110674631 on Build GRCh38
rs971515207 Chr.X: 110674507-110674507 on Build GRCh38
rs982282762 Chr.X: 110674514-110674514 on Build GRCh38
rs764377400 Chr.X: 110674398-110674398 on Build GRCh38
rs182515684 Chr.X: 110674605-110674605 on Build GRCh38
rs866429935 Chr.X: 110674443-110674443 on Build GRCh38
rs1032872066 Chr.X: 110674434-110674434 on Build GRCh38
rs5943043 Chr.X: 110674164-110674164 on Build GRCh38
rs183602384 Chr.X: 110674402-110674402 on Build GRCh38
rs746719422 Chr.X: 110674459-110674460 on Build GRCh38
rs769945270 Chr.X: 110674222-110674222 on Build GRCh38
rs796243093 Chr.X: 110674457-110674458 on Build GRCh38
rs938208649 Chr.X: 110674189-110674189 on Build GRCh38
rs759701623 Chr.X: 110674190-110674192 on Build GRCh38
rs780421754 Chr.X: 110674264-110674264 on Build GRCh38
rs1052748303 Chr.X: 110674145-110674145 on Build GRCh38
rs758816812 Chr.X: 110674487-110674506 on Build GRCh38
rs930702961 Chr.X: 110674632-110674632 on Build GRCh38
rs1055141792 Chr.X: 110674203-110674203 on Build GRCh38
rs900441298 Chr.X: 110674407-110674407 on Build GRCh38
rs1043191360 Chr.X: 110674296-110674296 on Build GRCh38
rs899139782 Chr.X: 110674235-110674235 on Build GRCh38
rs756457567 Chr.X: 110674261-110674261 on Build GRCh38
rs928203469 Chr.X: 110674186-110674186 on Build GRCh38
rs1048154358 Chr.X: 110674637-110674637 on Build GRCh38
rs1013834296 Chr.X: 110674447-110674447 on Build GRCh38
rs775884139 Chr.X: 110674381-110674381 on Build GRCh38
rs200106486 Chr.X: 110674460-110674461 on Build GRCh38

View Full Product Details