Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20592058-20592635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AY
Entrez Gene ID
9086
Gene Name
eukaryotic translation initiation factor 1A, Y-linked
Gene Aliases
eIF-4C
UniGene
Hs.461178
Gene Chromosome Location
Chr.Y: 20575711-20593154 on Build GRCh38
Transcripts
NM_001278612.1 NM_004681.3

SNP Detail

SNP ID Chromosome Location
rs760879760 Chr.Y: 20592369-20592369 on Build GRCh38
rs887831747 Chr.Y: 20592256-20592256 on Build GRCh38
rs747272238 Chr.Y: 20592590-20592590 on Build GRCh38
rs1039141753 Chr.Y: 20592448-20592448 on Build GRCh38
rs758018269 Chr.Y: 20592462-20592462 on Build GRCh38
rs766586400 Chr.Y: 20592377-20592377 on Build GRCh38
rs942223906 Chr.Y: 20592353-20592353 on Build GRCh38
rs997759500 Chr.Y: 20592553-20592553 on Build GRCh38

View Full Product Details