Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197043772-197044370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
UniGene
Hs.292549
Gene Chromosome Location
Chr.3: 197042560-197299272 on Build GRCh38
Transcripts
NM_001098424.1 NM_001204386.1 NM_001204387.1 NM_001204388.1 NM_001290983.1 NM_004087.2 XM_005269289.3 XM_011512502.2 XM_011512503.1 XM_011512505.1 XM_011512506.1 XM_017005800.1 XM_017005801.1 XM_017005802.1 XM_017005803.1 XM_017005804.1 XM_017005805.1 XM_017005806.1 XM_017005807.1 XM_017005808.1 XM_017005809.1 XM_017005810.1 XM_017005811.1 XM_017005812.1 XM_017005813.1 XM_017005814.1 XM_017005815.1 XM_017005816.1 XM_017005817.1 XM_017005818.1 XM_017005819.1 XM_017005820.1 XM_017005821.1 XM_017005822.1 XM_017005823.1 XM_017005824.1

SNP Detail

SNP ID Chromosome Location
rs918352569 Chr.3: 197044092-197044092 on Build GRCh38
rs747170339 Chr.3: 197044213-197044213 on Build GRCh38
rs985418369 Chr.3: 197044207-197044207 on Build GRCh38
rs942948178 Chr.3: 197044194-197044194 on Build GRCh38
rs866958660 Chr.3: 197044031-197044031 on Build GRCh38
rs933619004 Chr.3: 197043795-197043795 on Build GRCh38
rs950362885 Chr.3: 197044121-197044121 on Build GRCh38
rs922473488 Chr.3: 197044004-197044004 on Build GRCh38
rs1010483246 Chr.3: 197044160-197044160 on Build GRCh38
rs562101334 Chr.3: 197043811-197043812 on Build GRCh38
rs986875925 Chr.3: 197044168-197044168 on Build GRCh38
rs896075736 Chr.3: 197043907-197043907 on Build GRCh38
rs1051385417 Chr.3: 197043890-197043890 on Build GRCh38
rs929425365 Chr.3: 197044102-197044103 on Build GRCh38
rs910918081 Chr.3: 197044051-197044051 on Build GRCh38
rs1024897408 Chr.3: 197044009-197044009 on Build GRCh38
rs1034159018 Chr.3: 197044062-197044062 on Build GRCh38
rs910959806 Chr.3: 197043822-197043823 on Build GRCh38
rs1023003354 Chr.3: 197043946-197043946 on Build GRCh38
rs546929312 Chr.3: 197044214-197044214 on Build GRCh38
rs1048416144 Chr.3: 197043903-197043903 on Build GRCh38
rs758491203 Chr.3: 197043982-197043982 on Build GRCh38
rs951130427 Chr.3: 197044156-197044156 on Build GRCh38
rs942893031 Chr.3: 197043925-197043925 on Build GRCh38
rs899354310 Chr.3: 197043894-197043894 on Build GRCh38
rs1037860805 Chr.3: 197043866-197043866 on Build GRCh38
rs973795307 Chr.3: 197044082-197044082 on Build GRCh38
rs747862661 Chr.3: 197043985-197043985 on Build GRCh38
rs45448597 Chr.3: 197044178-197044178 on Build GRCh38
rs780239784 Chr.3: 197044020-197044020 on Build GRCh38
rs1031484011 Chr.3: 197043952-197043952 on Build GRCh38
rs12630899 Chr.3: 197044112-197044112 on Build GRCh38
rs1000984403 Chr.3: 197043950-197043950 on Build GRCh38
rs936829874 Chr.3: 197044290-197044290 on Build GRCh38
rs962341510 Chr.3: 197044228-197044229 on Build GRCh38
rs1044776230 Chr.3: 197044133-197044133 on Build GRCh38
rs368544753 Chr.3: 197043904-197043904 on Build GRCh38
rs979965502 Chr.3: 197044035-197044035 on Build GRCh38
rs925463276 Chr.3: 197044222-197044222 on Build GRCh38
rs1049408977 Chr.3: 197043807-197043808 on Build GRCh38
rs892505834 Chr.3: 197043967-197043967 on Build GRCh38
rs903996003 Chr.3: 197043945-197043945 on Build GRCh38
rs751986883 Chr.3: 197044134-197044134 on Build GRCh38
rs973741509 Chr.3: 197044307-197044307 on Build GRCh38
rs891392739 Chr.3: 197043917-197043917 on Build GRCh38
rs1047892095 Chr.3: 197044109-197044109 on Build GRCh38
rs918283456 Chr.3: 197043808-197043808 on Build GRCh38
rs1052564322 Chr.3: 197044344-197044344 on Build GRCh38
rs1029532321 Chr.3: 197044176-197044176 on Build GRCh38
rs929629693 Chr.3: 197043847-197043847 on Build GRCh38
rs375724869 Chr.3: 197044275-197044275 on Build GRCh38
rs78560510 Chr.3: 197044085-197044085 on Build GRCh38
rs993684256 Chr.3: 197043896-197043896 on Build GRCh38
rs138046498 Chr.3: 197043937-197043937 on Build GRCh38
rs896052400 Chr.3: 197044113-197044114 on Build GRCh38
rs183201961 Chr.3: 197044154-197044154 on Build GRCh38
rs973889806 Chr.3: 197044136-197044136 on Build GRCh38
rs543227581 Chr.3: 197043981-197043981 on Build GRCh38
rs891885080 Chr.3: 197044159-197044159 on Build GRCh38

View Full Product Details