Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98795719-98796313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCBLD2
Entrez Gene ID
131566
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
CLCP1, ESDN
UniGene
Hs.203691
Gene Chromosome Location
Chr.3: 98795970-98901689 on Build GRCh38
Transcripts
NM_080927.3 XM_011512419.2 NM_001271142.1 NM_001271145.1 NM_001271146.1 NM_001271147.1 NM_001271148.1 NM_001323352.1 NM_001323353.1 NM_001323358.1 NM_001323359.1 NM_001323360.1 NM_001323362.1 NM_001323363.1 NM_001323364.1 NM_001323365.1 NM_001323366.1 NM_001323367.1 NM_001323368.1 NM_006100.3

Gene Symbol
ST3GAL6
Entrez Gene ID
10402
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 6
Gene Aliases
SIAT10, ST3GALVI
UniGene
Hs.148716
Gene Chromosome Location
Chr.3: 98732236-98795845 on Build GRCh38
Transcripts
NM_080927.3 XM_011512419.2 NM_001271142.1 NM_001271145.1 NM_001271146.1 NM_001271147.1 NM_001271148.1 NM_001323352.1 NM_001323353.1 NM_001323358.1 NM_001323359.1 NM_001323360.1 NM_001323362.1 NM_001323363.1 NM_001323364.1 NM_001323365.1 NM_001323366.1 NM_001323367.1 NM_001323368.1 NM_006100.3

SNP Detail

SNP ID Chromosome Location
rs1019406636 Chr.3: 98795887-98795887 on Build GRCh38
rs1043849986 Chr.3: 98796135-98796135 on Build GRCh38
rs532731439 Chr.3: 98795782-98795783 on Build GRCh38
rs940846233 Chr.3: 98796056-98796056 on Build GRCh38
rs138084992 Chr.3: 98796238-98796239 on Build GRCh38
rs931965909 Chr.3: 98795741-98795741 on Build GRCh38
rs993675345 Chr.3: 98796225-98796225 on Build GRCh38
rs972190306 Chr.3: 98795750-98795750 on Build GRCh38
rs1007964720 Chr.3: 98795910-98795910 on Build GRCh38
rs761931398 Chr.3: 98796261-98796261 on Build GRCh38
rs927860945 Chr.3: 98795819-98795819 on Build GRCh38
rs1003243672 Chr.3: 98796277-98796277 on Build GRCh38
rs774829860 Chr.3: 98796237-98796238 on Build GRCh38
rs1023317964 Chr.3: 98796001-98796001 on Build GRCh38
rs1017094917 Chr.3: 98796036-98796037 on Build GRCh38
rs960590242 Chr.3: 98796087-98796087 on Build GRCh38
rs772701226 Chr.3: 98796255-98796257 on Build GRCh38
rs927890463 Chr.3: 98796047-98796047 on Build GRCh38
rs1032662942 Chr.3: 98795989-98795989 on Build GRCh38
rs982270788 Chr.3: 98796039-98796039 on Build GRCh38
rs746716628 Chr.3: 98796096-98796096 on Build GRCh38
rs903298244 Chr.3: 98796195-98796195 on Build GRCh38
rs572869243 Chr.3: 98795994-98795994 on Build GRCh38
rs957962057 Chr.3: 98795993-98795993 on Build GRCh38
rs548273584 Chr.3: 98796173-98796173 on Build GRCh38
rs993642381 Chr.3: 98795985-98795985 on Build GRCh38
rs999750949 Chr.3: 98796236-98796236 on Build GRCh38
rs35887694 Chr.3: 98796251-98796252 on Build GRCh38
rs768936531 Chr.3: 98796078-98796078 on Build GRCh38
rs907655054 Chr.3: 98796044-98796044 on Build GRCh38
rs951140618 Chr.3: 98796016-98796016 on Build GRCh38
rs946538587 Chr.3: 98796134-98796134 on Build GRCh38
rs56200647 Chr.3: 98795854-98795854 on Build GRCh38
rs768718075 Chr.3: 98795910-98795911 on Build GRCh38
rs11552976 Chr.3: 98796172-98796172 on Build GRCh38
rs530848860 Chr.3: 98795878-98795879 on Build GRCh38
rs984203799 Chr.3: 98795789-98795789 on Build GRCh38
rs78857045 Chr.3: 98796253-98796253 on Build GRCh38
rs72934634 Chr.3: 98796272-98796272 on Build GRCh38
rs566206841 Chr.3: 98796251-98796251 on Build GRCh38
rs553072321 Chr.3: 98795938-98795938 on Build GRCh38
rs1032192439 Chr.3: 98796249-98796249 on Build GRCh38
rs932249067 Chr.3: 98796156-98796156 on Build GRCh38
rs529867976 Chr.3: 98796262-98796262 on Build GRCh38
rs1005264094 Chr.3: 98796019-98796019 on Build GRCh38
rs1050353680 Chr.3: 98796165-98796165 on Build GRCh38
rs765921906 Chr.3: 98796276-98796277 on Build GRCh38
rs771767646 Chr.3: 98795846-98795846 on Build GRCh38
rs1025369797 Chr.3: 98795761-98795761 on Build GRCh38
rs909518105 Chr.3: 98795793-98795793 on Build GRCh38
rs1050404230 Chr.3: 98795794-98795794 on Build GRCh38
rs980754405 Chr.3: 98795816-98795816 on Build GRCh38
rs889161166 Chr.3: 98795890-98795891 on Build GRCh38
rs574164935 Chr.3: 98795835-98795835 on Build GRCh38
rs939195900 Chr.3: 98795828-98795829 on Build GRCh38
rs397774026 Chr.3: 98796252-98796253 on Build GRCh38
rs749018725 Chr.3: 98795885-98795888 on Build GRCh38
rs969524924 Chr.3: 98796277-98796277 on Build GRCh38
rs902114996 Chr.3: 98796199-98796199 on Build GRCh38
rs377226898 Chr.3: 98795980-98795980 on Build GRCh38
rs370984642 Chr.3: 98796087-98796087 on Build GRCh38
rs564983260 Chr.3: 98796115-98796115 on Build GRCh38
rs970430326 Chr.3: 98796013-98796013 on Build GRCh38
rs1042188027 Chr.3: 98796187-98796187 on Build GRCh38
rs374594340 Chr.3: 98796276-98796276 on Build GRCh38
rs3836215 Chr.3: 98796236-98796237 on Build GRCh38

View Full Product Details