Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183446703-183447280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2AIP
Entrez Gene ID
55602
Gene Name
CDKN2A interacting protein
Gene Aliases
CARF
UniGene
Hs.592508
Gene Chromosome Location
Chr.4: 183444591-183449064 on Build GRCh38
Transcripts
NM_001317343.1 NM_017632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5801476 c.1527_1528delTG p.Y511fs*35 Chr.4: 183447211-183447212 on Build GRCh38
1695292 c.1409C>G p.A470G Chr.4: 183447093-183447093 on Build GRCh38
5742606 c.1314_1315insT p.N441fs*2 Chr.4: 183446998-183446999 on Build GRCh38
4657640 c.1250C>G p.S417* Chr.4: 183446934-183446934 on Build GRCh38
279792 c.1459G>T p.E487* Chr.4: 183447143-183447143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138463372 Chr.4: 183446956-183446956 on Build GRCh38
rs773531189 Chr.4: 183446729-183446729 on Build GRCh38
rs753862628 Chr.4: 183446919-183446919 on Build GRCh38
rs982831205 Chr.4: 183446837-183446837 on Build GRCh38
rs201608702 Chr.4: 183447098-183447098 on Build GRCh38
rs141991851 Chr.4: 183446920-183446920 on Build GRCh38
rs759169753 Chr.4: 183446763-183446763 on Build GRCh38
rs767343115 Chr.4: 183446769-183446769 on Build GRCh38
rs767688935 Chr.4: 183447028-183447028 on Build GRCh38
rs199963421 Chr.4: 183446995-183446995 on Build GRCh38
rs775066417 Chr.4: 183446908-183446908 on Build GRCh38
rs781564950 Chr.4: 183446933-183446933 on Build GRCh38
rs768213975 Chr.4: 183447155-183447155 on Build GRCh38
rs779507257 Chr.4: 183447141-183447141 on Build GRCh38
rs369366498 Chr.4: 183447043-183447043 on Build GRCh38
rs764106682 Chr.4: 183446784-183446784 on Build GRCh38
rs750315909 Chr.4: 183447013-183447013 on Build GRCh38
rs753652052 Chr.4: 183446792-183446792 on Build GRCh38
rs1039633713 Chr.4: 183446897-183446897 on Build GRCh38
rs755280246 Chr.4: 183446811-183446811 on Build GRCh38
rs769981592 Chr.4: 183446850-183446850 on Build GRCh38
rs760761742 Chr.4: 183447240-183447240 on Build GRCh38
rs35970597 Chr.4: 183446840-183446840 on Build GRCh38
rs373518027 Chr.4: 183447055-183447055 on Build GRCh38
rs576981981 Chr.4: 183446815-183446815 on Build GRCh38
rs778018161 Chr.4: 183447056-183447056 on Build GRCh38
rs150114244 Chr.4: 183446948-183446948 on Build GRCh38
rs201828834 Chr.4: 183446757-183446757 on Build GRCh38
rs755040601 Chr.4: 183447150-183447150 on Build GRCh38
rs763782262 Chr.4: 183446917-183446917 on Build GRCh38
rs745397785 Chr.4: 183446961-183446961 on Build GRCh38
rs73870527 Chr.4: 183446728-183446728 on Build GRCh38
rs777982179 Chr.4: 183446942-183446942 on Build GRCh38
rs903244741 Chr.4: 183446952-183446952 on Build GRCh38
rs749748152 Chr.4: 183446859-183446859 on Build GRCh38
rs759751969 Chr.4: 183447022-183447022 on Build GRCh38
rs746485773 Chr.4: 183447148-183447148 on Build GRCh38
rs200444062 Chr.4: 183446941-183446941 on Build GRCh38
rs779397340 Chr.4: 183447082-183447082 on Build GRCh38
rs757862367 Chr.4: 183447176-183447176 on Build GRCh38
rs755048889 Chr.4: 183446931-183446931 on Build GRCh38
rs751708633 Chr.4: 183446928-183446928 on Build GRCh38
rs934158255 Chr.4: 183446794-183446794 on Build GRCh38
rs747896830 Chr.4: 183447159-183447159 on Build GRCh38
rs773822857 Chr.4: 183446892-183446892 on Build GRCh38
rs769703165 Chr.4: 183447170-183447170 on Build GRCh38
rs1037113618 Chr.4: 183446765-183446765 on Build GRCh38
rs771344065 Chr.4: 183446882-183446884 on Build GRCh38
rs941238397 Chr.4: 183446880-183446880 on Build GRCh38
rs760469515 Chr.4: 183446913-183446913 on Build GRCh38
rs758563013 Chr.4: 183446798-183446798 on Build GRCh38
rs774410382 Chr.4: 183447235-183447235 on Build GRCh38
rs779975198 Chr.4: 183446805-183446805 on Build GRCh38
rs776285795 Chr.4: 183447157-183447157 on Build GRCh38
rs376830028 Chr.4: 183446806-183446806 on Build GRCh38
rs921136302 Chr.4: 183446904-183446904 on Build GRCh38
rs775466362 Chr.4: 183447235-183447235 on Build GRCh38
rs775191114 Chr.4: 183446964-183446964 on Build GRCh38
rs371120721 Chr.4: 183446751-183446751 on Build GRCh38
rs765179780 Chr.4: 183446924-183446924 on Build GRCh38
rs771618693 Chr.4: 183447110-183447110 on Build GRCh38
rs765276188 Chr.4: 183447005-183447005 on Build GRCh38
rs200973636 Chr.4: 183447063-183447063 on Build GRCh38
rs75299215 Chr.4: 183447232-183447232 on Build GRCh38
rs751951765 Chr.4: 183446736-183446736 on Build GRCh38
rs971659444 Chr.4: 183447084-183447084 on Build GRCh38
rs373370372 Chr.4: 183446778-183446778 on Build GRCh38
rs776319496 Chr.4: 183446974-183446974 on Build GRCh38
rs779344281 Chr.4: 183446955-183446955 on Build GRCh38
rs757585751 Chr.4: 183447071-183447071 on Build GRCh38
rs780182422 Chr.4: 183446896-183446896 on Build GRCh38
rs759008324 Chr.4: 183446895-183446895 on Build GRCh38
rs758832945 Chr.4: 183446734-183446734 on Build GRCh38
rs768536227 Chr.4: 183446973-183446973 on Build GRCh38
rs201076176 Chr.4: 183446823-183446823 on Build GRCh38
rs770528042 Chr.4: 183446888-183446888 on Build GRCh38
rs746369912 Chr.4: 183447094-183447094 on Build GRCh38
rs773336783 Chr.4: 183447192-183447192 on Build GRCh38
rs368760511 Chr.4: 183446770-183446770 on Build GRCh38
rs781511940 Chr.4: 183446814-183446814 on Build GRCh38
rs893129537 Chr.4: 183447131-183447131 on Build GRCh38
rs762965030 Chr.4: 183447208-183447208 on Build GRCh38
rs1013625715 Chr.4: 183447174-183447174 on Build GRCh38

View Full Product Details