Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 44388267-44388765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF10
Entrez Gene ID
2255
Gene Name
fibroblast growth factor 10
Gene Aliases
-
UniGene
Hs.248049
Gene Chromosome Location
Chr.5: 44301655-44389780 on Build GRCh38
Transcripts
NM_004465.1 XM_005248264.3 NR_108034.1

Gene Symbol
FGF10-AS1
Entrez Gene ID
101927075
Gene Name
FGF10 antisense RNA 1
Gene Aliases
-
UniGene
Hs.617390
Gene Chromosome Location
Chr.5: 44388732-44413989 on Build GRCh38
Transcripts
NM_004465.1 XM_005248264.3 NR_108034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6663466 c.201G>C p.V67V Chr.5: 44388482-44388482 on Build GRCh38
6663464 c.188C>T p.A63V Chr.5: 44388495-44388495 on Build GRCh38
5506850 c.274A>G p.I92V Chr.5: 44388409-44388409 on Build GRCh38
1068457 c.92C>T p.S31F Chr.5: 44388591-44388591 on Build GRCh38
3674457 c.286G>T p.G96W Chr.5: 44388397-44388397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527933059 Chr.5: 44388544-44388544 on Build GRCh38
rs747476439 Chr.5: 44388667-44388667 on Build GRCh38
rs768364731 Chr.5: 44388605-44388605 on Build GRCh38
rs146669375 Chr.5: 44388565-44388565 on Build GRCh38
rs767735212 Chr.5: 44388383-44388383 on Build GRCh38
rs758481605 Chr.5: 44388662-44388662 on Build GRCh38
rs200089060 Chr.5: 44388720-44388720 on Build GRCh38
rs761396841 Chr.5: 44388710-44388710 on Build GRCh38
rs751081612 Chr.5: 44388537-44388537 on Build GRCh38
rs104893885 Chr.5: 44388366-44388366 on Build GRCh38
rs200382374 Chr.5: 44388311-44388311 on Build GRCh38
rs745684641 Chr.5: 44388469-44388469 on Build GRCh38
rs778802173 Chr.5: 44388556-44388556 on Build GRCh38
rs768676783 Chr.5: 44388494-44388494 on Build GRCh38
rs201790067 Chr.5: 44388567-44388567 on Build GRCh38
rs374755098 Chr.5: 44388634-44388634 on Build GRCh38
rs780222015 Chr.5: 44388477-44388477 on Build GRCh38
rs201719161 Chr.5: 44388368-44388368 on Build GRCh38
rs201772112 Chr.5: 44388697-44388697 on Build GRCh38
rs140279180 Chr.5: 44388581-44388581 on Build GRCh38
rs775259892 Chr.5: 44388528-44388528 on Build GRCh38
rs201058164 Chr.5: 44388317-44388317 on Build GRCh38
rs200247528 Chr.5: 44388619-44388619 on Build GRCh38
rs1007631855 Chr.5: 44388579-44388579 on Build GRCh38
rs775751673 Chr.5: 44388346-44388346 on Build GRCh38
rs895685307 Chr.5: 44388300-44388300 on Build GRCh38
rs199700809 Chr.5: 44388335-44388335 on Build GRCh38
rs762407366 Chr.5: 44388380-44388380 on Build GRCh38
rs200141272 Chr.5: 44388701-44388701 on Build GRCh38
rs772109862 Chr.5: 44388561-44388561 on Build GRCh38
rs761116992 Chr.5: 44388514-44388514 on Build GRCh38
rs104893888 Chr.5: 44388443-44388443 on Build GRCh38
rs774523215 Chr.5: 44388501-44388501 on Build GRCh38
rs568356056 Chr.5: 44388296-44388296 on Build GRCh38
rs779192199 Chr.5: 44388647-44388647 on Build GRCh38
rs762625065 Chr.5: 44388592-44388592 on Build GRCh38
rs750371676 Chr.5: 44388546-44388546 on Build GRCh38
rs201659493 Chr.5: 44388666-44388666 on Build GRCh38
rs201676495 Chr.5: 44388492-44388492 on Build GRCh38
rs766097540 Chr.5: 44388635-44388635 on Build GRCh38
rs761359401 Chr.5: 44388628-44388628 on Build GRCh38
rs201987275 Chr.5: 44388352-44388352 on Build GRCh38
rs761372591 Chr.5: 44388291-44388292 on Build GRCh38
rs779848984 Chr.5: 44388552-44388552 on Build GRCh38
rs772647537 Chr.5: 44388509-44388509 on Build GRCh38
rs766290230 Chr.5: 44388724-44388724 on Build GRCh38
rs200701392 Chr.5: 44388363-44388363 on Build GRCh38
rs766989043 Chr.5: 44388631-44388631 on Build GRCh38
rs562602772 Chr.5: 44388746-44388746 on Build GRCh38
rs145373611 Chr.5: 44388586-44388586 on Build GRCh38
rs773650137 Chr.5: 44388512-44388512 on Build GRCh38
rs886060655 Chr.5: 44388539-44388539 on Build GRCh38
rs369132289 Chr.5: 44388688-44388688 on Build GRCh38
rs773315020 Chr.5: 44388425-44388425 on Build GRCh38
rs898009871 Chr.5: 44388499-44388499 on Build GRCh38
rs199850745 Chr.5: 44388504-44388504 on Build GRCh38
rs201038841 Chr.5: 44388507-44388507 on Build GRCh38
rs770003641 Chr.5: 44388345-44388345 on Build GRCh38
rs576181814 Chr.5: 44388613-44388615 on Build GRCh38
rs756978392 Chr.5: 44388325-44388325 on Build GRCh38
rs145059362 Chr.5: 44388343-44388343 on Build GRCh38
rs199689272 Chr.5: 44388455-44388455 on Build GRCh38
rs148032639 Chr.5: 44388642-44388642 on Build GRCh38
rs759054084 Chr.5: 44388386-44388386 on Build GRCh38
rs752464185 Chr.5: 44388411-44388411 on Build GRCh38
rs770100359 Chr.5: 44388700-44388700 on Build GRCh38
rs762714965 Chr.5: 44388529-44388529 on Build GRCh38
rs370941301 Chr.5: 44388359-44388359 on Build GRCh38
rs981726953 Chr.5: 44388722-44388722 on Build GRCh38
rs922473926 Chr.5: 44388293-44388293 on Build GRCh38
rs757950282 Chr.5: 44388415-44388415 on Build GRCh38
rs746217648 Chr.5: 44388696-44388696 on Build GRCh38
rs533983497 Chr.5: 44388304-44388304 on Build GRCh38
rs562084259 Chr.5: 44388487-44388487 on Build GRCh38
rs545941601 Chr.5: 44388422-44388422 on Build GRCh38
rs200056747 Chr.5: 44388440-44388440 on Build GRCh38
rs202211960 Chr.5: 44388521-44388521 on Build GRCh38
rs886060654 Chr.5: 44388497-44388497 on Build GRCh38
rs991153960 Chr.5: 44388350-44388350 on Build GRCh38
rs200886626 Chr.5: 44388663-44388663 on Build GRCh38
rs919539418 Chr.5: 44388745-44388745 on Build GRCh38
rs541818263 Chr.5: 44388471-44388471 on Build GRCh38
rs200535647 Chr.5: 44388557-44388557 on Build GRCh38
rs754826334 Chr.5: 44388643-44388643 on Build GRCh38
rs201168313 Chr.5: 44388553-44388553 on Build GRCh38
rs758040280 Chr.5: 44388310-44388310 on Build GRCh38
rs143865624 Chr.5: 44388692-44388692 on Build GRCh38
rs774516773 Chr.5: 44388294-44388294 on Build GRCh38
rs764969764 Chr.5: 44388387-44388387 on Build GRCh38
rs375875663 Chr.5: 44388298-44388298 on Build GRCh38
rs149009979 Chr.5: 44388467-44388467 on Build GRCh38
rs752902142 Chr.5: 44388650-44388650 on Build GRCh38
rs927001644 Chr.5: 44388290-44388290 on Build GRCh38
rs747501368 Chr.5: 44388612-44388613 on Build GRCh38
rs1006967829 Chr.5: 44388399-44388399 on Build GRCh38
rs199671312 Chr.5: 44388678-44388678 on Build GRCh38
rs201408864 Chr.5: 44388630-44388630 on Build GRCh38
rs755884036 Chr.5: 44388548-44388548 on Build GRCh38
rs199743095 Chr.5: 44388362-44388362 on Build GRCh38
rs201698360 Chr.5: 44388716-44388716 on Build GRCh38
rs199806461 Chr.5: 44388599-44388599 on Build GRCh38
rs17233910 Chr.5: 44388715-44388715 on Build GRCh38
rs144567587 Chr.5: 44388535-44388535 on Build GRCh38

View Full Product Details