Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142592403-142592951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF1
Entrez Gene ID
2246
Gene Name
fibroblast growth factor 1
Gene Aliases
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
UniGene
Hs.483635
Gene Chromosome Location
Chr.5: 142592178-142698070 on Build GRCh38
Transcripts
NM_000800.4 NM_001144892.2 NM_001144934.1 NM_001144935.1 NM_001257205.1 NM_001257206.1 NM_001257207.1 NM_001257208.1 NM_001257209.1 NM_001257210.1 NM_001257211.1 NM_001257212.1 NM_033136.3 NM_033137.2 XM_005268390.4 XM_011537607.2

SNP Detail

SNP ID Chromosome Location
rs759947308 Chr.5: 142592564-142592564 on Build GRCh38
rs555089327 Chr.5: 142592646-142592646 on Build GRCh38
rs986421828 Chr.5: 142592631-142592631 on Build GRCh38
rs374483190 Chr.5: 142592809-142592809 on Build GRCh38
rs892336489 Chr.5: 142592470-142592470 on Build GRCh38
rs931479125 Chr.5: 142592693-142592693 on Build GRCh38
rs761943239 Chr.5: 142592446-142592446 on Build GRCh38
rs193167271 Chr.5: 142592484-142592484 on Build GRCh38
rs1028063466 Chr.5: 142592863-142592863 on Build GRCh38
rs1055807974 Chr.5: 142592712-142592712 on Build GRCh38
rs397702956 Chr.5: 142592808-142592809 on Build GRCh38
rs986436670 Chr.5: 142592881-142592881 on Build GRCh38
rs1020806324 Chr.5: 142592803-142592803 on Build GRCh38
rs896413824 Chr.5: 142592766-142592766 on Build GRCh38
rs543839381 Chr.5: 142592733-142592733 on Build GRCh38
rs556184887 Chr.5: 142592926-142592926 on Build GRCh38
rs1021175505 Chr.5: 142592590-142592590 on Build GRCh38
rs563632049 Chr.5: 142592790-142592790 on Build GRCh38
rs538886886 Chr.5: 142592485-142592485 on Build GRCh38
rs1006255047 Chr.5: 142592741-142592741 on Build GRCh38
rs905936706 Chr.5: 142592720-142592720 on Build GRCh38
rs527769323 Chr.5: 142592826-142592826 on Build GRCh38
rs769754350 Chr.5: 142592516-142592516 on Build GRCh38
rs1046293453 Chr.5: 142592679-142592679 on Build GRCh38
rs1030556032 Chr.5: 142592800-142592800 on Build GRCh38
rs1000642371 Chr.5: 142592849-142592849 on Build GRCh38
rs920898844 Chr.5: 142592427-142592427 on Build GRCh38
rs953742758 Chr.5: 142592626-142592626 on Build GRCh38
rs370765374 Chr.5: 142592495-142592495 on Build GRCh38
rs1044436077 Chr.5: 142592524-142592524 on Build GRCh38
rs1057118963 Chr.5: 142592760-142592760 on Build GRCh38
rs1007690410 Chr.5: 142592827-142592827 on Build GRCh38
rs531884306 Chr.5: 142592565-142592565 on Build GRCh38
rs945314626 Chr.5: 142592678-142592678 on Build GRCh38
rs912816231 Chr.5: 142592635-142592635 on Build GRCh38
rs913560251 Chr.5: 142592671-142592671 on Build GRCh38
rs1044527092 Chr.5: 142592716-142592716 on Build GRCh38
rs773272684 Chr.5: 142592538-142592538 on Build GRCh38
rs556086885 Chr.5: 142592482-142592482 on Build GRCh38
rs756624321 Chr.5: 142592785-142592785 on Build GRCh38
rs532709555 Chr.5: 142592798-142592798 on Build GRCh38
rs766571572 Chr.5: 142592548-142592548 on Build GRCh38
rs551356625 Chr.5: 142592602-142592602 on Build GRCh38
rs62382130 Chr.5: 142592532-142592532 on Build GRCh38
rs755386094 Chr.5: 142592494-142592494 on Build GRCh38
rs932922123 Chr.5: 142592450-142592450 on Build GRCh38
rs17099022 Chr.5: 142592563-142592563 on Build GRCh38
rs995968664 Chr.5: 142592510-142592510 on Build GRCh38
rs968311258 Chr.5: 142592818-142592818 on Build GRCh38
rs1009407702 Chr.5: 142592571-142592571 on Build GRCh38
rs765552224 Chr.5: 142592816-142592816 on Build GRCh38
rs953794600 Chr.5: 142592867-142592867 on Build GRCh38
rs910239801 Chr.5: 142592431-142592431 on Build GRCh38
rs912183913 Chr.5: 142592899-142592899 on Build GRCh38
rs4440401 Chr.5: 142592529-142592529 on Build GRCh38
rs898588981 Chr.5: 142592502-142592502 on Build GRCh38
rs764644155 Chr.5: 142592793-142592793 on Build GRCh38
rs897808643 Chr.5: 142592771-142592771 on Build GRCh38
rs894604833 Chr.5: 142592727-142592727 on Build GRCh38
rs17224024 Chr.5: 142592804-142592805 on Build GRCh38
rs753241362 Chr.5: 142592675-142592675 on Build GRCh38
rs1053709760 Chr.5: 142592453-142592453 on Build GRCh38
rs754357784 Chr.5: 142592802-142592802 on Build GRCh38
rs985861366 Chr.5: 142592429-142592430 on Build GRCh38
rs927805110 Chr.5: 142592690-142592690 on Build GRCh38
rs985889146 Chr.5: 142592628-142592628 on Build GRCh38
rs17224045 Chr.5: 142592539-142592539 on Build GRCh38
rs920995353 Chr.5: 142592623-142592623 on Build GRCh38
rs1030941704 Chr.5: 142592614-142592614 on Build GRCh38
rs961667553 Chr.5: 142592915-142592915 on Build GRCh38
rs17224031 Chr.5: 142592660-142592660 on Build GRCh38
rs979129505 Chr.5: 142592615-142592615 on Build GRCh38
rs1002911783 Chr.5: 142592751-142592751 on Build GRCh38
rs558491807 Chr.5: 142592493-142592493 on Build GRCh38
rs759014367 Chr.5: 142592652-142592652 on Build GRCh38

View Full Product Details