Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142593545-142594095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF1
Entrez Gene ID
2246
Gene Name
fibroblast growth factor 1
Gene Aliases
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
UniGene
Hs.483635
Gene Chromosome Location
Chr.5: 142592178-142698070 on Build GRCh38
Transcripts
NM_000800.4 NM_001144892.2 NM_001144934.1 NM_001144935.1 NM_001257205.1 NM_001257206.1 NM_001257207.1 NM_001257208.1 NM_001257209.1 NM_001257210.1 NM_001257211.1 NM_001257212.1 NM_033136.3 NM_033137.2 XM_005268390.4 XM_011537607.2

SNP Detail

SNP ID Chromosome Location
rs17217618 Chr.5: 142593659-142593659 on Build GRCh38
rs1025498082 Chr.5: 142594076-142594076 on Build GRCh38
rs979179491 Chr.5: 142593636-142593636 on Build GRCh38
rs542022817 Chr.5: 142593851-142593851 on Build GRCh38
rs980328178 Chr.5: 142593571-142593571 on Build GRCh38
rs918406985 Chr.5: 142593830-142593830 on Build GRCh38
rs557412026 Chr.5: 142593709-142593709 on Build GRCh38
rs1020936833 Chr.5: 142594067-142594067 on Build GRCh38
rs774691952 Chr.5: 142593788-142593788 on Build GRCh38
rs929783372 Chr.5: 142593997-142593997 on Build GRCh38
rs528727010 Chr.5: 142593816-142593816 on Build GRCh38
rs1055429598 Chr.5: 142594059-142594059 on Build GRCh38
rs112939431 Chr.5: 142593818-142593818 on Build GRCh38
rs967600395 Chr.5: 142594072-142594072 on Build GRCh38
rs1014481041 Chr.5: 142594066-142594066 on Build GRCh38
rs943625418 Chr.5: 142593613-142593613 on Build GRCh38
rs34000 Chr.5: 142593936-142593936 on Build GRCh38
rs148171786 Chr.5: 142593613-142593613 on Build GRCh38
rs902295668 Chr.5: 142593696-142593696 on Build GRCh38
rs1011040976 Chr.5: 142593649-142593649 on Build GRCh38
rs757002483 Chr.5: 142593825-142593825 on Build GRCh38
rs926025172 Chr.5: 142593938-142593938 on Build GRCh38
rs896902475 Chr.5: 142593765-142593765 on Build GRCh38
rs1001241390 Chr.5: 142594054-142594054 on Build GRCh38
rs760035567 Chr.5: 142593801-142593801 on Build GRCh38
rs529757070 Chr.5: 142593893-142593893 on Build GRCh38
rs943932917 Chr.5: 142593964-142593964 on Build GRCh38
rs761110393 Chr.5: 142594000-142594000 on Build GRCh38
rs1040843167 Chr.5: 142593764-142593764 on Build GRCh38
rs1034704886 Chr.5: 142593984-142593984 on Build GRCh38
rs561907876 Chr.5: 142593861-142593861 on Build GRCh38
rs987533924 Chr.5: 142593852-142593852 on Build GRCh38
rs59014099 Chr.5: 142593991-142593991 on Build GRCh38
rs17224010 Chr.5: 142593731-142593731 on Build GRCh38
rs993005265 Chr.5: 142593598-142593598 on Build GRCh38
rs929434421 Chr.5: 142593847-142593847 on Build GRCh38
rs776627640 Chr.5: 142593797-142593797 on Build GRCh38
rs1034673800 Chr.5: 142593755-142593755 on Build GRCh38
rs574993539 Chr.5: 142593641-142593641 on Build GRCh38
rs943322799 Chr.5: 142593894-142593894 on Build GRCh38
rs960225942 Chr.5: 142593593-142593593 on Build GRCh38
rs1020417176 Chr.5: 142593910-142593910 on Build GRCh38
rs78257478 Chr.5: 142594064-142594064 on Build GRCh38
rs1041287415 Chr.5: 142593965-142593965 on Build GRCh38
rs1034583798 Chr.5: 142593578-142593578 on Build GRCh38
rs141826868 Chr.5: 142593762-142593762 on Build GRCh38
rs953412914 Chr.5: 142593940-142593940 on Build GRCh38
rs946114242 Chr.5: 142593634-142593634 on Build GRCh38
rs888138240 Chr.5: 142593798-142593798 on Build GRCh38
rs558510755 Chr.5: 142593797-142593797 on Build GRCh38
rs748097101 Chr.5: 142593960-142593960 on Build GRCh38
rs34001 Chr.5: 142594034-142594034 on Build GRCh38
rs753054937 Chr.5: 142593962-142593962 on Build GRCh38
rs994476961 Chr.5: 142594071-142594071 on Build GRCh38
rs967885358 Chr.5: 142593920-142593920 on Build GRCh38
rs960276487 Chr.5: 142593841-142593841 on Build GRCh38
rs1003315301 Chr.5: 142593977-142593977 on Build GRCh38
rs33999 Chr.5: 142593784-142593784 on Build GRCh38
rs17224003 Chr.5: 142593770-142593770 on Build GRCh38
rs569580970 Chr.5: 142593942-142593942 on Build GRCh38

View Full Product Details