Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157269175-157269771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1

SNP Detail

SNP ID Chromosome Location
rs73302141 Chr.5: 157269677-157269677 on Build GRCh38
rs574373785 Chr.5: 157269655-157269655 on Build GRCh38
rs898629066 Chr.5: 157269740-157269740 on Build GRCh38
rs558375868 Chr.5: 157269567-157269567 on Build GRCh38
rs1030443015 Chr.5: 157269736-157269736 on Build GRCh38
rs569453739 Chr.5: 157269356-157269356 on Build GRCh38
rs531889954 Chr.5: 157269405-157269405 on Build GRCh38
rs764914386 Chr.5: 157269621-157269621 on Build GRCh38
rs929295097 Chr.5: 157269222-157269222 on Build GRCh38
rs1009621507 Chr.5: 157269438-157269438 on Build GRCh38
rs188054066 Chr.5: 157269724-157269724 on Build GRCh38
rs754685458 Chr.5: 157269446-157269446 on Build GRCh38
rs951335132 Chr.5: 157269352-157269352 on Build GRCh38
rs368840333 Chr.5: 157269342-157269342 on Build GRCh38
rs5872507 Chr.5: 157269588-157269589 on Build GRCh38
rs1053386387 Chr.5: 157269663-157269663 on Build GRCh38
rs1030081751 Chr.5: 157269442-157269442 on Build GRCh38
rs537710418 Chr.5: 157269196-157269197 on Build GRCh38
rs956375345 Chr.5: 157269392-157269392 on Build GRCh38
rs919145832 Chr.5: 157269538-157269538 on Build GRCh38
rs554599611 Chr.5: 157269620-157269620 on Build GRCh38
rs568372445 Chr.5: 157269511-157269511 on Build GRCh38
rs1013094293 Chr.5: 157269253-157269253 on Build GRCh38
rs575083674 Chr.5: 157269576-157269576 on Build GRCh38
rs538522375 Chr.5: 157269563-157269563 on Build GRCh38
rs963162296 Chr.5: 157269532-157269532 on Build GRCh38
rs146714317 Chr.5: 157269536-157269536 on Build GRCh38
rs375891852 Chr.5: 157269510-157269510 on Build GRCh38
rs1057433119 Chr.5: 157269556-157269556 on Build GRCh38
rs1047574260 Chr.5: 157269490-157269490 on Build GRCh38
rs935946828 Chr.5: 157269550-157269550 on Build GRCh38
rs559880446 Chr.5: 157269694-157269694 on Build GRCh38
rs895850015 Chr.5: 157269250-157269250 on Build GRCh38
rs1057236302 Chr.5: 157269229-157269229 on Build GRCh38
rs912901697 Chr.5: 157269609-157269610 on Build GRCh38
rs149802355 Chr.5: 157269490-157269491 on Build GRCh38
rs995939364 Chr.5: 157269264-157269264 on Build GRCh38
rs767007 Chr.5: 157269485-157269485 on Build GRCh38
rs948856512 Chr.5: 157269584-157269584 on Build GRCh38
rs527541255 Chr.5: 157269370-157269370 on Build GRCh38
rs954400133 Chr.5: 157269739-157269739 on Build GRCh38
rs767006 Chr.5: 157269413-157269413 on Build GRCh38
rs534168810 Chr.5: 157269523-157269523 on Build GRCh38
rs566791942 Chr.5: 157269547-157269547 on Build GRCh38
rs1054278758 Chr.5: 157269605-157269606 on Build GRCh38
rs1044994034 Chr.5: 157269282-157269282 on Build GRCh38
rs756937412 Chr.5: 157269196-157269196 on Build GRCh38
rs917250827 Chr.5: 157269579-157269579 on Build GRCh38
rs73814079 Chr.5: 157269617-157269617 on Build GRCh38
rs954576153 Chr.5: 157269482-157269482 on Build GRCh38

View Full Product Details