Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83917797-83918239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R4
Entrez Gene ID
51167
Gene Name
cytochrome b5 reductase 4
Gene Aliases
NCB5OR, cb5/cb5R, dJ676J13.1
UniGene
Hs.5741
Gene Chromosome Location
Chr.6: 83859651-83960427 on Build GRCh38
Transcripts
NM_016230.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082121 c.462C>T p.S154S Chr.6: 83918021-83918021 on Build GRCh38
6175171 c.475A>G p.T159A Chr.6: 83918034-83918034 on Build GRCh38
3630279 c.479T>G p.L160R Chr.6: 83918038-83918038 on Build GRCh38
1697595 c.502C>T p.P168S Chr.6: 83918061-83918061 on Build GRCh38
4402221 c.460A>G p.S154G Chr.6: 83918019-83918019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045653168 Chr.6: 83918041-83918041 on Build GRCh38
rs150785197 Chr.6: 83917889-83917889 on Build GRCh38
rs568242390 Chr.6: 83918117-83918117 on Build GRCh38
rs774237554 Chr.6: 83917948-83917948 on Build GRCh38
rs375928964 Chr.6: 83917943-83917943 on Build GRCh38
rs554094102 Chr.6: 83917914-83917914 on Build GRCh38
rs778928130 Chr.6: 83918101-83918101 on Build GRCh38
rs758410641 Chr.6: 83918065-83918065 on Build GRCh38
rs535689350 Chr.6: 83918118-83918118 on Build GRCh38
rs372240764 Chr.6: 83917964-83917964 on Build GRCh38
rs551778438 Chr.6: 83918050-83918050 on Build GRCh38
rs539808404 Chr.6: 83918214-83918214 on Build GRCh38
rs1009048130 Chr.6: 83918193-83918193 on Build GRCh38
rs892191959 Chr.6: 83917997-83917997 on Build GRCh38
rs1009534650 Chr.6: 83917910-83917910 on Build GRCh38
rs62419732 Chr.6: 83918021-83918021 on Build GRCh38
rs562284906 Chr.6: 83917832-83917832 on Build GRCh38
rs773984745 Chr.6: 83918004-83918004 on Build GRCh38
rs569196681 Chr.6: 83918184-83918184 on Build GRCh38
rs888952754 Chr.6: 83918071-83918071 on Build GRCh38
rs983309117 Chr.6: 83918005-83918005 on Build GRCh38
rs563992791 Chr.6: 83918066-83918066 on Build GRCh38
rs370959517 Chr.6: 83918072-83918072 on Build GRCh38
rs765159688 Chr.6: 83917988-83917988 on Build GRCh38
rs750502574 Chr.6: 83918055-83918055 on Build GRCh38
rs200526921 Chr.6: 83918108-83918108 on Build GRCh38
rs572692453 Chr.6: 83917933-83917933 on Build GRCh38
rs757104649 Chr.6: 83917947-83917947 on Build GRCh38
rs751762835 Chr.6: 83918075-83918075 on Build GRCh38
rs770988539 Chr.6: 83918099-83918099 on Build GRCh38
rs145537929 Chr.6: 83918049-83918049 on Build GRCh38
rs771855490 Chr.6: 83918014-83918014 on Build GRCh38
rs775227179 Chr.6: 83918015-83918015 on Build GRCh38
rs759329551 Chr.6: 83918009-83918009 on Build GRCh38
rs902709585 Chr.6: 83918190-83918190 on Build GRCh38
rs777340060 Chr.6: 83917828-83917828 on Build GRCh38
rs918668325 Chr.6: 83917846-83917846 on Build GRCh38
rs376263148 Chr.6: 83918025-83918025 on Build GRCh38
rs984280964 Chr.6: 83917849-83917849 on Build GRCh38
rs1018811850 Chr.6: 83918097-83918101 on Build GRCh38
rs1042574044 Chr.6: 83918121-83918121 on Build GRCh38
rs749241943 Chr.6: 83918094-83918094 on Build GRCh38
rs1011081908 Chr.6: 83917859-83917859 on Build GRCh38
rs1026082425 Chr.6: 83918202-83918202 on Build GRCh38
rs755900651 Chr.6: 83918088-83918088 on Build GRCh38
rs754346231 Chr.6: 83918035-83918035 on Build GRCh38
rs761812600 Chr.6: 83918101-83918102 on Build GRCh38
rs61763821 Chr.6: 83917874-83917874 on Build GRCh38
rs948249675 Chr.6: 83917882-83917882 on Build GRCh38
rs7773061 Chr.6: 83918091-83918091 on Build GRCh38
rs930077383 Chr.6: 83917847-83917847 on Build GRCh38
rs144658748 Chr.6: 83917938-83917938 on Build GRCh38
rs550847499 Chr.6: 83918155-83918155 on Build GRCh38
rs1045343114 Chr.6: 83917905-83917905 on Build GRCh38
rs544491825 Chr.6: 83917908-83917908 on Build GRCh38
rs150033774 Chr.6: 83918033-83918033 on Build GRCh38
rs200538689 Chr.6: 83917976-83917976 on Build GRCh38
rs372530372 Chr.6: 83917916-83917916 on Build GRCh38
rs770612795 Chr.6: 83917991-83917991 on Build GRCh38
rs749216986 Chr.6: 83917987-83917987 on Build GRCh38
rs964395443 Chr.6: 83917928-83917928 on Build GRCh38
rs777601401 Chr.6: 83917982-83917982 on Build GRCh38
rs745763836 Chr.6: 83918007-83918007 on Build GRCh38

View Full Product Details