Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 93240487-93241041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA7
Entrez Gene ID
2045
Gene Name
EPH receptor A7
Gene Aliases
EHK-3, EHK3, EK11, HEK11
UniGene
Hs.73962
Gene Chromosome Location
Chr.6: 93240020-93419687 on Build GRCh38
Transcripts
NM_001288629.1 NM_004440.3 XM_005248669.2 XM_017010365.1

SNP Detail

SNP ID Chromosome Location
rs1011370398 Chr.6: 93240636-93240636 on Build GRCh38
rs377056780 Chr.6: 93240846-93240846 on Build GRCh38
rs944504284 Chr.6: 93240848-93240848 on Build GRCh38
rs192616485 Chr.6: 93240529-93240529 on Build GRCh38
rs993161795 Chr.6: 93240980-93240980 on Build GRCh38
rs1023824976 Chr.6: 93240636-93240636 on Build GRCh38
rs945325131 Chr.6: 93240626-93240626 on Build GRCh38
rs142329466 Chr.6: 93240747-93240748 on Build GRCh38
rs924516467 Chr.6: 93240849-93240849 on Build GRCh38
rs998175766 Chr.6: 93240784-93240784 on Build GRCh38
rs546195509 Chr.6: 93240625-93240625 on Build GRCh38
rs138297561 Chr.6: 93240886-93240886 on Build GRCh38
rs543439076 Chr.6: 93240527-93240527 on Build GRCh38
rs982356763 Chr.6: 93240699-93240699 on Build GRCh38
rs539777307 Chr.6: 93241003-93241003 on Build GRCh38
rs183876070 Chr.6: 93240825-93240825 on Build GRCh38
rs938584358 Chr.6: 93240522-93240522 on Build GRCh38
rs1031539926 Chr.6: 93240594-93240594 on Build GRCh38
rs528743058 Chr.6: 93240737-93240737 on Build GRCh38
rs532104624 Chr.6: 93240558-93240558 on Build GRCh38
rs763224115 Chr.6: 93241013-93241013 on Build GRCh38
rs985574095 Chr.6: 93240768-93240768 on Build GRCh38
rs62414212 Chr.6: 93240876-93240876 on Build GRCh38
rs775983195 Chr.6: 93240898-93240898 on Build GRCh38
rs868289191 Chr.6: 93240885-93240885 on Build GRCh38
rs560040839 Chr.6: 93240707-93240707 on Build GRCh38
rs903917257 Chr.6: 93240705-93240705 on Build GRCh38
rs971748832 Chr.6: 93240714-93240714 on Build GRCh38
rs950393039 Chr.6: 93240806-93240806 on Build GRCh38
rs927100847 Chr.6: 93240924-93240924 on Build GRCh38
rs900779579 Chr.6: 93240748-93240748 on Build GRCh38
rs896471202 Chr.6: 93240609-93240609 on Build GRCh38
rs986255849 Chr.6: 93240761-93240761 on Build GRCh38
rs969497471 Chr.6: 93240694-93240694 on Build GRCh38
rs558429473 Chr.6: 93241011-93241011 on Build GRCh38
rs34175783 Chr.6: 93240788-93240789 on Build GRCh38
rs1030030878 Chr.6: 93240794-93240794 on Build GRCh38
rs960023090 Chr.6: 93240972-93240972 on Build GRCh38
rs71542006 Chr.6: 93240726-93240727 on Build GRCh38
rs961724593 Chr.6: 93240711-93240711 on Build GRCh38
rs770384012 Chr.6: 93240536-93240536 on Build GRCh38
rs867988170 Chr.6: 93240689-93240689 on Build GRCh38
rs570776061 Chr.6: 93240923-93240923 on Build GRCh38
rs1018178471 Chr.6: 93240740-93240740 on Build GRCh38
rs1042499017 Chr.6: 93240704-93240704 on Build GRCh38
rs10806473 Chr.6: 93240848-93240848 on Build GRCh38

View Full Product Details