Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49488424-49488818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPQ
Entrez Gene ID
55166
Gene Name
centromere protein Q
Gene Aliases
C6orf139, CENP-Q
UniGene
Hs.88663
Gene Chromosome Location
Chr.6: 49463327-49493107 on Build GRCh38
Transcripts
NM_018132.3 XM_005249205.1 XM_011514723.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6106923 c.585G>C p.E195D Chr.6: 49488459-49488459 on Build GRCh38
3628252 c.554T>C p.I185T Chr.6: 49488428-49488428 on Build GRCh38
4504060 c.655C>T p.L219F Chr.6: 49488664-49488664 on Build GRCh38
306943 c.570G>T p.V190V Chr.6: 49488444-49488444 on Build GRCh38
5466877 c.597+8T>G p.? Chr.6: 49488479-49488479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747940505 Chr.6: 49488711-49488711 on Build GRCh38
rs779541033 Chr.6: 49488587-49488587 on Build GRCh38
rs1026088831 Chr.6: 49488620-49488620 on Build GRCh38
rs535758644 Chr.6: 49488791-49488791 on Build GRCh38
rs765700964 Chr.6: 49488571-49488572 on Build GRCh38
rs537129987 Chr.6: 49488545-49488545 on Build GRCh38
rs757805814 Chr.6: 49488683-49488683 on Build GRCh38
rs904400288 Chr.6: 49488561-49488561 on Build GRCh38
rs753144692 Chr.6: 49488519-49488519 on Build GRCh38
rs746597675 Chr.6: 49488695-49488695 on Build GRCh38
rs145201362 Chr.6: 49488656-49488656 on Build GRCh38
rs768753866 Chr.6: 49488459-49488459 on Build GRCh38
rs554980196 Chr.6: 49488555-49488555 on Build GRCh38
rs769610726 Chr.6: 49488609-49488609 on Build GRCh38
rs771803855 Chr.6: 49488583-49488583 on Build GRCh38
rs572606702 Chr.6: 49488779-49488779 on Build GRCh38
rs921716922 Chr.6: 49488719-49488719 on Build GRCh38
rs750612401 Chr.6: 49488490-49488490 on Build GRCh38
rs373627672 Chr.6: 49488494-49488494 on Build GRCh38
rs768522874 Chr.6: 49488592-49488592 on Build GRCh38
rs373972605 Chr.6: 49488705-49488705 on Build GRCh38
rs756996355 Chr.6: 49488735-49488735 on Build GRCh38
rs535770550 Chr.6: 49488684-49488684 on Build GRCh38
rs745459104 Chr.6: 49488447-49488447 on Build GRCh38
rs751997954 Chr.6: 49488497-49488497 on Build GRCh38
rs746830717 Chr.6: 49488591-49488591 on Build GRCh38
rs759884825 Chr.6: 49488455-49488456 on Build GRCh38
rs1055465147 Chr.6: 49488758-49488758 on Build GRCh38
rs933011375 Chr.6: 49488529-49488529 on Build GRCh38
rs373167722 Chr.6: 49488465-49488465 on Build GRCh38
rs774512323 Chr.6: 49488638-49488638 on Build GRCh38
rs564696569 Chr.6: 49488785-49488785 on Build GRCh38
rs764488347 Chr.6: 49488665-49488665 on Build GRCh38
rs756673195 Chr.6: 49488664-49488664 on Build GRCh38
rs766491181 Chr.6: 49488496-49488496 on Build GRCh38
rs775415028 Chr.6: 49488454-49488454 on Build GRCh38
rs762913311 Chr.6: 49488622-49488622 on Build GRCh38
rs759911830 Chr.6: 49488644-49488644 on Build GRCh38
rs891733540 Chr.6: 49488544-49488544 on Build GRCh38
rs778553504 Chr.6: 49488564-49488564 on Build GRCh38
rs751201150 Chr.6: 49488662-49488663 on Build GRCh38
rs184852635 Chr.6: 49488784-49488784 on Build GRCh38
rs781683360 Chr.6: 49488518-49488518 on Build GRCh38
rs745359247 Chr.6: 49488580-49488580 on Build GRCh38
rs553995796 Chr.6: 49488712-49488712 on Build GRCh38
rs569698110 Chr.6: 49488450-49488450 on Build GRCh38
rs773396952 Chr.6: 49488615-49488615 on Build GRCh38
rs367656058 Chr.6: 49488556-49488556 on Build GRCh38
rs912929311 Chr.6: 49488782-49488782 on Build GRCh38
rs369521294 Chr.6: 49488594-49488594 on Build GRCh38
rs542704670 Chr.6: 49488645-49488645 on Build GRCh38
rs140237607 Chr.6: 49488629-49488629 on Build GRCh38
rs202062394 Chr.6: 49488601-49488601 on Build GRCh38
rs755013031 Chr.6: 49488547-49488547 on Build GRCh38
rs549443844 Chr.6: 49488731-49488731 on Build GRCh38
rs1051840394 Chr.6: 49488537-49488537 on Build GRCh38
rs765300057 Chr.6: 49488471-49488471 on Build GRCh38
rs376329132 Chr.6: 49488478-49488478 on Build GRCh38
rs754821181 Chr.6: 49488701-49488701 on Build GRCh38
rs35246867 Chr.6: 49488493-49488494 on Build GRCh38
rs964207345 Chr.6: 49488693-49488693 on Build GRCh38
rs772985397 Chr.6: 49488723-49488723 on Build GRCh38
rs951346630 Chr.6: 49488623-49488623 on Build GRCh38
rs749988875 Chr.6: 49488671-49488671 on Build GRCh38
rs760521971 Chr.6: 49488456-49488456 on Build GRCh38
rs755299407 Chr.6: 49488510-49488510 on Build GRCh38

View Full Product Details