Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53268167-53268700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL5
Entrez Gene ID
60481
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
HELO1, SCA38, dJ483K16.1
UniGene
Hs.520189
Gene Chromosome Location
Chr.6: 53267398-53349179 on Build GRCh38
Transcripts
NM_001242828.1 NM_001242830.1 NM_001301856.1 NM_021814.4

SNP Detail

SNP ID Chromosome Location
rs146109873 Chr.6: 53268225-53268225 on Build GRCh38
rs956267546 Chr.6: 53268590-53268590 on Build GRCh38
rs1032257596 Chr.6: 53268589-53268589 on Build GRCh38
rs1035767723 Chr.6: 53268231-53268231 on Build GRCh38
rs754722098 Chr.6: 53268297-53268297 on Build GRCh38
rs763609128 Chr.6: 53268383-53268383 on Build GRCh38
rs866260162 Chr.6: 53268323-53268323 on Build GRCh38
rs968840293 Chr.6: 53268316-53268316 on Build GRCh38
rs796159129 Chr.6: 53268420-53268420 on Build GRCh38
rs931105956 Chr.6: 53268414-53268414 on Build GRCh38
rs921213351 Chr.6: 53268409-53268409 on Build GRCh38
rs140131369 Chr.6: 53268255-53268255 on Build GRCh38
rs371438650 Chr.6: 53268547-53268547 on Build GRCh38
rs957738927 Chr.6: 53268559-53268559 on Build GRCh38
rs1013112869 Chr.6: 53268269-53268269 on Build GRCh38
rs895468002 Chr.6: 53268488-53268488 on Build GRCh38
rs146708366 Chr.6: 53268548-53268548 on Build GRCh38
rs11555161 Chr.6: 53268213-53268213 on Build GRCh38
rs143266597 Chr.6: 53268341-53268341 on Build GRCh38
rs41273872 Chr.6: 53268400-53268400 on Build GRCh38
rs931160255 Chr.6: 53268194-53268194 on Build GRCh38
rs79049968 Chr.6: 53268420-53268421 on Build GRCh38
rs896034790 Chr.6: 53268244-53268244 on Build GRCh38
rs139081058 Chr.6: 53268595-53268595 on Build GRCh38
rs1000294992 Chr.6: 53268567-53268567 on Build GRCh38
rs182388209 Chr.6: 53268426-53268426 on Build GRCh38
rs1050463070 Chr.6: 53268319-53268319 on Build GRCh38
rs911875282 Chr.6: 53268373-53268373 on Build GRCh38
rs886823671 Chr.6: 53268427-53268427 on Build GRCh38
rs1013900929 Chr.6: 53268422-53268422 on Build GRCh38
rs1031725351 Chr.6: 53268332-53268332 on Build GRCh38
rs745458407 Chr.6: 53268419-53268420 on Build GRCh38
rs1048706 Chr.6: 53268210-53268210 on Build GRCh38
rs932098184 Chr.6: 53268315-53268321 on Build GRCh38
rs1006181275 Chr.6: 53268352-53268352 on Build GRCh38
rs375923542 Chr.6: 53268427-53268427 on Build GRCh38
rs987840147 Chr.6: 53268561-53268561 on Build GRCh38
rs1012560127 Chr.6: 53268493-53268493 on Build GRCh38
rs887818140 Chr.6: 53268327-53268327 on Build GRCh38
rs542385881 Chr.6: 53268315-53268315 on Build GRCh38
rs777581304 Chr.6: 53268439-53268439 on Build GRCh38
rs987449777 Chr.6: 53268366-53268366 on Build GRCh38
rs956336429 Chr.6: 53268351-53268351 on Build GRCh38
rs572371808 Chr.6: 53268421-53268421 on Build GRCh38
rs41273874 Chr.6: 53268436-53268436 on Build GRCh38
rs1009395756 Chr.6: 53268660-53268660 on Build GRCh38
rs1024837403 Chr.6: 53268425-53268426 on Build GRCh38
rs901991896 Chr.6: 53268309-53268309 on Build GRCh38
rs1044614724 Chr.6: 53268546-53268546 on Build GRCh38
rs539507173 Chr.6: 53268224-53268224 on Build GRCh38
rs975312004 Chr.6: 53268406-53268406 on Build GRCh38

View Full Product Details