Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75091003-75091526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
UniGene
Hs.101302
Gene Chromosome Location
Chr.6: 75084326-75206262 on Build GRCh38
Transcripts
NM_004370.5 NM_080645.2 XM_011535434.1 XM_011535435.1 XM_011535436.2 XM_017010252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6107748 c.8676A>G p.K2892K Chr.6: 75091499-75091499 on Build GRCh38
3629763 c.8661G>A p.G2887G Chr.6: 75091514-75091514 on Build GRCh38
4511459 c.8696C>T p.A2899V Chr.6: 75091379-75091379 on Build GRCh38
7202932 c.8706C>A p.N2902K Chr.6: 75091369-75091369 on Build GRCh38
6905022 c.8740C>A p.Q2914K Chr.6: 75091335-75091335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772735275 Chr.6: 75091291-75091294 on Build GRCh38
rs746952026 Chr.6: 75091291-75091291 on Build GRCh38
rs371386011 Chr.6: 75091193-75091193 on Build GRCh38
rs781729406 Chr.6: 75091153-75091153 on Build GRCh38
rs368910082 Chr.6: 75091477-75091477 on Build GRCh38
rs191411385 Chr.6: 75091465-75091465 on Build GRCh38
rs924650208 Chr.6: 75091190-75091190 on Build GRCh38
rs770795119 Chr.6: 75091420-75091423 on Build GRCh38
rs540177129 Chr.6: 75091214-75091214 on Build GRCh38
rs777667204 Chr.6: 75091434-75091434 on Build GRCh38
rs996988585 Chr.6: 75091486-75091486 on Build GRCh38
rs964265590 Chr.6: 75091092-75091092 on Build GRCh38
rs758646803 Chr.6: 75091298-75091298 on Build GRCh38
rs542833805 Chr.6: 75091164-75091164 on Build GRCh38
rs189211970 Chr.6: 75091439-75091439 on Build GRCh38
rs747884941 Chr.6: 75091305-75091305 on Build GRCh38
rs756976210 Chr.6: 75091224-75091225 on Build GRCh38
rs750491475 Chr.6: 75091388-75091388 on Build GRCh38
rs183918103 Chr.6: 75091378-75091378 on Build GRCh38
rs745441898 Chr.6: 75091471-75091471 on Build GRCh38
rs775109401 Chr.6: 75091454-75091454 on Build GRCh38
rs567716939 Chr.6: 75091396-75091396 on Build GRCh38
rs746233474 Chr.6: 75091206-75091206 on Build GRCh38
rs3765496 Chr.6: 75091215-75091215 on Build GRCh38
rs773958923 Chr.6: 75091352-75091352 on Build GRCh38
rs745307208 Chr.6: 75091448-75091448 on Build GRCh38
rs747296733 Chr.6: 75091397-75091397 on Build GRCh38
rs763196298 Chr.6: 75091471-75091472 on Build GRCh38
rs763115734 Chr.6: 75091482-75091482 on Build GRCh38
rs1047419746 Chr.6: 75091363-75091363 on Build GRCh38
rs776535931 Chr.6: 75091445-75091446 on Build GRCh38
rs1001691731 Chr.6: 75091457-75091457 on Build GRCh38
rs770849364 Chr.6: 75091443-75091443 on Build GRCh38
rs769648368 Chr.6: 75091308-75091308 on Build GRCh38
rs367575408 Chr.6: 75091129-75091129 on Build GRCh38
rs575461502 Chr.6: 75091140-75091140 on Build GRCh38
rs201582509 Chr.6: 75091312-75091312 on Build GRCh38
rs1024414351 Chr.6: 75091417-75091417 on Build GRCh38
rs980219263 Chr.6: 75091048-75091048 on Build GRCh38
rs516533 Chr.6: 75091077-75091077 on Build GRCh38
rs749317169 Chr.6: 75091330-75091330 on Build GRCh38
rs760970675 Chr.6: 75091368-75091368 on Build GRCh38
rs532425473 Chr.6: 75091257-75091257 on Build GRCh38
rs192087820 Chr.6: 75091293-75091293 on Build GRCh38
rs188452781 Chr.6: 75091186-75091186 on Build GRCh38
rs375435011 Chr.6: 75091119-75091119 on Build GRCh38
rs775597496 Chr.6: 75091466-75091467 on Build GRCh38
rs900259952 Chr.6: 75091478-75091478 on Build GRCh38
rs771583873 Chr.6: 75091452-75091452 on Build GRCh38
rs759392551 Chr.6: 75091451-75091454 on Build GRCh38
rs749754621 Chr.6: 75091394-75091394 on Build GRCh38
rs777006427 Chr.6: 75091466-75091466 on Build GRCh38
rs948553114 Chr.6: 75091246-75091246 on Build GRCh38
rs757269039 Chr.6: 75091280-75091280 on Build GRCh38
rs529883980 Chr.6: 75091301-75091301 on Build GRCh38
rs778849073 Chr.6: 75091447-75091447 on Build GRCh38
rs376238729 Chr.6: 75091332-75091332 on Build GRCh38
rs989433982 Chr.6: 75091114-75091114 on Build GRCh38
rs754295132 Chr.6: 75091374-75091374 on Build GRCh38
rs762116400 Chr.6: 75091473-75091473 on Build GRCh38
rs765050760 Chr.6: 75091455-75091456 on Build GRCh38
rs779113685 Chr.6: 75091288-75091288 on Build GRCh38
rs553097287 Chr.6: 75091089-75091089 on Build GRCh38
rs759065663 Chr.6: 75091361-75091361 on Build GRCh38
rs1024780633 Chr.6: 75091025-75091025 on Build GRCh38
rs755860124 Chr.6: 75091403-75091403 on Build GRCh38
rs544211538 Chr.6: 75091277-75091277 on Build GRCh38
rs970226708 Chr.6: 75091029-75091029 on Build GRCh38
rs780369344 Chr.6: 75091395-75091395 on Build GRCh38
rs796108876 Chr.6: 75091200-75091200 on Build GRCh38
rs572860110 Chr.6: 75091197-75091197 on Build GRCh38
rs780457606 Chr.6: 75091380-75091380 on Build GRCh38
rs149516438 Chr.6: 75091126-75091126 on Build GRCh38
rs371399251 Chr.6: 75091362-75091362 on Build GRCh38
rs968946844 Chr.6: 75091136-75091136 on Build GRCh38

View Full Product Details