Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106186251-106186816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG5
Entrez Gene ID
9474
Gene Name
autophagy related 5
Gene Aliases
APG5, APG5-LIKE, APG5L, ASP, hAPG5
UniGene
Hs.486063
Gene Chromosome Location
Chr.6: 106184476-106325820 on Build GRCh38
Transcripts
NM_001286106.1 NM_001286107.1 NM_001286108.1 NM_001286111.1 NM_004849.3 NR_104402.1 NR_104403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3154369 c.704delA p.K235fs*4 Chr.6: 106186664-106186664 on Build GRCh38
3154368 c.705G>T p.K235N Chr.6: 106186663-106186663 on Build GRCh38
7143213 c.774G>C p.L258L Chr.6: 106186594-106186594 on Build GRCh38
5756718 c.697G>A p.E233K Chr.6: 106186671-106186671 on Build GRCh38
3857249 c.782C>T p.P261L Chr.6: 106186586-106186586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138207825 Chr.6: 106186513-106186513 on Build GRCh38
rs140455408 Chr.6: 106186397-106186397 on Build GRCh38
rs1045854658 Chr.6: 106186309-106186309 on Build GRCh38
rs577930222 Chr.6: 106186521-106186521 on Build GRCh38
rs185329702 Chr.6: 106186704-106186704 on Build GRCh38
rs376181818 Chr.6: 106186500-106186500 on Build GRCh38
rs1016540259 Chr.6: 106186475-106186475 on Build GRCh38
rs748346595 Chr.6: 106186598-106186598 on Build GRCh38
rs866313330 Chr.6: 106186313-106186313 on Build GRCh38
rs778187845 Chr.6: 106186718-106186718 on Build GRCh38
rs76183704 Chr.6: 106186298-106186298 on Build GRCh38
rs148318473 Chr.6: 106186337-106186337 on Build GRCh38
rs760275075 Chr.6: 106186673-106186673 on Build GRCh38
rs1055011670 Chr.6: 106186363-106186363 on Build GRCh38
rs143781161 Chr.6: 106186782-106186782 on Build GRCh38
rs757282744 Chr.6: 106186769-106186769 on Build GRCh38
rs934860513 Chr.6: 106186334-106186334 on Build GRCh38
rs934767863 Chr.6: 106186581-106186581 on Build GRCh38
rs978167288 Chr.6: 106186787-106186787 on Build GRCh38
rs959228606 Chr.6: 106186485-106186485 on Build GRCh38
rs563775563 Chr.6: 106186633-106186633 on Build GRCh38
rs561574434 Chr.6: 106186757-106186757 on Build GRCh38
rs751072722 Chr.6: 106186501-106186501 on Build GRCh38
rs1019841454 Chr.6: 106186705-106186705 on Build GRCh38
rs898608321 Chr.6: 106186670-106186670 on Build GRCh38
rs749781045 Chr.6: 106186297-106186298 on Build GRCh38
rs748337769 Chr.6: 106186708-106186708 on Build GRCh38
rs954332901 Chr.6: 106186275-106186275 on Build GRCh38
rs903234655 Chr.6: 106186333-106186333 on Build GRCh38
rs147026203 Chr.6: 106186430-106186430 on Build GRCh38
rs143306121 Chr.6: 106186317-106186317 on Build GRCh38
rs749734336 Chr.6: 106186625-106186625 on Build GRCh38
rs977759554 Chr.6: 106186392-106186392 on Build GRCh38
rs770143894 Chr.6: 106186617-106186617 on Build GRCh38
rs749682741 Chr.6: 106186724-106186724 on Build GRCh38
rs949307058 Chr.6: 106186467-106186467 on Build GRCh38
rs773836998 Chr.6: 106186660-106186660 on Build GRCh38
rs1039257418 Chr.6: 106186427-106186427 on Build GRCh38
rs990688397 Chr.6: 106186444-106186444 on Build GRCh38
rs543456136 Chr.6: 106186539-106186539 on Build GRCh38
rs753585749 Chr.6: 106186685-106186685 on Build GRCh38
rs553100488 Chr.6: 106186614-106186614 on Build GRCh38
rs902074127 Chr.6: 106186699-106186699 on Build GRCh38
rs779507869 Chr.6: 106186718-106186725 on Build GRCh38
rs539148837 Chr.6: 106186271-106186271 on Build GRCh38
rs886092534 Chr.6: 106186781-106186781 on Build GRCh38
rs916415841 Chr.6: 106186448-106186448 on Build GRCh38
rs369833045 Chr.6: 106186532-106186532 on Build GRCh38
rs914763225 Chr.6: 106186630-106186630 on Build GRCh38
rs372102128 Chr.6: 106186688-106186688 on Build GRCh38
rs759096052 Chr.6: 106186663-106186663 on Build GRCh38
rs781601442 Chr.6: 106186585-106186585 on Build GRCh38
rs752467751 Chr.6: 106186522-106186522 on Build GRCh38
rs966789861 Chr.6: 106186353-106186353 on Build GRCh38
rs757158272 Chr.6: 106186548-106186548 on Build GRCh38
rs142878178 Chr.6: 106186672-106186672 on Build GRCh38
rs1032001446 Chr.6: 106186788-106186788 on Build GRCh38
rs1008257091 Chr.6: 106186417-106186417 on Build GRCh38
rs41292420 Chr.6: 106186694-106186694 on Build GRCh38
rs556179726 Chr.6: 106186301-106186301 on Build GRCh38
rs1047874045 Chr.6: 106186758-106186758 on Build GRCh38
rs534388758 Chr.6: 106186307-106186307 on Build GRCh38
rs747155799 Chr.6: 106186558-106186558 on Build GRCh38
rs573329878 Chr.6: 106186525-106186525 on Build GRCh38
rs780219305 Chr.6: 106186549-106186549 on Build GRCh38
rs763881439 Chr.6: 106186680-106186680 on Build GRCh38
rs763827614 Chr.6: 106186529-106186529 on Build GRCh38
rs751653699 Chr.6: 106186695-106186695 on Build GRCh38
rs1019267162 Chr.6: 106186300-106186300 on Build GRCh38
rs950494829 Chr.6: 106186446-106186446 on Build GRCh38
rs542567718 Chr.6: 106186280-106186280 on Build GRCh38
rs773459851 Chr.6: 106186622-106186622 on Build GRCh38
rs374442938 Chr.6: 106186711-106186711 on Build GRCh38
rs755063582 Chr.6: 106186701-106186701 on Build GRCh38
rs755978408 Chr.6: 106186526-106186526 on Build GRCh38

View Full Product Details