Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143502382-143502862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUCA2
Entrez Gene ID
2519
Gene Name
fucosidase, alpha-L- 2, plasma
Gene Aliases
dJ20N2.5
UniGene
Hs.17680
Gene Chromosome Location
Chr.6: 143494812-143511883 on Build GRCh38
Transcripts
NM_032020.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667756 c.828C>A p.T276T Chr.6: 143502490-143502490 on Build GRCh38
7410891 c.899T>C p.L300P Chr.6: 143502419-143502419 on Build GRCh38
5803671 c.847C>A p.P283T Chr.6: 143502471-143502471 on Build GRCh38
5793304 c.772G>T p.V258F Chr.6: 143502546-143502546 on Build GRCh38
594791 c.839G>A p.R280H Chr.6: 143502479-143502479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770509613 Chr.6: 143502404-143502405 on Build GRCh38
rs867114560 Chr.6: 143502456-143502456 on Build GRCh38
rs766600314 Chr.6: 143502549-143502549 on Build GRCh38
rs773786894 Chr.6: 143502487-143502487 on Build GRCh38
rs543674576 Chr.6: 143502479-143502479 on Build GRCh38
rs998668080 Chr.6: 143502719-143502719 on Build GRCh38
rs3789790 Chr.6: 143502774-143502774 on Build GRCh38
rs3789791 Chr.6: 143502830-143502830 on Build GRCh38
rs368459684 Chr.6: 143502579-143502579 on Build GRCh38
rs386706559 Chr.6: 143502621-143502622 on Build GRCh38
rs762353818 Chr.6: 143502539-143502539 on Build GRCh38
rs192022651 Chr.6: 143502689-143502689 on Build GRCh38
rs917136204 Chr.6: 143502759-143502759 on Build GRCh38
rs920412875 Chr.6: 143502642-143502642 on Build GRCh38
rs762302563 Chr.6: 143502412-143502412 on Build GRCh38
rs754920115 Chr.6: 143502471-143502471 on Build GRCh38
rs781151176 Chr.6: 143502472-143502472 on Build GRCh38
rs899926684 Chr.6: 143502710-143502710 on Build GRCh38
rs937194978 Chr.6: 143502647-143502647 on Build GRCh38
rs778661901 Chr.6: 143502601-143502601 on Build GRCh38
rs552475776 Chr.6: 143502695-143502695 on Build GRCh38
rs773761687 Chr.6: 143502626-143502626 on Build GRCh38
rs200430749 Chr.6: 143502619-143502620 on Build GRCh38
rs748940896 Chr.6: 143502485-143502485 on Build GRCh38
rs3789788 Chr.6: 143502621-143502621 on Build GRCh38
rs765636975 Chr.6: 143502558-143502558 on Build GRCh38
rs751628619 Chr.6: 143502550-143502550 on Build GRCh38
rs374985227 Chr.6: 143502421-143502421 on Build GRCh38
rs758628368 Chr.6: 143502434-143502434 on Build GRCh38
rs773708500 Chr.6: 143502542-143502542 on Build GRCh38
rs186717192 Chr.6: 143502674-143502674 on Build GRCh38
rs745355753 Chr.6: 143502526-143502526 on Build GRCh38
rs760005569 Chr.6: 143502532-143502532 on Build GRCh38
rs773840697 Chr.6: 143502404-143502404 on Build GRCh38
rs753611764 Chr.6: 143502589-143502589 on Build GRCh38
rs763323872 Chr.6: 143502545-143502545 on Build GRCh38
rs117472393 Chr.6: 143502680-143502680 on Build GRCh38
rs79184916 Chr.6: 143502607-143502607 on Build GRCh38
rs375228394 Chr.6: 143502553-143502553 on Build GRCh38
rs370706578 Chr.6: 143502449-143502449 on Build GRCh38
rs771445547 Chr.6: 143502533-143502533 on Build GRCh38
rs374520739 Chr.6: 143502409-143502409 on Build GRCh38
rs761147553 Chr.6: 143502407-143502407 on Build GRCh38
rs544662413 Chr.6: 143502780-143502780 on Build GRCh38
rs767444690 Chr.6: 143502557-143502557 on Build GRCh38
rs879439002 Chr.6: 143502621-143502622 on Build GRCh38
rs3789789 Chr.6: 143502622-143502622 on Build GRCh38
rs368673865 Chr.6: 143502428-143502428 on Build GRCh38
rs766633320 Chr.6: 143502451-143502451 on Build GRCh38
rs1042086929 Chr.6: 143502785-143502785 on Build GRCh38
rs182596147 Chr.6: 143502630-143502630 on Build GRCh38
rs777484258 Chr.6: 143502484-143502484 on Build GRCh38
rs139695584 Chr.6: 143502470-143502470 on Build GRCh38
rs770412068 Chr.6: 143502623-143502623 on Build GRCh38
rs1000303499 Chr.6: 143502424-143502424 on Build GRCh38
rs200310924 Chr.6: 143502581-143502581 on Build GRCh38
rs376770369 Chr.6: 143502812-143502812 on Build GRCh38
rs776903052 Chr.6: 143502410-143502410 on Build GRCh38
rs756946564 Chr.6: 143502591-143502591 on Build GRCh38
rs770287715 Chr.6: 143502541-143502541 on Build GRCh38
rs747993291 Chr.6: 143502475-143502475 on Build GRCh38

View Full Product Details