Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30791199-30791709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188B
Entrez Gene ID
84182
Gene Name
family with sequence similarity 188 member B
Gene Aliases
C7orf67
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30771417-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Gene Symbol
INMT-FAM188B
Entrez Gene ID
100526825
Gene Name
INMT-FAM188B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30752135-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963906 c.742G>T p.V248F Chr.7: 30791243-30791243 on Build GRCh38
6659135 c.1045C>T p.R349W Chr.7: 30791546-30791546 on Build GRCh38
1734198 c.1046G>A p.R349Q Chr.7: 30791547-30791547 on Build GRCh38
4004450 c.971G>A p.R324K Chr.7: 30791472-30791472 on Build GRCh38
4004449 c.965C>T p.T322M Chr.7: 30791466-30791466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767264500 Chr.7: 30791337-30791337 on Build GRCh38
rs151089199 Chr.7: 30791564-30791564 on Build GRCh38
rs778689188 Chr.7: 30791542-30791542 on Build GRCh38
rs757734675 Chr.7: 30791450-30791450 on Build GRCh38
rs749022893 Chr.7: 30791405-30791405 on Build GRCh38
rs755140213 Chr.7: 30791539-30791539 on Build GRCh38
rs776232171 Chr.7: 30791554-30791554 on Build GRCh38
rs779760592 Chr.7: 30791298-30791298 on Build GRCh38
rs377214150 Chr.7: 30791544-30791544 on Build GRCh38
rs758991160 Chr.7: 30791368-30791368 on Build GRCh38
rs16875355 Chr.7: 30791528-30791528 on Build GRCh38
rs749051508 Chr.7: 30791464-30791465 on Build GRCh38
rs753048755 Chr.7: 30791366-30791366 on Build GRCh38
rs765287774 Chr.7: 30791233-30791233 on Build GRCh38
rs761212892 Chr.7: 30791327-30791327 on Build GRCh38
rs751081117 Chr.7: 30791319-30791319 on Build GRCh38
rs763461332 Chr.7: 30791503-30791503 on Build GRCh38
rs12701034 Chr.7: 30791466-30791466 on Build GRCh38
rs373295305 Chr.7: 30791419-30791419 on Build GRCh38
rs1033387793 Chr.7: 30791638-30791638 on Build GRCh38
rs368051174 Chr.7: 30791452-30791452 on Build GRCh38
rs764738856 Chr.7: 30791437-30791437 on Build GRCh38
rs563349387 Chr.7: 30791485-30791485 on Build GRCh38
rs752503699 Chr.7: 30791599-30791599 on Build GRCh38
rs774318644 Chr.7: 30791480-30791480 on Build GRCh38
rs1027425922 Chr.7: 30791600-30791600 on Build GRCh38
rs763950875 Chr.7: 30791310-30791310 on Build GRCh38
rs62446940 Chr.7: 30791647-30791647 on Build GRCh38
rs951706021 Chr.7: 30791433-30791433 on Build GRCh38
rs537168510 Chr.7: 30791676-30791676 on Build GRCh38
rs536189237 Chr.7: 30791563-30791563 on Build GRCh38
rs142944427 Chr.7: 30791390-30791390 on Build GRCh38
rs770545132 Chr.7: 30791548-30791548 on Build GRCh38
rs759236849 Chr.7: 30791339-30791339 on Build GRCh38
rs768009257 Chr.7: 30791516-30791516 on Build GRCh38
rs758831576 Chr.7: 30791436-30791436 on Build GRCh38
rs759294830 Chr.7: 30791562-30791562 on Build GRCh38
rs776501754 Chr.7: 30791224-30791224 on Build GRCh38
rs12672119 Chr.7: 30791565-30791565 on Build GRCh38
rs764575197 Chr.7: 30791512-30791512 on Build GRCh38
rs376245817 Chr.7: 30791341-30791341 on Build GRCh38
rs778007380 Chr.7: 30791264-30791264 on Build GRCh38
rs114125123 Chr.7: 30791654-30791654 on Build GRCh38
rs776135787 Chr.7: 30791677-30791677 on Build GRCh38
rs777621925 Chr.7: 30791611-30791611 on Build GRCh38
rs374575606 Chr.7: 30791382-30791382 on Build GRCh38
rs778413193 Chr.7: 30791375-30791375 on Build GRCh38
rs570097105 Chr.7: 30791673-30791673 on Build GRCh38
rs1053755722 Chr.7: 30791356-30791356 on Build GRCh38
rs756302522 Chr.7: 30791467-30791467 on Build GRCh38
rs372857757 Chr.7: 30791572-30791572 on Build GRCh38
rs367583446 Chr.7: 30791408-30791408 on Build GRCh38
rs766204953 Chr.7: 30791499-30791499 on Build GRCh38
rs779186239 Chr.7: 30791598-30791598 on Build GRCh38
rs369576794 Chr.7: 30791622-30791622 on Build GRCh38
rs17159453 Chr.7: 30791286-30791286 on Build GRCh38
rs776392657 Chr.7: 30791502-30791502 on Build GRCh38
rs759416100 Chr.7: 30791301-30791301 on Build GRCh38
rs201348400 Chr.7: 30791351-30791351 on Build GRCh38
rs368335229 Chr.7: 30791501-30791501 on Build GRCh38
rs746100538 Chr.7: 30791396-30791396 on Build GRCh38
rs780649449 Chr.7: 30791246-30791246 on Build GRCh38
rs1044328023 Chr.7: 30791592-30791592 on Build GRCh38
rs1009191340 Chr.7: 30791556-30791556 on Build GRCh38
rs762908365 Chr.7: 30791306-30791306 on Build GRCh38
rs35897481 Chr.7: 30791472-30791472 on Build GRCh38
rs369701153 Chr.7: 30791389-30791389 on Build GRCh38
rs1019147693 Chr.7: 30791411-30791411 on Build GRCh38
rs181084371 Chr.7: 30791338-30791338 on Build GRCh38
rs770017395 Chr.7: 30791401-30791401 on Build GRCh38
rs750524835 Chr.7: 30791517-30791517 on Build GRCh38
rs768406432 Chr.7: 30791479-30791479 on Build GRCh38
rs571250988 Chr.7: 30791574-30791574 on Build GRCh38
rs369880517 Chr.7: 30791245-30791245 on Build GRCh38
rs762499754 Chr.7: 30791238-30791238 on Build GRCh38
rs376449663 Chr.7: 30791251-30791251 on Build GRCh38
rs771981738 Chr.7: 30791488-30791488 on Build GRCh38
rs1026637517 Chr.7: 30791653-30791653 on Build GRCh38
rs781526610 Chr.7: 30791391-30791391 on Build GRCh38
rs757707897 Chr.7: 30791513-30791513 on Build GRCh38
rs758184504 Chr.7: 30791608-30791608 on Build GRCh38
rs541721487 Chr.7: 30791418-30791418 on Build GRCh38
rs749146324 Chr.7: 30791474-30791474 on Build GRCh38
rs746967511 Chr.7: 30791615-30791615 on Build GRCh38
rs139782481 Chr.7: 30791679-30791679 on Build GRCh38
rs772671203 Chr.7: 30791258-30791258 on Build GRCh38
rs772615564 Chr.7: 30791490-30791490 on Build GRCh38
rs370149704 Chr.7: 30791431-30791431 on Build GRCh38
rs199816608 Chr.7: 30791253-30791253 on Build GRCh38
rs202006369 Chr.7: 30791593-30791593 on Build GRCh38
rs774263848 Chr.7: 30791407-30791407 on Build GRCh38
rs370929595 Chr.7: 30791304-30791304 on Build GRCh38
rs560122053 Chr.7: 30791443-30791443 on Build GRCh38
rs574245366 Chr.7: 30791409-30791409 on Build GRCh38
rs780292877 Chr.7: 30791525-30791525 on Build GRCh38
rs61741901 Chr.7: 30791287-30791287 on Build GRCh38
rs1002659832 Chr.7: 30791438-30791438 on Build GRCh38
rs763615532 Chr.7: 30791240-30791240 on Build GRCh38
rs1056328377 Chr.7: 30791273-30791273 on Build GRCh38
rs374435049 Chr.7: 30791432-30791432 on Build GRCh38
rs61741898 Chr.7: 30791280-30791280 on Build GRCh38
rs371926512 Chr.7: 30791231-30791231 on Build GRCh38
rs370172815 Chr.7: 30791546-30791546 on Build GRCh38
rs202164342 Chr.7: 30791302-30791302 on Build GRCh38
rs751028070 Chr.7: 30791243-30791243 on Build GRCh38
rs61739692 Chr.7: 30791237-30791237 on Build GRCh38
rs1049903498 Chr.7: 30791305-30791305 on Build GRCh38
rs200938852 Chr.7: 30791547-30791547 on Build GRCh38
rs980919941 Chr.7: 30791629-30791630 on Build GRCh38
rs775267181 Chr.7: 30791402-30791402 on Build GRCh38
rs756178537 Chr.7: 30791521-30791521 on Build GRCh38
rs753870329 Chr.7: 30791580-30791580 on Build GRCh38

View Full Product Details