Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 21908119-21908667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA7L
Entrez Gene ID
55536
Gene Name
cell division cycle associated 7 like
Gene Aliases
JPO2, R1, RAM2
UniGene
Hs.744069
Gene Chromosome Location
Chr.7: 21900899-21945924 on Build GRCh38
Transcripts
NM_001127370.2 NM_001127371.2 NM_018719.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279752 c.538C>A p.L180I Chr.7: 21908273-21908273 on Build GRCh38
1088471 c.620G>T p.S207I Chr.7: 21908191-21908191 on Build GRCh38
4164264 c.604G>A p.D202N Chr.7: 21908207-21908207 on Build GRCh38
1088472 c.448A>G p.T150A Chr.7: 21908363-21908363 on Build GRCh38
1088473 c.428G>A p.R143Q Chr.7: 21908383-21908383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750462397 Chr.7: 21908342-21908342 on Build GRCh38
rs760707123 Chr.7: 21908275-21908275 on Build GRCh38
rs757277115 Chr.7: 21908252-21908252 on Build GRCh38
rs543881067 Chr.7: 21908530-21908530 on Build GRCh38
rs972761529 Chr.7: 21908404-21908404 on Build GRCh38
rs35281045 Chr.7: 21908250-21908250 on Build GRCh38
rs778015525 Chr.7: 21908548-21908558 on Build GRCh38
rs778026662 Chr.7: 21908156-21908156 on Build GRCh38
rs1029355987 Chr.7: 21908634-21908634 on Build GRCh38
rs774001726 Chr.7: 21908311-21908313 on Build GRCh38
rs766864649 Chr.7: 21908304-21908304 on Build GRCh38
rs142981617 Chr.7: 21908459-21908459 on Build GRCh38
rs949553854 Chr.7: 21908196-21908196 on Build GRCh38
rs80103615 Chr.7: 21908623-21908623 on Build GRCh38
rs780509440 Chr.7: 21908348-21908348 on Build GRCh38
rs779213060 Chr.7: 21908527-21908527 on Build GRCh38
rs753436839 Chr.7: 21908510-21908511 on Build GRCh38
rs766453729 Chr.7: 21908511-21908511 on Build GRCh38
rs983978413 Chr.7: 21908367-21908367 on Build GRCh38
rs951405512 Chr.7: 21908569-21908569 on Build GRCh38
rs770128778 Chr.7: 21908184-21908184 on Build GRCh38
rs778688425 Chr.7: 21908513-21908515 on Build GRCh38
rs776702624 Chr.7: 21908395-21908395 on Build GRCh38
rs767651522 Chr.7: 21908168-21908168 on Build GRCh38
rs1026546380 Chr.7: 21908604-21908628 on Build GRCh38
rs201363125 Chr.7: 21908227-21908227 on Build GRCh38
rs760730241 Chr.7: 21908312-21908312 on Build GRCh38
rs141140287 Chr.7: 21908383-21908383 on Build GRCh38
rs756139901 Chr.7: 21908434-21908439 on Build GRCh38
rs747763741 Chr.7: 21908382-21908382 on Build GRCh38
rs146976111 Chr.7: 21908229-21908229 on Build GRCh38
rs765748356 Chr.7: 21908519-21908519 on Build GRCh38
rs147641498 Chr.7: 21908310-21908310 on Build GRCh38
rs758712356 Chr.7: 21908236-21908236 on Build GRCh38
rs371638574 Chr.7: 21908387-21908387 on Build GRCh38
rs773872806 Chr.7: 21908303-21908303 on Build GRCh38
rs371377746 Chr.7: 21908207-21908207 on Build GRCh38
rs1028888016 Chr.7: 21908301-21908301 on Build GRCh38
rs759460317 Chr.7: 21908400-21908400 on Build GRCh38
rs765485528 Chr.7: 21908311-21908320 on Build GRCh38
rs746309699 Chr.7: 21908179-21908179 on Build GRCh38
rs780597605 Chr.7: 21908424-21908424 on Build GRCh38
rs776714118 Chr.7: 21908257-21908260 on Build GRCh38
rs201577034 Chr.7: 21908507-21908507 on Build GRCh38
rs758047531 Chr.7: 21908557-21908557 on Build GRCh38
rs764953629 Chr.7: 21908403-21908403 on Build GRCh38
rs745803542 Chr.7: 21908479-21908479 on Build GRCh38
rs752469383 Chr.7: 21908408-21908408 on Build GRCh38
rs975194611 Chr.7: 21908613-21908613 on Build GRCh38
rs1052204522 Chr.7: 21908145-21908145 on Build GRCh38
rs755459234 Chr.7: 21908516-21908516 on Build GRCh38
rs753650366 Chr.7: 21908232-21908232 on Build GRCh38
rs758727113 Chr.7: 21908538-21908539 on Build GRCh38
rs987310773 Chr.7: 21908390-21908390 on Build GRCh38
rs201160995 Chr.7: 21908502-21908502 on Build GRCh38
rs767757196 Chr.7: 21908415-21908415 on Build GRCh38
rs765943506 Chr.7: 21908510-21908514 on Build GRCh38
rs755859959 Chr.7: 21908139-21908139 on Build GRCh38
rs150333184 Chr.7: 21908444-21908444 on Build GRCh38
rs748459254 Chr.7: 21908533-21908536 on Build GRCh38
rs975330798 Chr.7: 21908427-21908427 on Build GRCh38
rs201734057 Chr.7: 21908299-21908299 on Build GRCh38
rs749722138 Chr.7: 21908354-21908354 on Build GRCh38
rs148205645 Chr.7: 21908346-21908346 on Build GRCh38
rs759370631 Chr.7: 21908321-21908321 on Build GRCh38
rs758630757 Chr.7: 21908155-21908155 on Build GRCh38
rs904686660 Chr.7: 21908247-21908247 on Build GRCh38
rs558132059 Chr.7: 21908451-21908451 on Build GRCh38
rs761733292 Chr.7: 21908398-21908400 on Build GRCh38
rs761198537 Chr.7: 21908209-21908209 on Build GRCh38
rs200891942 Chr.7: 21908392-21908392 on Build GRCh38
rs771498809 Chr.7: 21908308-21908308 on Build GRCh38
rs376335429 Chr.7: 21908425-21908425 on Build GRCh38
rs376745541 Chr.7: 21908178-21908178 on Build GRCh38
rs1005631468 Chr.7: 21908588-21908588 on Build GRCh38
rs746675295 Chr.7: 21908544-21908544 on Build GRCh38
rs137919998 Chr.7: 21908447-21908447 on Build GRCh38
rs200919907 Chr.7: 21908203-21908203 on Build GRCh38
rs747150378 Chr.7: 21908251-21908251 on Build GRCh38
rs1053429818 Chr.7: 21908545-21908545 on Build GRCh38
rs771515346 Chr.7: 21908169-21908169 on Build GRCh38
rs753026977 Chr.7: 21908538-21908538 on Build GRCh38
rs759717318 Chr.7: 21908256-21908257 on Build GRCh38
rs963859297 Chr.7: 21908606-21908606 on Build GRCh38
rs755449955 Chr.7: 21908432-21908432 on Build GRCh38
rs770218756 Chr.7: 21908273-21908273 on Build GRCh38
rs750316250 Chr.7: 21908434-21908436 on Build GRCh38
rs554719860 Chr.7: 21908505-21908505 on Build GRCh38
rs369568135 Chr.7: 21908453-21908453 on Build GRCh38
rs767220245 Chr.7: 21908398-21908412 on Build GRCh38
rs753872064 Chr.7: 21908513-21908513 on Build GRCh38
rs746374294 Chr.7: 21908267-21908267 on Build GRCh38
rs779282166 Chr.7: 21908369-21908369 on Build GRCh38
rs775772395 Chr.7: 21908200-21908200 on Build GRCh38
rs781136077 Chr.7: 21908339-21908339 on Build GRCh38
rs1031964759 Chr.7: 21908270-21908270 on Build GRCh38
rs373779507 Chr.7: 21908322-21908322 on Build GRCh38
rs767559031 Chr.7: 21908223-21908223 on Build GRCh38
rs374550676 Chr.7: 21908300-21908300 on Build GRCh38
rs746526455 Chr.7: 21908639-21908639 on Build GRCh38
rs909753364 Chr.7: 21908371-21908371 on Build GRCh38
rs1026030280 Chr.7: 21908565-21908565 on Build GRCh38
rs747008088 Chr.7: 21908157-21908157 on Build GRCh38
rs1046584240 Chr.7: 21908225-21908225 on Build GRCh38
rs367949098 Chr.7: 21908362-21908362 on Build GRCh38
rs781750531 Chr.7: 21908260-21908260 on Build GRCh38
rs528806227 Chr.7: 21908280-21908280 on Build GRCh38
rs750543368 Chr.7: 21908422-21908422 on Build GRCh38
rs563675406 Chr.7: 21908201-21908201 on Build GRCh38
rs776221731 Chr.7: 21908463-21908463 on Build GRCh38
rs778114786 Chr.7: 21908249-21908249 on Build GRCh38
rs370049835 Chr.7: 21908504-21908504 on Build GRCh38
rs142227347 Chr.7: 21908313-21908313 on Build GRCh38
rs768997741 Chr.7: 21908363-21908363 on Build GRCh38
rs761972880 Chr.7: 21908410-21908410 on Build GRCh38
rs746581280 Chr.7: 21908454-21908454 on Build GRCh38
rs549044913 Chr.7: 21908513-21908514 on Build GRCh38
rs530173538 Chr.7: 21908142-21908142 on Build GRCh38
rs560042862 Chr.7: 21908230-21908230 on Build GRCh38
rs143179934 Chr.7: 21908335-21908335 on Build GRCh38

View Full Product Details