Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 51029482-51030061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
UniGene
Hs.99141
Gene Chromosome Location
Chr.7: 51016212-51316818 on Build GRCh38
Transcripts
NM_001287436.1 NM_015198.3 XM_005271750.1 XM_005271751.1 XM_005271756.1 XM_011515234.1 XM_011515235.1 XM_011515236.1 XM_011515237.1 XM_011515238.1 XM_011515239.1 XM_011515240.1 XM_011515241.1 XM_017011898.1 XM_017011899.1 XM_017011900.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4782881 c.1808C>T p.A603V Chr.7: 51029534-51029534 on Build GRCh38
4635515 c.1581G>T p.Q527H Chr.7: 51029515-51029515 on Build GRCh38
5049108 c.1575C>G p.C525W Chr.7: 51029521-51029521 on Build GRCh38
3639585 c.1800C>T p.I600I Chr.7: 51029542-51029542 on Build GRCh38
1451386 c.1562C>T p.A521V Chr.7: 51029534-51029534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954603455 Chr.7: 51029846-51029846 on Build GRCh38
rs993549550 Chr.7: 51029795-51029795 on Build GRCh38
rs113851285 Chr.7: 51029914-51029914 on Build GRCh38
rs904033113 Chr.7: 51029745-51029745 on Build GRCh38
rs772733046 Chr.7: 51029923-51029923 on Build GRCh38
rs940591589 Chr.7: 51029658-51029658 on Build GRCh38
rs867297566 Chr.7: 51029982-51029982 on Build GRCh38
rs770351240 Chr.7: 51029546-51029546 on Build GRCh38
rs537020830 Chr.7: 51029927-51029927 on Build GRCh38
rs986305017 Chr.7: 51029868-51029868 on Build GRCh38
rs560953709 Chr.7: 51029596-51029596 on Build GRCh38
rs1046828179 Chr.7: 51029957-51029957 on Build GRCh38
rs368010769 Chr.7: 51029541-51029541 on Build GRCh38
rs772404228 Chr.7: 51029524-51029524 on Build GRCh38
rs142839981 Chr.7: 51029538-51029538 on Build GRCh38
rs945420831 Chr.7: 51029950-51029950 on Build GRCh38
rs61739178 Chr.7: 51029558-51029558 on Build GRCh38
rs747070996 Chr.7: 51029537-51029537 on Build GRCh38
rs1019880396 Chr.7: 51029856-51029856 on Build GRCh38
rs763523295 Chr.7: 51029587-51029587 on Build GRCh38
rs567175877 Chr.7: 51029557-51029557 on Build GRCh38
rs557126384 Chr.7: 51030024-51030024 on Build GRCh38
rs146068844 Chr.7: 51029688-51029688 on Build GRCh38
rs183984683 Chr.7: 51029706-51029706 on Build GRCh38
rs549892880 Chr.7: 51029643-51029643 on Build GRCh38
rs181046742 Chr.7: 51029835-51029835 on Build GRCh38
rs986294121 Chr.7: 51029588-51029588 on Build GRCh38
rs1041905904 Chr.7: 51029828-51029828 on Build GRCh38
rs1034139292 Chr.7: 51029739-51029739 on Build GRCh38
rs981350558 Chr.7: 51029678-51029678 on Build GRCh38
rs750197318 Chr.7: 51029633-51029633 on Build GRCh38
rs906593035 Chr.7: 51029717-51029717 on Build GRCh38
rs753126307 Chr.7: 51029953-51029953 on Build GRCh38
rs894398876 Chr.7: 51029743-51029743 on Build GRCh38
rs749182983 Chr.7: 51029503-51029503 on Build GRCh38
rs910839032 Chr.7: 51029595-51029595 on Build GRCh38
rs375259494 Chr.7: 51029622-51029622 on Build GRCh38
rs368473607 Chr.7: 51029547-51029547 on Build GRCh38
rs192749584 Chr.7: 51029637-51029637 on Build GRCh38
rs200382652 Chr.7: 51029540-51029540 on Build GRCh38
rs569801763 Chr.7: 51029645-51029645 on Build GRCh38
rs1055570599 Chr.7: 51030009-51030009 on Build GRCh38
rs781504529 Chr.7: 51029924-51029924 on Build GRCh38
rs779046375 Chr.7: 51029631-51029631 on Build GRCh38
rs566095868 Chr.7: 51029792-51029792 on Build GRCh38
rs760161537 Chr.7: 51029564-51029564 on Build GRCh38
rs945468464 Chr.7: 51029705-51029705 on Build GRCh38
rs778835962 Chr.7: 51029514-51029514 on Build GRCh38
rs892895533 Chr.7: 51029748-51029748 on Build GRCh38
rs757057355 Chr.7: 51029504-51029504 on Build GRCh38
rs1008473716 Chr.7: 51029844-51029844 on Build GRCh38
rs372180151 Chr.7: 51029562-51029562 on Build GRCh38
rs188327267 Chr.7: 51029812-51029812 on Build GRCh38
rs1002749883 Chr.7: 51029729-51029729 on Build GRCh38
rs1006070015 Chr.7: 51029758-51029758 on Build GRCh38
rs910686662 Chr.7: 51029907-51029907 on Build GRCh38
rs370469180 Chr.7: 51029528-51029528 on Build GRCh38
rs17656599 Chr.7: 51029519-51029519 on Build GRCh38
rs961765280 Chr.7: 51029791-51029791 on Build GRCh38
rs767107999 Chr.7: 51029561-51029561 on Build GRCh38
rs755107149 Chr.7: 51029615-51029615 on Build GRCh38
rs753442654 Chr.7: 51029599-51029599 on Build GRCh38
rs778190807 Chr.7: 51029973-51029973 on Build GRCh38
rs748041914 Chr.7: 51029779-51029779 on Build GRCh38
rs756595454 Chr.7: 51029961-51029961 on Build GRCh38

View Full Product Details