Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117719368-117719916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTTNBP2
Entrez Gene ID
83992
Gene Name
cortactin binding protein 2
Gene Aliases
C7orf8, CORTBP2, Orf4
UniGene
Hs.592285
Gene Chromosome Location
Chr.7: 117710648-117873507 on Build GRCh38
Transcripts
NM_033427.2 XM_005250634.4 XM_005250635.3 XM_011516615.2 XM_017012706.1 XM_017012708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6255685 c.4600C>A p.L1534I Chr.7: 117719548-117719548 on Build GRCh38
319741 c.4567G>T p.G1523C Chr.7: 117719581-117719581 on Build GRCh38
5406304 c.4617C>T p.S1539S Chr.7: 117719531-117719531 on Build GRCh38
7117411 c.4556G>C p.R1519T Chr.7: 117719592-117719592 on Build GRCh38
6108441 c.4611G>T p.M1537I Chr.7: 117719537-117719537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764465645 Chr.7: 117719417-117719417 on Build GRCh38
rs765070933 Chr.7: 117719458-117719458 on Build GRCh38
rs144520488 Chr.7: 117719635-117719635 on Build GRCh38
rs756392237 Chr.7: 117719614-117719614 on Build GRCh38
rs769892311 Chr.7: 117719786-117719786 on Build GRCh38
rs369909482 Chr.7: 117719492-117719492 on Build GRCh38
rs779821431 Chr.7: 117719448-117719448 on Build GRCh38
rs200405061 Chr.7: 117719709-117719709 on Build GRCh38
rs774415522 Chr.7: 117719467-117719468 on Build GRCh38
rs112489206 Chr.7: 117719870-117719870 on Build GRCh38
rs775949032 Chr.7: 117719630-117719630 on Build GRCh38
rs561308163 Chr.7: 117719393-117719393 on Build GRCh38
rs74635469 Chr.7: 117719736-117719736 on Build GRCh38
rs759092792 Chr.7: 117719647-117719647 on Build GRCh38
rs770665037 Chr.7: 117719655-117719655 on Build GRCh38
rs754717140 Chr.7: 117719842-117719845 on Build GRCh38
rs752538500 Chr.7: 117719585-117719585 on Build GRCh38
rs1013370805 Chr.7: 117719493-117719493 on Build GRCh38
rs894584369 Chr.7: 117719817-117719817 on Build GRCh38
rs746644129 Chr.7: 117719599-117719599 on Build GRCh38
rs979046903 Chr.7: 117719489-117719489 on Build GRCh38
rs747688830 Chr.7: 117719545-117719545 on Build GRCh38
rs376142744 Chr.7: 117719557-117719557 on Build GRCh38
rs869210388 Chr.7: 117719846-117719846 on Build GRCh38
rs1051893112 Chr.7: 117719666-117719666 on Build GRCh38
rs771179521 Chr.7: 117719610-117719610 on Build GRCh38
rs770702632 Chr.7: 117719563-117719563 on Build GRCh38
rs572755683 Chr.7: 117719647-117719647 on Build GRCh38
rs777218215 Chr.7: 117719760-117719760 on Build GRCh38
rs747884214 Chr.7: 117719483-117719483 on Build GRCh38
rs772821399 Chr.7: 117719583-117719584 on Build GRCh38
rs1002925671 Chr.7: 117719456-117719456 on Build GRCh38
rs760285636 Chr.7: 117719614-117719615 on Build GRCh38
rs546876647 Chr.7: 117719424-117719424 on Build GRCh38
rs555247736 Chr.7: 117719503-117719503 on Build GRCh38
rs770393389 Chr.7: 117719532-117719532 on Build GRCh38
rs535673418 Chr.7: 117719426-117719426 on Build GRCh38
rs375775074 Chr.7: 117719757-117719757 on Build GRCh38
rs1017561244 Chr.7: 117719759-117719759 on Build GRCh38
rs764018285 Chr.7: 117719685-117719685 on Build GRCh38
rs571557190 Chr.7: 117719569-117719569 on Build GRCh38
rs557495204 Chr.7: 117719895-117719895 on Build GRCh38
rs977949373 Chr.7: 117719749-117719749 on Build GRCh38
rs1037882666 Chr.7: 117719410-117719411 on Build GRCh38
rs558175040 Chr.7: 117719705-117719705 on Build GRCh38
rs897950547 Chr.7: 117719418-117719418 on Build GRCh38
rs577893870 Chr.7: 117719821-117719821 on Build GRCh38
rs188569775 Chr.7: 117719658-117719658 on Build GRCh38
rs775511227 Chr.7: 117719668-117719668 on Build GRCh38
rs142195236 Chr.7: 117719570-117719570 on Build GRCh38
rs545326991 Chr.7: 117719823-117719823 on Build GRCh38
rs777492122 Chr.7: 117719643-117719643 on Build GRCh38
rs1038128706 Chr.7: 117719833-117719833 on Build GRCh38
rs968614109 Chr.7: 117719478-117719478 on Build GRCh38
rs34712344 Chr.7: 117719805-117719806 on Build GRCh38
rs867143743 Chr.7: 117719735-117719735 on Build GRCh38
rs963992792 Chr.7: 117719552-117719552 on Build GRCh38
rs771904829 Chr.7: 117719580-117719592 on Build GRCh38
rs749751383 Chr.7: 117719636-117719636 on Build GRCh38
rs916985638 Chr.7: 117719565-117719565 on Build GRCh38
rs749152010 Chr.7: 117719501-117719501 on Build GRCh38
rs199539621 Chr.7: 117719659-117719659 on Build GRCh38
rs534510422 Chr.7: 117719696-117719696 on Build GRCh38
rs151225619 Chr.7: 117719588-117719588 on Build GRCh38
rs762811438 Chr.7: 117719678-117719678 on Build GRCh38
rs1057108172 Chr.7: 117719487-117719487 on Build GRCh38
rs149265601 Chr.7: 117719609-117719609 on Build GRCh38
rs770713034 Chr.7: 117719657-117719657 on Build GRCh38
rs867374363 Chr.7: 117719438-117719438 on Build GRCh38
rs939147250 Chr.7: 117719781-117719781 on Build GRCh38
rs568993434 Chr.7: 117719592-117719592 on Build GRCh38
rs375406711 Chr.7: 117719470-117719470 on Build GRCh38
rs764103771 Chr.7: 117719589-117719589 on Build GRCh38
rs1031994496 Chr.7: 117719888-117719888 on Build GRCh38
rs569839918 Chr.7: 117719407-117719407 on Build GRCh38
rs768858221 Chr.7: 117719707-117719707 on Build GRCh38
rs148511220 Chr.7: 117719530-117719530 on Build GRCh38
rs752869900 Chr.7: 117719469-117719469 on Build GRCh38
rs757633013 Chr.7: 117719502-117719502 on Build GRCh38
rs780406244 Chr.7: 117719538-117719538 on Build GRCh38
rs781557273 Chr.7: 117719510-117719510 on Build GRCh38
rs773009121 Chr.7: 117719548-117719548 on Build GRCh38
rs777426780 Chr.7: 117719435-117719435 on Build GRCh38
rs765967816 Chr.7: 117719741-117719742 on Build GRCh38
rs771708340 Chr.7: 117719641-117719641 on Build GRCh38
rs765261471 Chr.7: 117719576-117719576 on Build GRCh38
rs550774805 Chr.7: 117719591-117719591 on Build GRCh38
rs769428096 Chr.7: 117719664-117719664 on Build GRCh38

View Full Product Details