Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121124188-121124675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPED1
Entrez Gene ID
79974
Gene Name
cadherin like and PC-esterase domain containing 1
Gene Aliases
C7orf58
UniGene
Hs.189652
Gene Chromosome Location
Chr.7: 120988697-121297444 on Build GRCh38
Transcripts
NM_001105533.1 NM_024913.4 XM_017012649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3632412 c.947G>A p.R316K Chr.7: 121124359-121124359 on Build GRCh38
5654104 c.1011G>C p.A337A Chr.7: 121124423-121124423 on Build GRCh38
3669680 c.981G>T p.M327I Chr.7: 121124393-121124393 on Build GRCh38
106908 c.942C>T p.F314F Chr.7: 121124354-121124354 on Build GRCh38
7285638 c.950C>T p.A317V Chr.7: 121124362-121124362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1524498 Chr.7: 121124423-121124423 on Build GRCh38
rs1037105793 Chr.7: 121124604-121124604 on Build GRCh38
rs150453350 Chr.7: 121124438-121124438 on Build GRCh38
rs760046352 Chr.7: 121124294-121124294 on Build GRCh38
rs1024782408 Chr.7: 121124594-121124594 on Build GRCh38
rs776152875 Chr.7: 121124489-121124498 on Build GRCh38
rs372022088 Chr.7: 121124606-121124606 on Build GRCh38
rs995413163 Chr.7: 121124235-121124235 on Build GRCh38
rs798948 Chr.7: 121124519-121124519 on Build GRCh38
rs774679211 Chr.7: 121124393-121124393 on Build GRCh38
rs763846150 Chr.7: 121124506-121124506 on Build GRCh38
rs757211766 Chr.7: 121124515-121124515 on Build GRCh38
rs764125990 Chr.7: 121124425-121124425 on Build GRCh38
rs17143165 Chr.7: 121124389-121124389 on Build GRCh38
rs989578159 Chr.7: 121124530-121124530 on Build GRCh38
rs768853490 Chr.7: 121124448-121124448 on Build GRCh38
rs574151683 Chr.7: 121124482-121124482 on Build GRCh38
rs1034437184 Chr.7: 121124291-121124291 on Build GRCh38
rs1019626547 Chr.7: 121124251-121124251 on Build GRCh38
rs143044610 Chr.7: 121124401-121124401 on Build GRCh38
rs969593123 Chr.7: 121124542-121124542 on Build GRCh38
rs548687153 Chr.7: 121124231-121124231 on Build GRCh38
rs944420640 Chr.7: 121124436-121124436 on Build GRCh38
rs764591660 Chr.7: 121124500-121124531 on Build GRCh38
rs906891113 Chr.7: 121124372-121124372 on Build GRCh38
rs148370748 Chr.7: 121124422-121124422 on Build GRCh38
rs187097612 Chr.7: 121124424-121124424 on Build GRCh38
rs16870557 Chr.7: 121124593-121124593 on Build GRCh38
rs368546611 Chr.7: 121124290-121124290 on Build GRCh38
rs1044026135 Chr.7: 121124366-121124366 on Build GRCh38
rs897834795 Chr.7: 121124598-121124598 on Build GRCh38
rs913579908 Chr.7: 121124473-121124473 on Build GRCh38
rs370215040 Chr.7: 121124444-121124444 on Build GRCh38
rs752480478 Chr.7: 121124325-121124325 on Build GRCh38
rs561105658 Chr.7: 121124239-121124239 on Build GRCh38
rs747113701 Chr.7: 121124359-121124359 on Build GRCh38
rs989615656 Chr.7: 121124320-121124320 on Build GRCh38
rs764871220 Chr.7: 121124490-121124490 on Build GRCh38
rs554039793 Chr.7: 121124495-121124495 on Build GRCh38
rs763384936 Chr.7: 121124495-121124499 on Build GRCh38
rs1001951994 Chr.7: 121124276-121124276 on Build GRCh38
rs894590216 Chr.7: 121124565-121124565 on Build GRCh38
rs200112614 Chr.7: 121124306-121124306 on Build GRCh38
rs368245288 Chr.7: 121124404-121124404 on Build GRCh38
rs547003345 Chr.7: 121124475-121124476 on Build GRCh38
rs546770429 Chr.7: 121124267-121124267 on Build GRCh38
rs762832898 Chr.7: 121124500-121124500 on Build GRCh38
rs140560169 Chr.7: 121124354-121124354 on Build GRCh38
rs774607731 Chr.7: 121124506-121124507 on Build GRCh38
rs770109987 Chr.7: 121124376-121124376 on Build GRCh38
rs368701131 Chr.7: 121124468-121124468 on Build GRCh38
rs756463039 Chr.7: 121124432-121124432 on Build GRCh38
rs1044005002 Chr.7: 121124256-121124256 on Build GRCh38
rs751343033 Chr.7: 121124514-121124514 on Build GRCh38
rs372184352 Chr.7: 121124426-121124426 on Build GRCh38
rs762046137 Chr.7: 121124516-121124519 on Build GRCh38
rs201986877 Chr.7: 121124307-121124307 on Build GRCh38
rs769221141 Chr.7: 121124391-121124391 on Build GRCh38
rs928279994 Chr.7: 121124566-121124566 on Build GRCh38
rs147947186 Chr.7: 121124375-121124375 on Build GRCh38
rs189964151 Chr.7: 121124536-121124536 on Build GRCh38
rs770416298 Chr.7: 121124495-121124495 on Build GRCh38
rs777511519 Chr.7: 121124336-121124336 on Build GRCh38
rs745371610 Chr.7: 121124491-121124492 on Build GRCh38
rs892400882 Chr.7: 121124242-121124242 on Build GRCh38
rs749676033 Chr.7: 121124445-121124445 on Build GRCh38
rs948320383 Chr.7: 121124360-121124360 on Build GRCh38
rs774754179 Chr.7: 121124454-121124454 on Build GRCh38
rs770879282 Chr.7: 121124361-121124361 on Build GRCh38
rs144932954 Chr.7: 121124247-121124247 on Build GRCh38
rs374353575 Chr.7: 121124320-121124320 on Build GRCh38
rs534136212 Chr.7: 121124494-121124494 on Build GRCh38
rs745940245 Chr.7: 121124429-121124429 on Build GRCh38
rs749986213 Chr.7: 121124373-121124373 on Build GRCh38
rs200186503 Chr.7: 121124461-121124461 on Build GRCh38
rs776195156 Chr.7: 121124494-121124495 on Build GRCh38
rs758414206 Chr.7: 121124335-121124335 on Build GRCh38
rs112116837 Chr.7: 121124534-121124534 on Build GRCh38
rs748643141 Chr.7: 121124460-121124460 on Build GRCh38
rs776172827 Chr.7: 121124478-121124478 on Build GRCh38
rs776064081 Chr.7: 121124378-121124378 on Build GRCh38
rs781464866 Chr.7: 121124427-121124427 on Build GRCh38
rs778776131 Chr.7: 121124364-121124364 on Build GRCh38

View Full Product Details