Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27748336-27748831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4542762 c.324G>C p.Q108H Chr.8: 27748519-27748519 on Build GRCh38
6622020 c.278C>T p.T93M Chr.8: 27748565-27748565 on Build GRCh38
7184253 c.334C>T p.H112Y Chr.8: 27748509-27748509 on Build GRCh38
3899318 c.328G>T p.G110C Chr.8: 27748515-27748515 on Build GRCh38
6368812 c.308A>T p.D103V Chr.8: 27748535-27748535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039324631 Chr.8: 27748777-27748777 on Build GRCh38
rs747145585 Chr.8: 27748445-27748445 on Build GRCh38
rs563681976 Chr.8: 27748672-27748672 on Build GRCh38
rs772227286 Chr.8: 27748706-27748706 on Build GRCh38
rs781515866 Chr.8: 27748588-27748588 on Build GRCh38
rs372998761 Chr.8: 27748463-27748463 on Build GRCh38
rs376182770 Chr.8: 27748484-27748484 on Build GRCh38
rs577630274 Chr.8: 27748653-27748653 on Build GRCh38
rs749035925 Chr.8: 27748483-27748483 on Build GRCh38
rs766980234 Chr.8: 27748579-27748579 on Build GRCh38
rs996001561 Chr.8: 27748655-27748655 on Build GRCh38
rs1047228806 Chr.8: 27748759-27748759 on Build GRCh38
rs765484544 Chr.8: 27748558-27748558 on Build GRCh38
rs929493438 Chr.8: 27748714-27748714 on Build GRCh38
rs111836868 Chr.8: 27748658-27748658 on Build GRCh38
rs76860931 Chr.8: 27748416-27748416 on Build GRCh38
rs748422590 Chr.8: 27748644-27748644 on Build GRCh38
rs763676522 Chr.8: 27748537-27748537 on Build GRCh38
rs555691437 Chr.8: 27748393-27748393 on Build GRCh38
rs778559077 Chr.8: 27748629-27748629 on Build GRCh38
rs17477488 Chr.8: 27748638-27748638 on Build GRCh38
rs572117238 Chr.8: 27748427-27748427 on Build GRCh38
rs768473604 Chr.8: 27748562-27748562 on Build GRCh38
rs1031645307 Chr.8: 27748690-27748690 on Build GRCh38
rs888073291 Chr.8: 27748436-27748436 on Build GRCh38
rs150602989 Chr.8: 27748793-27748793 on Build GRCh38
rs186962048 Chr.8: 27748705-27748705 on Build GRCh38
rs760181505 Chr.8: 27748533-27748533 on Build GRCh38
rs542990072 Chr.8: 27748709-27748709 on Build GRCh38
rs751624701 Chr.8: 27748582-27748582 on Build GRCh38
rs909645673 Chr.8: 27748771-27748771 on Build GRCh38
rs948873386 Chr.8: 27748356-27748356 on Build GRCh38
rs1022399787 Chr.8: 27748615-27748615 on Build GRCh38
rs919385764 Chr.8: 27748710-27748710 on Build GRCh38
rs370914554 Chr.8: 27748505-27748505 on Build GRCh38
rs1018115736 Chr.8: 27748447-27748447 on Build GRCh38
rs773596531 Chr.8: 27748780-27748780 on Build GRCh38
rs867818444 Chr.8: 27748438-27748438 on Build GRCh38
rs980707227 Chr.8: 27748379-27748379 on Build GRCh38
rs200083256 Chr.8: 27748564-27748564 on Build GRCh38
rs752085662 Chr.8: 27748645-27748656 on Build GRCh38
rs865838174 Chr.8: 27748539-27748539 on Build GRCh38
rs541493473 Chr.8: 27748608-27748608 on Build GRCh38
rs144207300 Chr.8: 27748679-27748679 on Build GRCh38
rs776286032 Chr.8: 27748541-27748541 on Build GRCh38
rs781570900 Chr.8: 27748466-27748466 on Build GRCh38
rs762121706 Chr.8: 27748542-27748542 on Build GRCh38
rs376553863 Chr.8: 27748583-27748583 on Build GRCh38
rs908148833 Chr.8: 27748806-27748806 on Build GRCh38
rs772104417 Chr.8: 27748513-27748513 on Build GRCh38
rs372802643 Chr.8: 27748494-27748494 on Build GRCh38
rs956061886 Chr.8: 27748695-27748695 on Build GRCh38
rs112765590 Chr.8: 27748776-27748776 on Build GRCh38
rs998478227 Chr.8: 27748792-27748792 on Build GRCh38
rs752999918 Chr.8: 27748602-27748602 on Build GRCh38
rs559598233 Chr.8: 27748766-27748766 on Build GRCh38
rs774856802 Chr.8: 27748527-27748527 on Build GRCh38
rs960947386 Chr.8: 27748684-27748684 on Build GRCh38
rs970151336 Chr.8: 27748713-27748713 on Build GRCh38
rs774305168 Chr.8: 27748485-27748485 on Build GRCh38
rs998135749 Chr.8: 27748497-27748497 on Build GRCh38

View Full Product Details