Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103420877-103421354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF13
Entrez Gene ID
25879
Gene Name
DDB1 and CUL4 associated factor 13
Gene Aliases
GM83, HSPC064, WDSOF1
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103414714-103443453 on Build GRCh38
Transcripts
NM_015420.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3727687 c.764G>A p.R255H Chr.8: 103421012-103421012 on Build GRCh38
6438587 c.784G>A p.G262S Chr.8: 103421032-103421032 on Build GRCh38
5565198 c.752G>A p.R251Q Chr.8: 103421000-103421000 on Build GRCh38
5873649 c.751C>T p.R251W Chr.8: 103420999-103420999 on Build GRCh38
6640441 c.763C>T p.R255C Chr.8: 103421011-103421011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761301834 Chr.8: 103421041-103421041 on Build GRCh38
rs764680089 Chr.8: 103421243-103421243 on Build GRCh38
rs991300389 Chr.8: 103420949-103420949 on Build GRCh38
rs959931104 Chr.8: 103421170-103421170 on Build GRCh38
rs767792186 Chr.8: 103421055-103421055 on Build GRCh38
rs373378051 Chr.8: 103421021-103421021 on Build GRCh38
rs907474666 Chr.8: 103420921-103420921 on Build GRCh38
rs770970108 Chr.8: 103420938-103420938 on Build GRCh38
rs536797299 Chr.8: 103421042-103421042 on Build GRCh38
rs187330014 Chr.8: 103421289-103421289 on Build GRCh38
rs147879440 Chr.8: 103421046-103421046 on Build GRCh38
rs774171051 Chr.8: 103421016-103421016 on Build GRCh38
rs906916813 Chr.8: 103421262-103421262 on Build GRCh38
rs967407867 Chr.8: 103420902-103420902 on Build GRCh38
rs763005420 Chr.8: 103420933-103420933 on Build GRCh38
rs778048907 Chr.8: 103421011-103421011 on Build GRCh38
rs745814983 Chr.8: 103421095-103421095 on Build GRCh38
rs199708023 Chr.8: 103421059-103421059 on Build GRCh38
rs920026512 Chr.8: 103420993-103420993 on Build GRCh38
rs760281498 Chr.8: 103420966-103420966 on Build GRCh38
rs578163618 Chr.8: 103421298-103421298 on Build GRCh38
rs377711032 Chr.8: 103421104-103421104 on Build GRCh38
rs144660563 Chr.8: 103421026-103421026 on Build GRCh38
rs761385547 Chr.8: 103421127-103421127 on Build GRCh38
rs753328868 Chr.8: 103420980-103420980 on Build GRCh38
rs540644850 Chr.8: 103421177-103421177 on Build GRCh38
rs778134995 Chr.8: 103421058-103421058 on Build GRCh38
rs879916405 Chr.8: 103421160-103421160 on Build GRCh38
rs914342983 Chr.8: 103421164-103421164 on Build GRCh38
rs759245083 Chr.8: 103420958-103420958 on Build GRCh38
rs376534034 Chr.8: 103421141-103421141 on Build GRCh38
rs539634733 Chr.8: 103420906-103420906 on Build GRCh38
rs776276050 Chr.8: 103421123-103421123 on Build GRCh38
rs772725147 Chr.8: 103421245-103421245 on Build GRCh38
rs760009356 Chr.8: 103421252-103421252 on Build GRCh38
rs1044043050 Chr.8: 103421220-103421220 on Build GRCh38
rs200624174 Chr.8: 103421308-103421308 on Build GRCh38
rs771058066 Chr.8: 103421015-103421015 on Build GRCh38
rs756718921 Chr.8: 103421130-103421130 on Build GRCh38
rs146305428 Chr.8: 103420971-103420971 on Build GRCh38
rs1002958303 Chr.8: 103420954-103420954 on Build GRCh38
rs780424597 Chr.8: 103420990-103420990 on Build GRCh38
rs548751419 Chr.8: 103420918-103420918 on Build GRCh38
rs372455118 Chr.8: 103420926-103420926 on Build GRCh38
rs753949875 Chr.8: 103421148-103421148 on Build GRCh38
rs777843588 Chr.8: 103421148-103421148 on Build GRCh38
rs1035377655 Chr.8: 103421158-103421158 on Build GRCh38
rs753177667 Chr.8: 103421284-103421284 on Build GRCh38
rs755468577 Chr.8: 103421054-103421054 on Build GRCh38
rs776192350 Chr.8: 103421034-103421034 on Build GRCh38
rs201279500 Chr.8: 103421326-103421326 on Build GRCh38
rs944317482 Chr.8: 103421022-103421022 on Build GRCh38
rs745753981 Chr.8: 103421020-103421020 on Build GRCh38
rs768037483 Chr.8: 103421274-103421274 on Build GRCh38
rs1021759155 Chr.8: 103421097-103421097 on Build GRCh38
rs752015905 Chr.8: 103420997-103420997 on Build GRCh38
rs866732236 Chr.8: 103421030-103421031 on Build GRCh38
rs752299153 Chr.8: 103420964-103420964 on Build GRCh38
rs775306856 Chr.8: 103421025-103421025 on Build GRCh38
rs866596670 Chr.8: 103421247-103421247 on Build GRCh38
rs376638763 Chr.8: 103421056-103421056 on Build GRCh38
rs781549097 Chr.8: 103421000-103421000 on Build GRCh38
rs371332126 Chr.8: 103421117-103421117 on Build GRCh38
rs757256806 Chr.8: 103421149-103421151 on Build GRCh38
rs749435725 Chr.8: 103421077-103421077 on Build GRCh38
rs1022900631 Chr.8: 103421259-103421259 on Build GRCh38
rs375740775 Chr.8: 103420955-103420955 on Build GRCh38
rs374390026 Chr.8: 103421057-103421057 on Build GRCh38
rs1011747605 Chr.8: 103421081-103421081 on Build GRCh38
rs183706608 Chr.8: 103421031-103421031 on Build GRCh38
rs868582110 Chr.8: 103421031-103421032 on Build GRCh38
rs763709416 Chr.8: 103421033-103421033 on Build GRCh38
rs757289460 Chr.8: 103421092-103421092 on Build GRCh38
rs372518680 Chr.8: 103421012-103421012 on Build GRCh38
rs919999085 Chr.8: 103421283-103421283 on Build GRCh38
rs963506314 Chr.8: 103421266-103421266 on Build GRCh38
rs758946959 Chr.8: 103420988-103420988 on Build GRCh38
rs768357734 Chr.8: 103421119-103421119 on Build GRCh38
rs767286775 Chr.8: 103420961-103420961 on Build GRCh38
rs954159923 Chr.8: 103421032-103421032 on Build GRCh38
rs775308251 Chr.8: 103421110-103421110 on Build GRCh38
rs757452235 Chr.8: 103421316-103421316 on Build GRCh38
rs61744470 Chr.8: 103421072-103421072 on Build GRCh38
rs201585324 Chr.8: 103421065-103421065 on Build GRCh38
rs755309968 Chr.8: 103420999-103420999 on Build GRCh38
rs748408774 Chr.8: 103421005-103421005 on Build GRCh38
rs973271433 Chr.8: 103421268-103421268 on Build GRCh38
rs72675463 Chr.8: 103421320-103421320 on Build GRCh38
rs991373931 Chr.8: 103421188-103421188 on Build GRCh38
rs770091405 Chr.8: 103421010-103421010 on Build GRCh38

View Full Product Details