Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169822928-169823527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf112
Entrez Gene ID
55732
Gene Name
chromosome 1 open reading frame 112
Gene Aliases
-
UniGene
Hs.443551
Gene Chromosome Location
Chr.1: 169794730-169854080 on Build GRCh38
Transcripts
NM_001320047.1 NM_001320048.1 NM_018186.3 XM_005245317.4 XM_011509735.2 XM_017001722.1 XM_017001723.1 XM_017001724.1 XM_017001725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5556475 c.846C>T p.H282H Chr.1: 169823470-169823470 on Build GRCh38
4518766 c.793_794CC>TT p.P265F Chr.1: 169823417-169823418 on Build GRCh38
7166036 c.1009C>T p.L337F Chr.1: 169823459-169823459 on Build GRCh38
7166035 c.835C>T p.L279F Chr.1: 169823459-169823459 on Build GRCh38
458412 c.840G>C p.Q280H Chr.1: 169823464-169823464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778518478 Chr.1: 169823433-169823433 on Build GRCh38
rs1014551804 Chr.1: 169822981-169822981 on Build GRCh38
rs962175733 Chr.1: 169823461-169823461 on Build GRCh38
rs965674915 Chr.1: 169823095-169823095 on Build GRCh38
rs1044303676 Chr.1: 169823504-169823504 on Build GRCh38
rs952002745 Chr.1: 169823406-169823406 on Build GRCh38
rs191333858 Chr.1: 169822994-169822994 on Build GRCh38
rs921831888 Chr.1: 169823127-169823127 on Build GRCh38
rs888556131 Chr.1: 169822959-169822959 on Build GRCh38
rs769967534 Chr.1: 169823108-169823108 on Build GRCh38
rs544784114 Chr.1: 169823242-169823242 on Build GRCh38
rs907736795 Chr.1: 169823451-169823451 on Build GRCh38
rs758865741 Chr.1: 169823270-169823273 on Build GRCh38
rs116550168 Chr.1: 169823325-169823325 on Build GRCh38
rs12121443 Chr.1: 169823501-169823501 on Build GRCh38
rs895479917 Chr.1: 169823497-169823497 on Build GRCh38
rs770970622 Chr.1: 169823471-169823471 on Build GRCh38
rs183195626 Chr.1: 169822996-169822996 on Build GRCh38
rs984652689 Chr.1: 169823172-169823172 on Build GRCh38
rs550447399 Chr.1: 169823452-169823452 on Build GRCh38
rs1044394736 Chr.1: 169823297-169823297 on Build GRCh38
rs773143775 Chr.1: 169823442-169823442 on Build GRCh38
rs77004647 Chr.1: 169823322-169823322 on Build GRCh38
rs1000387204 Chr.1: 169823001-169823001 on Build GRCh38
rs760674324 Chr.1: 169823454-169823454 on Build GRCh38
rs1037038762 Chr.1: 169822977-169822977 on Build GRCh38
rs377362176 Chr.1: 169823487-169823487 on Build GRCh38
rs527767023 Chr.1: 169823251-169823255 on Build GRCh38
rs186199419 Chr.1: 169823383-169823383 on Build GRCh38
rs1018635829 Chr.1: 169823366-169823366 on Build GRCh38
rs143298868 Chr.1: 169823156-169823156 on Build GRCh38
rs965105462 Chr.1: 169823313-169823313 on Build GRCh38
rs773493943 Chr.1: 169823122-169823122 on Build GRCh38
rs1012834168 Chr.1: 169823031-169823031 on Build GRCh38
rs902919469 Chr.1: 169822973-169822973 on Build GRCh38
rs891955703 Chr.1: 169823342-169823342 on Build GRCh38
rs895818541 Chr.1: 169822978-169822978 on Build GRCh38
rs201156524 Chr.1: 169823382-169823382 on Build GRCh38
rs199939162 Chr.1: 169823260-169823264 on Build GRCh38
rs750009902 Chr.1: 169823367-169823368 on Build GRCh38
rs1054632562 Chr.1: 169823338-169823338 on Build GRCh38
rs763390194 Chr.1: 169823502-169823502 on Build GRCh38
rs995054755 Chr.1: 169823179-169823179 on Build GRCh38
rs765757116 Chr.1: 169823411-169823414 on Build GRCh38
rs1033567799 Chr.1: 169823009-169823009 on Build GRCh38
rs372665901 Chr.1: 169823423-169823423 on Build GRCh38
rs370766365 Chr.1: 169823492-169823492 on Build GRCh38
rs778429524 Chr.1: 169823364-169823364 on Build GRCh38
rs985111902 Chr.1: 169823418-169823418 on Build GRCh38
rs1010360589 Chr.1: 169823054-169823054 on Build GRCh38
rs1018951890 Chr.1: 169823204-169823204 on Build GRCh38
rs10919251 Chr.1: 169823190-169823190 on Build GRCh38
rs200701654 Chr.1: 169823458-169823458 on Build GRCh38
rs377477737 Chr.1: 169823264-169823264 on Build GRCh38
rs748002417 Chr.1: 169823436-169823436 on Build GRCh38
rs934371784 Chr.1: 169823473-169823473 on Build GRCh38
rs940069000 Chr.1: 169822961-169822961 on Build GRCh38
rs762462964 Chr.1: 169823387-169823390 on Build GRCh38
rs950013567 Chr.1: 169823259-169823259 on Build GRCh38
rs1010021472 Chr.1: 169823350-169823350 on Build GRCh38
rs987556567 Chr.1: 169823203-169823203 on Build GRCh38
rs368821634 Chr.1: 169823166-169823166 on Build GRCh38
rs144662128 Chr.1: 169823441-169823441 on Build GRCh38
rs760900160 Chr.1: 169823249-169823267 on Build GRCh38
rs907663554 Chr.1: 169823186-169823186 on Build GRCh38
rs149158064 Chr.1: 169823049-169823049 on Build GRCh38
rs779775818 Chr.1: 169823401-169823401 on Build GRCh38
rs965704841 Chr.1: 169823378-169823378 on Build GRCh38
rs917285814 Chr.1: 169823229-169823229 on Build GRCh38
rs560241898 Chr.1: 169823284-169823284 on Build GRCh38
rs934427018 Chr.1: 169822987-169822987 on Build GRCh38
rs768403537 Chr.1: 169823430-169823430 on Build GRCh38
rs765527309 Chr.1: 169823494-169823494 on Build GRCh38
rs946901834 Chr.1: 169822958-169822959 on Build GRCh38
rs915501279 Chr.1: 169823356-169823356 on Build GRCh38
rs186063887 Chr.1: 169822956-169822956 on Build GRCh38
rs1037112867 Chr.1: 169823197-169823197 on Build GRCh38
rs749896630 Chr.1: 169823376-169823376 on Build GRCh38
rs1018604711 Chr.1: 169823059-169823059 on Build GRCh38
rs754047197 Chr.1: 169823259-169823260 on Build GRCh38

View Full Product Details