Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202943595-202944184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR1
Entrez Gene ID
51094
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
UniGene
Hs.5298
Gene Chromosome Location
Chr.1: 202940825-202958572 on Build GRCh38
Transcripts
NM_001290553.1 NM_001290557.1 NM_001290629.1 NM_015999.5 XM_017001407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5907815 c.680C>T p.S227F Chr.1: 202943883-202943883 on Build GRCh38
376985 c.751A>T p.I251F Chr.1: 202943812-202943812 on Build GRCh38
902406 c.758C>T p.A253V Chr.1: 202943805-202943805 on Build GRCh38
6124175 c.759G>A p.A253A Chr.1: 202943804-202943804 on Build GRCh38
5379253 c.703C>T p.R235W Chr.1: 202943860-202943860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530759857 Chr.1: 202944089-202944089 on Build GRCh38
rs200326086 Chr.1: 202943893-202943893 on Build GRCh38
rs3737884 Chr.1: 202944076-202944076 on Build GRCh38
rs781168088 Chr.1: 202943644-202943644 on Build GRCh38
rs766952010 Chr.1: 202943750-202943750 on Build GRCh38
rs759055002 Chr.1: 202943971-202943971 on Build GRCh38
rs35418566 Chr.1: 202944019-202944019 on Build GRCh38
rs769555479 Chr.1: 202943703-202943703 on Build GRCh38
rs978390686 Chr.1: 202943706-202943706 on Build GRCh38
rs775398892 Chr.1: 202943713-202943713 on Build GRCh38
rs1032656024 Chr.1: 202944120-202944120 on Build GRCh38
rs764912508 Chr.1: 202943773-202943773 on Build GRCh38
rs1022597134 Chr.1: 202943998-202943998 on Build GRCh38
rs1013085422 Chr.1: 202943997-202943997 on Build GRCh38
rs1020193174 Chr.1: 202944061-202944061 on Build GRCh38
rs1002040474 Chr.1: 202944005-202944005 on Build GRCh38
rs754563633 Chr.1: 202943771-202943771 on Build GRCh38
rs774498455 Chr.1: 202943865-202943865 on Build GRCh38
rs1034427688 Chr.1: 202944016-202944016 on Build GRCh38
rs544597076 Chr.1: 202943690-202943690 on Build GRCh38
rs757117798 Chr.1: 202943839-202943839 on Build GRCh38
rs376579283 Chr.1: 202943731-202943731 on Build GRCh38
rs768820014 Chr.1: 202943726-202943726 on Build GRCh38
rs372980347 Chr.1: 202943909-202943909 on Build GRCh38
rs752321453 Chr.1: 202943983-202943983 on Build GRCh38
rs368393127 Chr.1: 202944090-202944090 on Build GRCh38
rs748704262 Chr.1: 202944000-202944000 on Build GRCh38
rs867974443 Chr.1: 202943850-202943850 on Build GRCh38
rs528315216 Chr.1: 202944046-202944046 on Build GRCh38
rs772408783 Chr.1: 202943920-202943920 on Build GRCh38
rs903378532 Chr.1: 202943831-202943831 on Build GRCh38
rs114193146 Chr.1: 202944158-202944158 on Build GRCh38
rs761085928 Chr.1: 202943743-202943743 on Build GRCh38
rs372051219 Chr.1: 202943711-202943711 on Build GRCh38
rs563431428 Chr.1: 202944060-202944060 on Build GRCh38
rs780762393 Chr.1: 202943721-202943721 on Build GRCh38
rs764822494 Chr.1: 202943982-202943982 on Build GRCh38
rs753379357 Chr.1: 202943754-202943754 on Build GRCh38
rs75992419 Chr.1: 202943872-202943872 on Build GRCh38
rs540762954 Chr.1: 202943840-202943840 on Build GRCh38
rs773421059 Chr.1: 202943930-202943930 on Build GRCh38
rs893391988 Chr.1: 202943956-202943956 on Build GRCh38
rs767786384 Chr.1: 202943739-202943739 on Build GRCh38
rs560545929 Chr.1: 202943964-202943964 on Build GRCh38
rs563962655 Chr.1: 202944114-202944114 on Build GRCh38
rs550842311 Chr.1: 202944103-202944103 on Build GRCh38
rs913131902 Chr.1: 202943678-202943678 on Build GRCh38
rs768728075 Chr.1: 202943852-202943852 on Build GRCh38
rs925589365 Chr.1: 202943729-202943729 on Build GRCh38
rs369530077 Chr.1: 202943811-202943811 on Build GRCh38
rs773687498 Chr.1: 202943740-202943740 on Build GRCh38
rs974317527 Chr.1: 202944149-202944149 on Build GRCh38
rs762324992 Chr.1: 202944098-202944098 on Build GRCh38
rs944828835 Chr.1: 202943793-202943793 on Build GRCh38
rs749415989 Chr.1: 202943714-202943714 on Build GRCh38
rs752407216 Chr.1: 202943782-202943782 on Build GRCh38
rs74411226 Chr.1: 202943867-202943867 on Build GRCh38
rs143634072 Chr.1: 202943805-202943805 on Build GRCh38
rs145759019 Chr.1: 202943804-202943804 on Build GRCh38
rs761152960 Chr.1: 202943947-202943947 on Build GRCh38
rs963652215 Chr.1: 202944136-202944136 on Build GRCh38
rs749328304 Chr.1: 202943843-202943843 on Build GRCh38

View Full Product Details