Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27196940-27197487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD5
Entrez Gene ID
91452
Gene Name
acyl-CoA binding domain containing 5
Gene Aliases
-
UniGene
Hs.530597
Gene Chromosome Location
Chr.10: 27182624-27248674 on Build GRCh38
Transcripts
NM_001042473.3 NM_001271512.3 NM_001301251.1 NM_001301252.1 NM_001301253.1 NM_001301254.1 NM_145698.4 XM_006717528.3 XM_006717530.3 XM_006717531.2 XM_006717535.2 XM_017016884.1 XM_017016885.1 XM_017016886.1 XM_017016887.1 XM_017016888.1 XM_017016889.1 XM_017016890.1 XM_017016891.1 XM_017016892.1 XM_017016893.1 XM_017016894.1 XM_017016895.1 XM_017016896.1 XM_017016897.1 XM_017016898.1 XM_017016899.1 XM_017016900.1 XM_017016901.1 XM_017016902.1 XM_017016903.1 XM_017016904.1 XM_017016905.1 XM_017016906.1 XM_017016907.1 XM_017016908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848412 c.1602C>A p.N534K Chr.10: 27197433-27197433 on Build GRCh38
4920862 c.1462A>T p.K488* Chr.10: 27197441-27197441 on Build GRCh38
427433 c.1577G>C p.*526S Chr.10: 27197431-27197431 on Build GRCh38
427434 c.1472G>C p.*491S Chr.10: 27197431-27197431 on Build GRCh38
4920863 c.1594A>T p.K532* Chr.10: 27197441-27197441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755088288 Chr.10: 27197260-27197260 on Build GRCh38
rs766296599 Chr.10: 27197377-27197377 on Build GRCh38
rs781448515 Chr.10: 27197401-27197401 on Build GRCh38
rs566733356 Chr.10: 27197288-27197288 on Build GRCh38
rs780967522 Chr.10: 27197291-27197291 on Build GRCh38
rs34694483 Chr.10: 27197233-27197234 on Build GRCh38
rs754129843 Chr.10: 27197094-27197094 on Build GRCh38
rs758991531 Chr.10: 27197016-27197019 on Build GRCh38
rs547264197 Chr.10: 27197250-27197250 on Build GRCh38
rs764986408 Chr.10: 27197362-27197362 on Build GRCh38
rs756991490 Chr.10: 27197097-27197097 on Build GRCh38
rs764555246 Chr.10: 27197050-27197050 on Build GRCh38
rs750270419 Chr.10: 27197373-27197373 on Build GRCh38
rs765656824 Chr.10: 27197157-27197157 on Build GRCh38
rs770242818 Chr.10: 27197280-27197280 on Build GRCh38
rs763676485 Chr.10: 27197345-27197345 on Build GRCh38
rs35454569 Chr.10: 27197354-27197355 on Build GRCh38
rs948353396 Chr.10: 27197333-27197333 on Build GRCh38
rs575460374 Chr.10: 27197273-27197273 on Build GRCh38
rs983630209 Chr.10: 27197051-27197051 on Build GRCh38
rs937700286 Chr.10: 27197123-27197123 on Build GRCh38
rs771396498 Chr.10: 27197307-27197307 on Build GRCh38
rs12356269 Chr.10: 27196972-27196972 on Build GRCh38
rs990255042 Chr.10: 27196966-27196966 on Build GRCh38
rs11420737 Chr.10: 27197349-27197350 on Build GRCh38
rs763722858 Chr.10: 27196979-27196979 on Build GRCh38
rs979185553 Chr.10: 27197054-27197054 on Build GRCh38
rs762440079 Chr.10: 27197140-27197140 on Build GRCh38
rs1045818667 Chr.10: 27197335-27197335 on Build GRCh38
rs754960623 Chr.10: 27197382-27197382 on Build GRCh38
rs376326619 Chr.10: 27197429-27197429 on Build GRCh38
rs780161581 Chr.10: 27197214-27197214 on Build GRCh38
rs530375004 Chr.10: 27197222-27197222 on Build GRCh38
rs763577436 Chr.10: 27197048-27197049 on Build GRCh38
rs758345992 Chr.10: 27197375-27197375 on Build GRCh38
rs778268016 Chr.10: 27197420-27197420 on Build GRCh38
rs776220825 Chr.10: 27197346-27197346 on Build GRCh38
rs924986979 Chr.10: 27197064-27197064 on Build GRCh38
rs976304116 Chr.10: 27197016-27197020 on Build GRCh38
rs760039877 Chr.10: 27197340-27197340 on Build GRCh38
rs775058750 Chr.10: 27197331-27197331 on Build GRCh38
rs779232071 Chr.10: 27197434-27197434 on Build GRCh38
rs751525362 Chr.10: 27197379-27197379 on Build GRCh38
rs748345249 Chr.10: 27197405-27197405 on Build GRCh38
rs781505581 Chr.10: 27197261-27197261 on Build GRCh38
rs142191359 Chr.10: 27197049-27197050 on Build GRCh38
rs561705792 Chr.10: 27197172-27197172 on Build GRCh38
rs765043505 Chr.10: 27197142-27197142 on Build GRCh38
rs907112175 Chr.10: 27197391-27197391 on Build GRCh38
rs941102202 Chr.10: 27197263-27197263 on Build GRCh38
rs761489496 Chr.10: 27197351-27197351 on Build GRCh38
rs976098668 Chr.10: 27197006-27197006 on Build GRCh38
rs747118256 Chr.10: 27197221-27197221 on Build GRCh38
rs753496558 Chr.10: 27197011-27197011 on Build GRCh38
rs966680513 Chr.10: 27197008-27197008 on Build GRCh38
rs182263760 Chr.10: 27196978-27196978 on Build GRCh38
rs758308677 Chr.10: 27197191-27197191 on Build GRCh38
rs543344287 Chr.10: 27197336-27197337 on Build GRCh38
rs756452915 Chr.10: 27197412-27197412 on Build GRCh38
rs896426857 Chr.10: 27197236-27197236 on Build GRCh38

View Full Product Details