Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101600497-101601043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPCD
Entrez Gene ID
25911
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
Gene Aliases
-
UniGene
Hs.658128
Gene Chromosome Location
Chr.10: 101588332-101609653 on Build GRCh38
Transcripts
NM_015448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
914049 c.157C>T p.R53C Chr.10: 101600749-101600749 on Build GRCh38
3665449 c.211C>G p.P71A Chr.10: 101600803-101600803 on Build GRCh38
5571784 c.204A>G p.V68V Chr.10: 101600796-101600796 on Build GRCh38
5480758 c.166A>C p.S56R Chr.10: 101600758-101600758 on Build GRCh38
2058577 c.211C>T p.P71S Chr.10: 101600803-101600803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951232524 Chr.10: 101600581-101600581 on Build GRCh38
rs369533298 Chr.10: 101600836-101600836 on Build GRCh38
rs974491906 Chr.10: 101600749-101600749 on Build GRCh38
rs201411239 Chr.10: 101600815-101600815 on Build GRCh38
rs1011956971 Chr.10: 101600826-101600826 on Build GRCh38
rs146176614 Chr.10: 101600532-101600532 on Build GRCh38
rs201479943 Chr.10: 101600832-101600832 on Build GRCh38
rs756946398 Chr.10: 101600696-101600696 on Build GRCh38
rs562786902 Chr.10: 101600566-101600566 on Build GRCh38
rs779173242 Chr.10: 101600739-101600739 on Build GRCh38
rs770257580 Chr.10: 101600818-101600818 on Build GRCh38
rs763649960 Chr.10: 101600690-101600690 on Build GRCh38
rs377516854 Chr.10: 101600891-101600891 on Build GRCh38
rs945924035 Chr.10: 101600831-101600831 on Build GRCh38
rs753652551 Chr.10: 101600857-101600857 on Build GRCh38
rs777917445 Chr.10: 101600969-101600969 on Build GRCh38
rs1001588288 Chr.10: 101600589-101600589 on Build GRCh38
rs202146435 Chr.10: 101600697-101600697 on Build GRCh38
rs143867323 Chr.10: 101600803-101600803 on Build GRCh38
rs983936118 Chr.10: 101600612-101600612 on Build GRCh38
rs752829504 Chr.10: 101600867-101600867 on Build GRCh38
rs1047382012 Chr.10: 101600935-101600935 on Build GRCh38
rs374926340 Chr.10: 101600772-101600772 on Build GRCh38
rs547421783 Chr.10: 101600661-101600661 on Build GRCh38
rs1045827456 Chr.10: 101600569-101600569 on Build GRCh38
rs560293997 Chr.10: 101600651-101600651 on Build GRCh38
rs192405634 Chr.10: 101600877-101600877 on Build GRCh38
rs779015795 Chr.10: 101600862-101600862 on Build GRCh38
rs188708077 Chr.10: 101600650-101600650 on Build GRCh38
rs149463737 Chr.10: 101600750-101600750 on Build GRCh38
rs371077200 Chr.10: 101600902-101600902 on Build GRCh38
rs776963985 Chr.10: 101600808-101600808 on Build GRCh38
rs754942624 Chr.10: 101600719-101600719 on Build GRCh38
rs532382975 Chr.10: 101600807-101600807 on Build GRCh38
rs750048160 Chr.10: 101600705-101600705 on Build GRCh38
rs964222761 Chr.10: 101601017-101601017 on Build GRCh38
rs748269466 Chr.10: 101600745-101600745 on Build GRCh38
rs755874537 Chr.10: 101600709-101600709 on Build GRCh38
rs148547618 Chr.10: 101600961-101600961 on Build GRCh38
rs375865718 Chr.10: 101600866-101600866 on Build GRCh38
rs529988649 Chr.10: 101600626-101600626 on Build GRCh38
rs576911865 Chr.10: 101600530-101600530 on Build GRCh38
rs775939781 Chr.10: 101600820-101600820 on Build GRCh38
rs751048649 Chr.10: 101600694-101600694 on Build GRCh38
rs1025589376 Chr.10: 101600962-101600962 on Build GRCh38
rs773316550 Chr.10: 101600759-101600759 on Build GRCh38
rs913143638 Chr.10: 101600811-101600811 on Build GRCh38
rs971051539 Chr.10: 101600522-101600522 on Build GRCh38
rs771159961 Chr.10: 101600888-101600888 on Build GRCh38
rs570528685 Chr.10: 101600698-101600716 on Build GRCh38
rs146371249 Chr.10: 101600834-101600834 on Build GRCh38
rs935531682 Chr.10: 101600796-101600796 on Build GRCh38
rs749306413 Chr.10: 101600717-101600717 on Build GRCh38
rs951263139 Chr.10: 101600974-101600974 on Build GRCh38
rs754020616 Chr.10: 101600591-101600591 on Build GRCh38
rs372782769 Chr.10: 101600865-101600865 on Build GRCh38
rs754778307 Chr.10: 101600858-101600858 on Build GRCh38
rs761255778 Chr.10: 101600829-101600829 on Build GRCh38
rs770099808 Chr.10: 101600892-101600892 on Build GRCh38
rs548539421 Chr.10: 101600872-101600872 on Build GRCh38
rs777901354 Chr.10: 101600870-101600870 on Build GRCh38
rs766147472 Chr.10: 101600846-101600846 on Build GRCh38
rs199897145 Chr.10: 101600716-101600716 on Build GRCh38
rs983975335 Chr.10: 101601008-101601008 on Build GRCh38

View Full Product Details