Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96059200-96059729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJ
Entrez Gene ID
54619
Gene Name
cyclin J
Gene Aliases
bA690P14.1
UniGene
Hs.596479
Gene Chromosome Location
Chr.10: 96042848-96060870 on Build GRCh38
Transcripts
NM_001134375.1 NM_001134376.1 NM_019084.4 XM_005269921.3 XM_011539885.2 XM_017016354.1 XM_017016355.1 XM_017016356.1 NR_038444.1 NR_134320.1 NR_134321.1 NR_134322.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001134375.1 NM_001134376.1 NM_019084.4 XM_005269921.3 XM_011539885.2 XM_017016354.1 XM_017016355.1 XM_017016356.1 NR_038444.1 NR_134320.1 NR_134321.1 NR_134322.1

SNP Detail

SNP ID Chromosome Location
rs555377299 Chr.10: 96059547-96059547 on Build GRCh38
rs117301955 Chr.10: 96059705-96059705 on Build GRCh38
rs933612681 Chr.10: 96059411-96059411 on Build GRCh38
rs540255295 Chr.10: 96059697-96059697 on Build GRCh38
rs780216361 Chr.10: 96059703-96059704 on Build GRCh38
rs575099219 Chr.10: 96059554-96059554 on Build GRCh38
rs546464401 Chr.10: 96059706-96059706 on Build GRCh38
rs182197918 Chr.10: 96059453-96059453 on Build GRCh38
rs12573468 Chr.10: 96059590-96059590 on Build GRCh38
rs942951479 Chr.10: 96059345-96059345 on Build GRCh38
rs967895297 Chr.10: 96059655-96059656 on Build GRCh38
rs748120936 Chr.10: 96059665-96059665 on Build GRCh38
rs752949529 Chr.10: 96059532-96059532 on Build GRCh38
rs1046725687 Chr.10: 96059444-96059444 on Build GRCh38
rs1034864230 Chr.10: 96059555-96059555 on Build GRCh38
rs1043518 Chr.10: 96059511-96059511 on Build GRCh38
rs189994619 Chr.10: 96059301-96059301 on Build GRCh38
rs1043280283 Chr.10: 96059319-96059319 on Build GRCh38
rs1016749520 Chr.10: 96059487-96059487 on Build GRCh38
rs765957957 Chr.10: 96059357-96059360 on Build GRCh38
rs913267200 Chr.10: 96059273-96059273 on Build GRCh38
rs778781734 Chr.10: 96059594-96059595 on Build GRCh38
rs201266607 Chr.10: 96059589-96059589 on Build GRCh38
rs570317759 Chr.10: 96059266-96059266 on Build GRCh38
rs772297867 Chr.10: 96059589-96059590 on Build GRCh38
rs926096043 Chr.10: 96059340-96059340 on Build GRCh38
rs376778882 Chr.10: 96059375-96059375 on Build GRCh38
rs758655578 Chr.10: 96059548-96059548 on Build GRCh38
rs1020274810 Chr.10: 96059642-96059642 on Build GRCh38
rs202177702 Chr.10: 96059593-96059594 on Build GRCh38
rs56951868 Chr.10: 96059512-96059512 on Build GRCh38
rs926123520 Chr.10: 96059677-96059677 on Build GRCh38
rs960068777 Chr.10: 96059581-96059581 on Build GRCh38
rs537749922 Chr.10: 96059690-96059690 on Build GRCh38
rs960283393 Chr.10: 96059247-96059247 on Build GRCh38
rs3841417 Chr.10: 96059594-96059594 on Build GRCh38
rs546380227 Chr.10: 96059399-96059399 on Build GRCh38
rs200852141 Chr.10: 96059590-96059590 on Build GRCh38
rs780179038 Chr.10: 96059667-96059667 on Build GRCh38
rs942812932 Chr.10: 96059696-96059696 on Build GRCh38
rs199958778 Chr.10: 96059594-96059594 on Build GRCh38
rs144230436 Chr.10: 96059588-96059589 on Build GRCh38
rs946148762 Chr.10: 96059296-96059296 on Build GRCh38
rs927314188 Chr.10: 96059235-96059235 on Build GRCh38
rs1005291650 Chr.10: 96059480-96059480 on Build GRCh38

View Full Product Details